Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 152575057-152575568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMNL2
Entrez Gene ID
114793
Gene Name
formin like 2
Gene Aliases
FHOD2
UniGene
Hs.654630
Gene Chromosome Location
Chr.2: 152335170-152649834 on Build GRCh38
Transcripts
NM_052905.3 XM_005246263.2 XM_005246265.3 XM_011510530.1 XM_011510531.1 XM_011510532.2 XM_011510533.2 XM_011510534.1 XM_011510535.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3567984 c.645G>T p.V215V Chr.2: 152575184-152575184 on Build GRCh38
4776795 c.650A>C p.K217T Chr.2: 152575189-152575189 on Build GRCh38
5660267 c.658G>T p.D220Y Chr.2: 152575197-152575197 on Build GRCh38
5744740 c.685C>T p.R229C Chr.2: 152575224-152575224 on Build GRCh38
3770912 c.611C>T p.P204L Chr.2: 152575150-152575150 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202220302 Chr.2: 152575342-152575342 on Build GRCh38
rs889227102 Chr.2: 152575374-152575374 on Build GRCh38
rs56101947 Chr.2: 152575451-152575452 on Build GRCh38
rs768173003 Chr.2: 152575120-152575120 on Build GRCh38
rs149316753 Chr.2: 152575264-152575264 on Build GRCh38
rs759990144 Chr.2: 152575241-152575241 on Build GRCh38
rs201093394 Chr.2: 152575169-152575169 on Build GRCh38
rs776889088 Chr.2: 152575394-152575394 on Build GRCh38
rs756422064 Chr.2: 152575270-152575270 on Build GRCh38
rs747653250 Chr.2: 152575132-152575132 on Build GRCh38
rs781047719 Chr.2: 152575440-152575440 on Build GRCh38
rs769226788 Chr.2: 152575149-152575149 on Build GRCh38
rs558408286 Chr.2: 152575518-152575518 on Build GRCh38
rs1051459399 Chr.2: 152575322-152575322 on Build GRCh38
rs763399880 Chr.2: 152575196-152575196 on Build GRCh38
rs1049658007 Chr.2: 152575139-152575139 on Build GRCh38
rs892469771 Chr.2: 152575418-152575418 on Build GRCh38
rs1033029059 Chr.2: 152575412-152575412 on Build GRCh38
rs766397126 Chr.2: 152575368-152575368 on Build GRCh38
rs966028696 Chr.2: 152575441-152575441 on Build GRCh38
rs543575629 Chr.2: 152575530-152575530 on Build GRCh38
rs970803275 Chr.2: 152575485-152575485 on Build GRCh38
rs546535067 Chr.2: 152575099-152575099 on Build GRCh38
rs758578386 Chr.2: 152575155-152575157 on Build GRCh38
rs980636541 Chr.2: 152575181-152575181 on Build GRCh38
rs900052140 Chr.2: 152575373-152575373 on Build GRCh38
rs148451777 Chr.2: 152575504-152575504 on Build GRCh38
rs780272257 Chr.2: 152575270-152575270 on Build GRCh38
rs774546089 Chr.2: 152575229-152575229 on Build GRCh38
rs538415036 Chr.2: 152575279-152575279 on Build GRCh38
rs369372045 Chr.2: 152575349-152575349 on Build GRCh38
rs755064756 Chr.2: 152575461-152575462 on Build GRCh38
rs5835437 Chr.2: 152575451-152575451 on Build GRCh38
rs397742202 Chr.2: 152575460-152575461 on Build GRCh38
rs746863647 Chr.2: 152575112-152575112 on Build GRCh38
rs377750407 Chr.2: 152575281-152575281 on Build GRCh38
rs375873175 Chr.2: 152575355-152575355 on Build GRCh38
rs926434810 Chr.2: 152575146-152575146 on Build GRCh38
rs398090762 Chr.2: 152575461-152575461 on Build GRCh38
rs926494626 Chr.2: 152575266-152575266 on Build GRCh38
rs778009238 Chr.2: 152575094-152575094 on Build GRCh38
rs866238007 Chr.2: 152575114-152575114 on Build GRCh38
rs750731843 Chr.2: 152575101-152575103 on Build GRCh38
rs747211392 Chr.2: 152575269-152575270 on Build GRCh38
rs1025029197 Chr.2: 152575461-152575461 on Build GRCh38
rs562524270 Chr.2: 152575476-152575477 on Build GRCh38
rs776403363 Chr.2: 152575128-152575128 on Build GRCh38
rs568510802 Chr.2: 152575327-152575327 on Build GRCh38
rs201743391 Chr.2: 152575224-152575224 on Build GRCh38
rs544934941 Chr.2: 152575505-152575505 on Build GRCh38
rs752904116 Chr.2: 152575269-152575269 on Build GRCh38
rs1009604477 Chr.2: 152575426-152575426 on Build GRCh38
rs926774264 Chr.2: 152575481-152575481 on Build GRCh38
rs532903667 Chr.2: 152575111-152575111 on Build GRCh38
rs564346698 Chr.2: 152575452-152575452 on Build GRCh38
rs866589667 Chr.2: 152575318-152575318 on Build GRCh38
rs553794137 Chr.2: 152575371-152575371 on Build GRCh38
rs1007580167 Chr.2: 152575407-152575407 on Build GRCh38
rs201061768 Chr.2: 152575101-152575101 on Build GRCh38
rs929000233 Chr.2: 152575303-152575303 on Build GRCh38
rs190078985 Chr.2: 152575293-152575293 on Build GRCh38
rs372666879 Chr.2: 152575332-152575332 on Build GRCh38
rs73969130 Chr.2: 152575531-152575531 on Build GRCh38
rs972438898 Chr.2: 152575449-152575449 on Build GRCh38
rs370835802 Chr.2: 152575460-152575460 on Build GRCh38

View Full Product Details