Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 197742979-197743483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOLL
Entrez Gene ID
66037
Gene Name
boule homolog, RNA binding protein
Gene Aliases
BOULE
UniGene
Hs.169797
Gene Chromosome Location
Chr.2: 197726879-197786574 on Build GRCh38
Transcripts
NM_001284358.1 NM_001284361.1 NM_001284362.1 NM_033030.5 NM_197970.2 XM_006712715.3 XM_011511692.2 XM_011511693.2 XM_011511694.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7249941 c.800C>A p.A267E Chr.2: 197743089-197743089 on Build GRCh38
7186370 c.773C>T p.S258F Chr.2: 197743152-197743152 on Build GRCh38
7249940 c.836C>A p.A279E Chr.2: 197743089-197743089 on Build GRCh38
4976665 c.821C>G p.A274G Chr.2: 197743104-197743104 on Build GRCh38
1014557 c.799C>A p.H267N Chr.2: 197743126-197743126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915094318 Chr.2: 197743319-197743319 on Build GRCh38
rs750750648 Chr.2: 197743038-197743038 on Build GRCh38
rs569962962 Chr.2: 197743396-197743396 on Build GRCh38
rs371047737 Chr.2: 197743130-197743130 on Build GRCh38
rs867709728 Chr.2: 197743141-197743141 on Build GRCh38
rs1026043980 Chr.2: 197743262-197743262 on Build GRCh38
rs77407369 Chr.2: 197743325-197743325 on Build GRCh38
rs375368975 Chr.2: 197743017-197743017 on Build GRCh38
rs768964464 Chr.2: 197743166-197743167 on Build GRCh38
rs375573791 Chr.2: 197743107-197743107 on Build GRCh38
rs1041352577 Chr.2: 197743033-197743033 on Build GRCh38
rs1005379820 Chr.2: 197743283-197743283 on Build GRCh38
rs986619330 Chr.2: 197743134-197743134 on Build GRCh38
rs756335563 Chr.2: 197743220-197743220 on Build GRCh38
rs762920202 Chr.2: 197743126-197743126 on Build GRCh38
rs757500135 Chr.2: 197743021-197743021 on Build GRCh38
rs1022595580 Chr.2: 197743412-197743412 on Build GRCh38
rs993226779 Chr.2: 197743260-197743260 on Build GRCh38
rs781664870 Chr.2: 197743031-197743031 on Build GRCh38
rs759218045 Chr.2: 197743096-197743096 on Build GRCh38
rs758525501 Chr.2: 197743010-197743010 on Build GRCh38
rs889408474 Chr.2: 197743192-197743192 on Build GRCh38
rs746514908 Chr.2: 197743200-197743200 on Build GRCh38
rs968202492 Chr.2: 197743461-197743461 on Build GRCh38
rs552302899 Chr.2: 197743186-197743186 on Build GRCh38
rs775325443 Chr.2: 197743124-197743124 on Build GRCh38
rs929907125 Chr.2: 197743233-197743233 on Build GRCh38
rs369412307 Chr.2: 197743085-197743085 on Build GRCh38
rs944671676 Chr.2: 197743172-197743172 on Build GRCh38
rs142061895 Chr.2: 197743076-197743076 on Build GRCh38
rs201315715 Chr.2: 197743454-197743454 on Build GRCh38
rs754347318 Chr.2: 197743190-197743190 on Build GRCh38
rs1046996518 Chr.2: 197743247-197743247 on Build GRCh38
rs946892434 Chr.2: 197743120-197743120 on Build GRCh38
rs771349777 Chr.2: 197743088-197743088 on Build GRCh38
rs77631688 Chr.2: 197743384-197743384 on Build GRCh38
rs1038285132 Chr.2: 197743223-197743223 on Build GRCh38
rs1005245375 Chr.2: 197743058-197743058 on Build GRCh38
rs1033562570 Chr.2: 197743297-197743297 on Build GRCh38
rs775621268 Chr.2: 197743028-197743031 on Build GRCh38
rs778742081 Chr.2: 197743456-197743456 on Build GRCh38
rs372015936 Chr.2: 197743089-197743089 on Build GRCh38
rs750850101 Chr.2: 197743164-197743164 on Build GRCh38
rs73990704 Chr.2: 197743049-197743049 on Build GRCh38
rs749393154 Chr.2: 197743301-197743301 on Build GRCh38
rs771602159 Chr.2: 197743069-197743069 on Build GRCh38
rs959558737 Chr.2: 197743306-197743306 on Build GRCh38
rs115905913 Chr.2: 197743352-197743352 on Build GRCh38
rs756457901 Chr.2: 197743168-197743168 on Build GRCh38
rs977487921 Chr.2: 197743345-197743345 on Build GRCh38
rs993922126 Chr.2: 197743041-197743041 on Build GRCh38
rs1023796460 Chr.2: 197743310-197743310 on Build GRCh38
rs368659245 Chr.2: 197743079-197743079 on Build GRCh38
rs1025451306 Chr.2: 197743045-197743045 on Build GRCh38
rs762194519 Chr.2: 197743154-197743154 on Build GRCh38
rs777001507 Chr.2: 197743178-197743179 on Build GRCh38
rs751679778 Chr.2: 197743153-197743153 on Build GRCh38
rs767604523 Chr.2: 197743160-197743160 on Build GRCh38
rs988376058 Chr.2: 197743407-197743407 on Build GRCh38
rs749746878 Chr.2: 197743064-197743064 on Build GRCh38
rs780404273 Chr.2: 197743055-197743055 on Build GRCh38
rs888260275 Chr.2: 197743251-197743251 on Build GRCh38
rs191927163 Chr.2: 197743248-197743248 on Build GRCh38
rs989721357 Chr.2: 197743084-197743084 on Build GRCh38
rs571303349 Chr.2: 197743328-197743328 on Build GRCh38
rs915401766 Chr.2: 197743113-197743113 on Build GRCh38
rs376712356 Chr.2: 197743196-197743196 on Build GRCh38
rs200225773 Chr.2: 197743016-197743016 on Build GRCh38
rs904947475 Chr.2: 197743312-197743312 on Build GRCh38
rs910672423 Chr.2: 197743151-197743151 on Build GRCh38
rs760863103 Chr.2: 197743037-197743037 on Build GRCh38
rs1040378544 Chr.2: 197743197-197743197 on Build GRCh38
rs755418185 Chr.2: 197743191-197743191 on Build GRCh38
rs867917983 Chr.2: 197743093-197743094 on Build GRCh38
rs777916226 Chr.2: 197743280-197743280 on Build GRCh38
rs987882553 Chr.2: 197743400-197743400 on Build GRCh38
rs1050531914 Chr.2: 197743009-197743009 on Build GRCh38
rs963102526 Chr.2: 197743014-197743014 on Build GRCh38
rs150043201 Chr.2: 197743353-197743353 on Build GRCh38
rs780425498 Chr.2: 197743175-197743175 on Build GRCh38

View Full Product Details