Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 197776972-197777479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOLL
Entrez Gene ID
66037
Gene Name
boule homolog, RNA binding protein
Gene Aliases
BOULE
UniGene
Hs.169797
Gene Chromosome Location
Chr.2: 197726879-197786574 on Build GRCh38
Transcripts
NM_001284358.1 NM_001284361.1 NM_001284362.1 NM_033030.5 NM_197970.2 XM_006712715.3 XM_011511692.2 XM_011511693.2 XM_011511694.2 XM_017004773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6313123 c.259C>A p.Q87K Chr.2: 197777076-197777076 on Build GRCh38
6313122 c.295C>A p.Q99K Chr.2: 197777076-197777076 on Build GRCh38
5623916 c.253G>A p.D85N Chr.2: 197777082-197777082 on Build GRCh38
5614997 c.271C>G p.Q91E Chr.2: 197777064-197777064 on Build GRCh38
4959572 c.258G>A p.G86G Chr.2: 197777113-197777113 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886153919 Chr.2: 197777445-197777445 on Build GRCh38
rs1043414359 Chr.2: 197777275-197777275 on Build GRCh38
rs187157061 Chr.2: 197777425-197777425 on Build GRCh38
rs540651021 Chr.2: 197777251-197777251 on Build GRCh38
rs1038601991 Chr.2: 197777323-197777323 on Build GRCh38
rs778337759 Chr.2: 197777091-197777091 on Build GRCh38
rs773029311 Chr.2: 197777159-197777159 on Build GRCh38
rs1029340977 Chr.2: 197777452-197777452 on Build GRCh38
rs759555811 Chr.2: 197777018-197777018 on Build GRCh38
rs192331363 Chr.2: 197777285-197777285 on Build GRCh38
rs545874348 Chr.2: 197777208-197777208 on Build GRCh38
rs778337369 Chr.2: 197777052-197777052 on Build GRCh38
rs929361771 Chr.2: 197777345-197777345 on Build GRCh38
rs143816061 Chr.2: 197777331-197777331 on Build GRCh38
rs375500928 Chr.2: 197777338-197777338 on Build GRCh38
rs546502449 Chr.2: 197777126-197777129 on Build GRCh38
rs563794945 Chr.2: 197777319-197777319 on Build GRCh38
rs111746936 Chr.2: 197777110-197777110 on Build GRCh38
rs913303236 Chr.2: 197777255-197777255 on Build GRCh38
rs1026718750 Chr.2: 197777001-197777001 on Build GRCh38
rs544014030 Chr.2: 197777315-197777315 on Build GRCh38
rs757834928 Chr.2: 197777078-197777078 on Build GRCh38
rs752969093 Chr.2: 197777030-197777030 on Build GRCh38
rs1003510927 Chr.2: 197777368-197777368 on Build GRCh38
rs115950330 Chr.2: 197777253-197777253 on Build GRCh38
rs746350560 Chr.2: 197777087-197777087 on Build GRCh38
rs553373974 Chr.2: 197777249-197777249 on Build GRCh38
rs751862100 Chr.2: 197777072-197777072 on Build GRCh38
rs561671635 Chr.2: 197777265-197777265 on Build GRCh38
rs1022502373 Chr.2: 197777224-197777224 on Build GRCh38
rs901795961 Chr.2: 197777375-197777375 on Build GRCh38
rs985107529 Chr.2: 197777311-197777311 on Build GRCh38
rs781686303 Chr.2: 197777385-197777385 on Build GRCh38
rs758630839 Chr.2: 197777045-197777045 on Build GRCh38
rs377444840 Chr.2: 197777252-197777252 on Build GRCh38
rs138416463 Chr.2: 197777080-197777080 on Build GRCh38
rs1047784280 Chr.2: 197777350-197777350 on Build GRCh38
rs906629369 Chr.2: 197777359-197777359 on Build GRCh38
rs895399106 Chr.2: 197777373-197777373 on Build GRCh38
rs947494080 Chr.2: 197777269-197777269 on Build GRCh38
rs1055130670 Chr.2: 197777372-197777372 on Build GRCh38
rs1015944774 Chr.2: 197777119-197777119 on Build GRCh38
rs529472378 Chr.2: 197777305-197777305 on Build GRCh38
rs989026578 Chr.2: 197777254-197777254 on Build GRCh38
rs183238709 Chr.2: 197777374-197777374 on Build GRCh38
rs970271511 Chr.2: 197777144-197777144 on Build GRCh38
rs768161071 Chr.2: 197777089-197777089 on Build GRCh38
rs933637348 Chr.2: 197777286-197777286 on Build GRCh38
rs906554146 Chr.2: 197777081-197777081 on Build GRCh38
rs959617605 Chr.2: 197777146-197777146 on Build GRCh38
rs560534719 Chr.2: 197777371-197777371 on Build GRCh38
rs765366371 Chr.2: 197777020-197777020 on Build GRCh38
rs1000122600 Chr.2: 197777436-197777436 on Build GRCh38
rs752840846 Chr.2: 197777145-197777145 on Build GRCh38
rs577424738 Chr.2: 197777103-197777103 on Build GRCh38

View Full Product Details