Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 199928638-199929134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TYW5
Entrez Gene ID
129450
Gene Name
tRNA-yW synthesizing protein 5
Gene Aliases
C2orf60, hTYW5
UniGene
Hs.204619
Gene Chromosome Location
Chr.2: 199928911-199955736 on Build GRCh38
Transcripts
NM_001039693.2 NR_004862.1 NR_109905.1 NR_109906.1 NR_109907.1

SNP Detail

SNP ID Chromosome Location
rs947646225 Chr.2: 199929091-199929091 on Build GRCh38
rs748518301 Chr.2: 199928821-199928821 on Build GRCh38
rs1021918704 Chr.2: 199928720-199928720 on Build GRCh38
rs1034608748 Chr.2: 199928975-199928975 on Build GRCh38
rs1026205320 Chr.2: 199928862-199928862 on Build GRCh38
rs971991149 Chr.2: 199928777-199928777 on Build GRCh38
rs377634250 Chr.2: 199928845-199928846 on Build GRCh38
rs1010504156 Chr.2: 199928702-199928702 on Build GRCh38
rs539823337 Chr.2: 199928897-199928897 on Build GRCh38
rs934854598 Chr.2: 199929036-199929036 on Build GRCh38
rs762470876 Chr.2: 199929058-199929058 on Build GRCh38
rs948825434 Chr.2: 199928946-199928946 on Build GRCh38
rs942387252 Chr.2: 199928836-199928839 on Build GRCh38
rs759570893 Chr.2: 199928684-199928684 on Build GRCh38
rs951963003 Chr.2: 199928894-199928894 on Build GRCh38
rs981974118 Chr.2: 199928966-199928966 on Build GRCh38
rs16832051 Chr.2: 199929061-199929061 on Build GRCh38
rs796396018 Chr.2: 199928987-199928989 on Build GRCh38
rs867984346 Chr.2: 199928770-199928770 on Build GRCh38
rs189347859 Chr.2: 199928677-199928677 on Build GRCh38
rs1018774954 Chr.2: 199928738-199928738 on Build GRCh38
rs1024788980 Chr.2: 199929006-199929006 on Build GRCh38
rs1007755226 Chr.2: 199928731-199928731 on Build GRCh38
rs888831731 Chr.2: 199928730-199928730 on Build GRCh38
rs550526681 Chr.2: 199929018-199929018 on Build GRCh38
rs966293218 Chr.2: 199928742-199928742 on Build GRCh38
rs977906142 Chr.2: 199929017-199929019 on Build GRCh38
rs1041163370 Chr.2: 199928841-199928844 on Build GRCh38
rs562657419 Chr.2: 199929043-199929043 on Build GRCh38
rs146625788 Chr.2: 199928683-199928685 on Build GRCh38
rs564872349 Chr.2: 199928934-199928934 on Build GRCh38
rs942336168 Chr.2: 199929071-199929071 on Build GRCh38
rs989138931 Chr.2: 199928810-199928810 on Build GRCh38
rs752260188 Chr.2: 199928985-199928989 on Build GRCh38
rs915923831 Chr.2: 199928944-199928944 on Build GRCh38
rs542664650 Chr.2: 199928667-199928667 on Build GRCh38
rs532241373 Chr.2: 199928982-199928982 on Build GRCh38
rs553646268 Chr.2: 199928786-199928786 on Build GRCh38
rs967482085 Chr.2: 199929007-199929007 on Build GRCh38
rs1047246842 Chr.2: 199928899-199928899 on Build GRCh38
rs765227046 Chr.2: 199928835-199928836 on Build GRCh38

View Full Product Details