Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 216414972-216415468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMARCAL1
Entrez Gene ID
50485
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a like 1
Gene Aliases
HARP, HHARP
UniGene
Hs.516674
Gene Chromosome Location
Chr.2: 216412414-216483053 on Build GRCh38
Transcripts
NM_001127207.1 NM_014140.3 XM_005246631.2 XM_005246632.1 XM_017004228.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1631834 c.335G>A p.S112N Chr.2: 216415039-216415039 on Build GRCh38
4091329 c.740G>A p.R247H Chr.2: 216415444-216415444 on Build GRCh38
7102468 c.405A>G p.K135K Chr.2: 216415109-216415109 on Build GRCh38
4091328 c.730A>G p.N244D Chr.2: 216415434-216415434 on Build GRCh38
6784941 c.645G>A p.T215T Chr.2: 216415349-216415349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922754263 Chr.2: 216415391-216415391 on Build GRCh38
rs11555797 Chr.2: 216415045-216415045 on Build GRCh38
rs748188404 Chr.2: 216415192-216415192 on Build GRCh38
rs756864375 Chr.2: 216415128-216415128 on Build GRCh38
rs906445943 Chr.2: 216415248-216415248 on Build GRCh38
rs947471355 Chr.2: 216415091-216415091 on Build GRCh38
rs778786012 Chr.2: 216415424-216415424 on Build GRCh38
rs151122737 Chr.2: 216415250-216415250 on Build GRCh38
rs376295895 Chr.2: 216415152-216415152 on Build GRCh38
rs751534116 Chr.2: 216415379-216415379 on Build GRCh38
rs749052447 Chr.2: 216415313-216415313 on Build GRCh38
rs774029345 Chr.2: 216415253-216415253 on Build GRCh38
rs749145673 Chr.2: 216415409-216415409 on Build GRCh38
rs201962302 Chr.2: 216415436-216415436 on Build GRCh38
rs760336986 Chr.2: 216415333-216415333 on Build GRCh38
rs776290743 Chr.2: 216415057-216415057 on Build GRCh38
rs765779796 Chr.2: 216415350-216415350 on Build GRCh38
rs777999697 Chr.2: 216415210-216415210 on Build GRCh38
rs372708949 Chr.2: 216415017-216415017 on Build GRCh38
rs371098968 Chr.2: 216415366-216415366 on Build GRCh38
rs768737511 Chr.2: 216415318-216415318 on Build GRCh38
rs1002105105 Chr.2: 216415285-216415285 on Build GRCh38
rs755477133 Chr.2: 216415270-216415270 on Build GRCh38
rs775634898 Chr.2: 216415222-216415222 on Build GRCh38
rs530505647 Chr.2: 216415304-216415304 on Build GRCh38
rs780006964 Chr.2: 216415308-216415308 on Build GRCh38
rs756174064 Chr.2: 216415370-216415371 on Build GRCh38
rs897578907 Chr.2: 216415134-216415134 on Build GRCh38
rs754147208 Chr.2: 216415269-216415269 on Build GRCh38
rs754343294 Chr.2: 216415159-216415159 on Build GRCh38
rs137914185 Chr.2: 216415437-216415437 on Build GRCh38
rs752739012 Chr.2: 216415295-216415295 on Build GRCh38
rs759332408 Chr.2: 216415060-216415060 on Build GRCh38
rs747775289 Chr.2: 216415231-216415231 on Build GRCh38
rs781023326 Chr.2: 216415119-216415120 on Build GRCh38
rs761271788 Chr.2: 216415325-216415325 on Build GRCh38
rs544173171 Chr.2: 216415353-216415353 on Build GRCh38
rs35907255 Chr.2: 216415127-216415127 on Build GRCh38
rs780581267 Chr.2: 216415157-216415157 on Build GRCh38
rs905827328 Chr.2: 216415135-216415135 on Build GRCh38
rs781515824 Chr.2: 216415402-216415402 on Build GRCh38
rs754228770 Chr.2: 216415342-216415342 on Build GRCh38
rs753215593 Chr.2: 216415352-216415352 on Build GRCh38
rs777896137 Chr.2: 216415303-216415303 on Build GRCh38
rs765629645 Chr.2: 216415442-216415442 on Build GRCh38
rs6734114 Chr.2: 216415323-216415323 on Build GRCh38
rs767158204 Chr.2: 216415130-216415130 on Build GRCh38
rs761014406 Chr.2: 216415263-216415263 on Build GRCh38
rs202031614 Chr.2: 216415044-216415044 on Build GRCh38
rs778801075 Chr.2: 216415215-216415215 on Build GRCh38
rs62178625 Chr.2: 216415083-216415083 on Build GRCh38
rs35406881 Chr.2: 216415297-216415297 on Build GRCh38
rs758472664 Chr.2: 216415355-216415355 on Build GRCh38
rs754790488 Chr.2: 216415413-216415413 on Build GRCh38
rs748106387 Chr.2: 216415427-216415427 on Build GRCh38
rs757584821 Chr.2: 216415394-216415394 on Build GRCh38
rs772275806 Chr.2: 216415052-216415052 on Build GRCh38
rs1015452878 Chr.2: 216415237-216415237 on Build GRCh38
rs771609695 Chr.2: 216415327-216415327 on Build GRCh38
rs758479373 Chr.2: 216415301-216415301 on Build GRCh38
rs35048226 Chr.2: 216415307-216415307 on Build GRCh38
rs985760536 Chr.2: 216415397-216415397 on Build GRCh38
rs910120660 Chr.2: 216415400-216415400 on Build GRCh38
rs1053318350 Chr.2: 216415078-216415078 on Build GRCh38
rs774325413 Chr.2: 216415319-216415319 on Build GRCh38
rs763691819 Chr.2: 216415097-216415097 on Build GRCh38
rs146658939 Chr.2: 216415430-216415430 on Build GRCh38
rs149567782 Chr.2: 216415000-216415000 on Build GRCh38
rs370414331 Chr.2: 216415414-216415414 on Build GRCh38
rs755113204 Chr.2: 216415172-216415172 on Build GRCh38
rs778973735 Chr.2: 216415190-216415190 on Build GRCh38
rs762590281 Chr.2: 216415086-216415086 on Build GRCh38
rs751582760 Chr.2: 216415325-216415325 on Build GRCh38
rs765946032 Chr.2: 216415339-216415339 on Build GRCh38
rs1046678608 Chr.2: 216415444-216415444 on Build GRCh38
rs750062612 Chr.2: 216415262-216415262 on Build GRCh38
rs765105653 Chr.2: 216415069-216415069 on Build GRCh38
rs375140716 Chr.2: 216415428-216415428 on Build GRCh38
rs761340743 Chr.2: 216415255-216415255 on Build GRCh38
rs376048935 Chr.2: 216415328-216415328 on Build GRCh38
rs759842283 Chr.2: 216415348-216415348 on Build GRCh38
rs768649610 Chr.2: 216415249-216415249 on Build GRCh38
rs865803048 Chr.2: 216415166-216415166 on Build GRCh38
rs779348137 Chr.2: 216415294-216415294 on Build GRCh38
rs781425240 Chr.2: 216414996-216414996 on Build GRCh38
rs534474398 Chr.2: 216415440-216415440 on Build GRCh38
rs772212169 Chr.2: 216415200-216415200 on Build GRCh38
rs766284659 Chr.2: 216415018-216415018 on Build GRCh38
rs149510554 Chr.2: 216415439-216415439 on Build GRCh38
rs1033691702 Chr.2: 216415291-216415291 on Build GRCh38
rs867970870 Chr.2: 216415443-216415443 on Build GRCh38
rs766719997 Chr.2: 216415268-216415268 on Build GRCh38
rs745698233 Chr.2: 216415212-216415212 on Build GRCh38

View Full Product Details