Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218399879-218400411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDSP1
Entrez Gene ID
58190
Gene Name
CTD small phosphatase 1
Gene Aliases
NIF3, NLI-IF, NLIIF, SCP1
UniGene
Hs.444468
Gene Chromosome Location
Chr.2: 218398338-218405941 on Build GRCh38
Transcripts
NM_001206878.1 NM_021198.2 NM_182642.2 XM_011511569.2 XM_011511570.2 XM_017004615.1 XM_017004616.1 XM_017004617.1 XM_017004618.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6382065 c.6C>A p.D2E Chr.2: 218400096-218400096 on Build GRCh38
1614359 c.17_18insCA p.I7fs*104 Chr.2: 218400107-218400108 on Build GRCh38
5708073 c.67+1G>T p.? Chr.2: 218400158-218400158 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs879677750 Chr.2: 218400208-218400208 on Build GRCh38
rs1046591506 Chr.2: 218400328-218400328 on Build GRCh38
rs976581068 Chr.2: 218399960-218399960 on Build GRCh38
rs1012429643 Chr.2: 218400118-218400118 on Build GRCh38
rs1029507243 Chr.2: 218400248-218400248 on Build GRCh38
rs748796724 Chr.2: 218400131-218400131 on Build GRCh38
rs1012874192 Chr.2: 218399930-218399930 on Build GRCh38
rs773593761 Chr.2: 218400168-218400168 on Build GRCh38
rs190100918 Chr.2: 218400326-218400326 on Build GRCh38
rs772437283 Chr.2: 218400166-218400166 on Build GRCh38
rs771005069 Chr.2: 218400160-218400160 on Build GRCh38
rs866328917 Chr.2: 218400132-218400132 on Build GRCh38
rs555872843 Chr.2: 218400125-218400125 on Build GRCh38
rs533705122 Chr.2: 218400004-218400004 on Build GRCh38
rs946462430 Chr.2: 218400367-218400367 on Build GRCh38
rs765236244 Chr.2: 218400310-218400310 on Build GRCh38
rs919236601 Chr.2: 218400323-218400323 on Build GRCh38
rs549174201 Chr.2: 218400034-218400034 on Build GRCh38
rs893526239 Chr.2: 218400355-218400355 on Build GRCh38
rs754114536 Chr.2: 218400259-218400259 on Build GRCh38
rs780044082 Chr.2: 218400159-218400160 on Build GRCh38
rs938172990 Chr.2: 218400338-218400338 on Build GRCh38
rs774858271 Chr.2: 218400163-218400163 on Build GRCh38
rs2579958 Chr.2: 218400371-218400371 on Build GRCh38
rs766562413 Chr.2: 218400180-218400180 on Build GRCh38
rs928175899 Chr.2: 218400334-218400334 on Build GRCh38
rs116233374 Chr.2: 218400343-218400343 on Build GRCh38
rs762243089 Chr.2: 218400165-218400165 on Build GRCh38
rs868227078 Chr.2: 218400225-218400225 on Build GRCh38
rs551378353 Chr.2: 218399950-218399950 on Build GRCh38
rs761064461 Chr.2: 218400179-218400179 on Build GRCh38
rs906675880 Chr.2: 218400135-218400135 on Build GRCh38
rs919485493 Chr.2: 218400109-218400109 on Build GRCh38
rs999744959 Chr.2: 218400369-218400369 on Build GRCh38
rs942020089 Chr.2: 218400038-218400038 on Build GRCh38
rs1037617948 Chr.2: 218399992-218399992 on Build GRCh38
rs1001911108 Chr.2: 218400154-218400154 on Build GRCh38
rs916679563 Chr.2: 218400344-218400344 on Build GRCh38
rs768119984 Chr.2: 218400155-218400155 on Build GRCh38
rs997902910 Chr.2: 218400217-218400217 on Build GRCh38
rs745478840 Chr.2: 218400120-218400120 on Build GRCh38
rs953858327 Chr.2: 218400260-218400260 on Build GRCh38
rs893505654 Chr.2: 218400206-218400206 on Build GRCh38
rs12620498 Chr.2: 218400262-218400262 on Build GRCh38
rs932483188 Chr.2: 218399989-218399992 on Build GRCh38
rs895399532 Chr.2: 218399925-218399925 on Build GRCh38
rs1022553157 Chr.2: 218399937-218399937 on Build GRCh38
rs867989709 Chr.2: 218400172-218400172 on Build GRCh38
rs957662181 Chr.2: 218399913-218399913 on Build GRCh38
rs922403988 Chr.2: 218399955-218399955 on Build GRCh38
rs909819301 Chr.2: 218399994-218399994 on Build GRCh38
rs759655402 Chr.2: 218400302-218400302 on Build GRCh38
rs1010973882 Chr.2: 218400211-218400211 on Build GRCh38
rs577208564 Chr.2: 218400230-218400230 on Build GRCh38
rs751455805 Chr.2: 218400322-218400322 on Build GRCh38
rs757010032 Chr.2: 218400096-218400096 on Build GRCh38
rs769541036 Chr.2: 218400122-218400122 on Build GRCh38
rs1056476748 Chr.2: 218400069-218400069 on Build GRCh38
rs955801892 Chr.2: 218399959-218399959 on Build GRCh38
rs771210899 Chr.2: 218400302-218400303 on Build GRCh38
rs954785777 Chr.2: 218400033-218400033 on Build GRCh38
rs1010863052 Chr.2: 218399917-218399917 on Build GRCh38
rs1021281848 Chr.2: 218399932-218399932 on Build GRCh38
rs12620519 Chr.2: 218400144-218400144 on Build GRCh38
rs777751173 Chr.2: 218400075-218400075 on Build GRCh38
rs537523584 Chr.2: 218400123-218400123 on Build GRCh38
rs910629366 Chr.2: 218400269-218400269 on Build GRCh38
rs976972132 Chr.2: 218399940-218399940 on Build GRCh38
rs749482555 Chr.2: 218400177-218400178 on Build GRCh38
rs780862243 Chr.2: 218400105-218400105 on Build GRCh38
rs986251159 Chr.2: 218400267-218400267 on Build GRCh38
rs770050335 Chr.2: 218400171-218400174 on Build GRCh38
rs781231169 Chr.2: 218400347-218400347 on Build GRCh38
rs1054799880 Chr.2: 218400189-218400189 on Build GRCh38
rs558120517 Chr.2: 218400278-218400278 on Build GRCh38
rs939475911 Chr.2: 218400063-218400063 on Build GRCh38
rs973831823 Chr.2: 218400319-218400319 on Build GRCh38
rs910598314 Chr.2: 218400035-218400035 on Build GRCh38
rs966596518 Chr.2: 218399938-218399938 on Build GRCh38
rs902138844 Chr.2: 218400380-218400380 on Build GRCh38
rs922392329 Chr.2: 218399981-218399981 on Build GRCh38
rs773537084 Chr.2: 218400176-218400177 on Build GRCh38
rs963434812 Chr.2: 218400295-218400295 on Build GRCh38
rs897778020 Chr.2: 218400255-218400255 on Build GRCh38
rs941341241 Chr.2: 218400031-218400031 on Build GRCh38
rs985335025 Chr.2: 218400256-218400256 on Build GRCh38
rs566992143 Chr.2: 218400090-218400090 on Build GRCh38
rs748226699 Chr.2: 218400164-218400165 on Build GRCh38
rs962693934 Chr.2: 218400268-218400268 on Build GRCh38
rs186986949 Chr.2: 218400240-218400240 on Build GRCh38
rs891441567 Chr.2: 218400372-218400372 on Build GRCh38
rs566808221 Chr.2: 218399993-218399993 on Build GRCh38
rs967046517 Chr.2: 218400233-218400233 on Build GRCh38
rs1037987118 Chr.2: 218400098-218400098 on Build GRCh38
rs918664389 Chr.2: 218400051-218400051 on Build GRCh38
rs953990363 Chr.2: 218400234-218400235 on Build GRCh38

View Full Product Details