Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219279698-219280221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB2
Entrez Gene ID
3300
Gene Name
DnaJ heat shock protein family (Hsp40) member B2
Gene Aliases
CMT2T, DSMA5, HSJ-1, HSJ1, HSPF3
UniGene
Hs.77768
Gene Chromosome Location
Chr.2: 219279318-219286900 on Build GRCh38
Transcripts
NM_001039550.1 NM_006736.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6897634 c.34C>T p.R12* Chr.2: 219279867-219279867 on Build GRCh38
5834389 c.27C>T p.D9D Chr.2: 219279860-219279860 on Build GRCh38
4133479 c.42G>A p.A14A Chr.2: 219279875-219279875 on Build GRCh38
6897635 c.34C>T p.R12* Chr.2: 219279867-219279867 on Build GRCh38
442344 c.27C>T p.D9D Chr.2: 219279860-219279860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757100106 Chr.2: 219279934-219279934 on Build GRCh38
rs991804706 Chr.2: 219280183-219280183 on Build GRCh38
rs754824979 Chr.2: 219280048-219280048 on Build GRCh38
rs970821372 Chr.2: 219279896-219279896 on Build GRCh38
rs3770223 Chr.2: 219280197-219280197 on Build GRCh38
rs766354069 Chr.2: 219279875-219279875 on Build GRCh38
rs770735247 Chr.2: 219279798-219279798 on Build GRCh38
rs914530440 Chr.2: 219280082-219280082 on Build GRCh38
rs777721294 Chr.2: 219279787-219279787 on Build GRCh38
rs368141673 Chr.2: 219279799-219279799 on Build GRCh38
rs776134515 Chr.2: 219279810-219279810 on Build GRCh38
rs781558713 Chr.2: 219280064-219280064 on Build GRCh38
rs375674959 Chr.2: 219279924-219279924 on Build GRCh38
rs1044556723 Chr.2: 219279910-219279910 on Build GRCh38
rs767774707 Chr.2: 219279860-219279860 on Build GRCh38
rs887348297 Chr.2: 219279825-219279825 on Build GRCh38
rs370352788 Chr.2: 219279944-219279944 on Build GRCh38
rs751321002 Chr.2: 219279967-219279967 on Build GRCh38
rs996442021 Chr.2: 219280075-219280075 on Build GRCh38
rs777809028 Chr.2: 219279914-219279914 on Build GRCh38
rs750308678 Chr.2: 219279775-219279775 on Build GRCh38
rs758270233 Chr.2: 219279784-219279784 on Build GRCh38
rs569003467 Chr.2: 219279960-219279960 on Build GRCh38
rs375597355 Chr.2: 219279783-219279783 on Build GRCh38
rs563695457 Chr.2: 219279736-219279736 on Build GRCh38
rs373472689 Chr.2: 219279827-219279827 on Build GRCh38
rs781035633 Chr.2: 219279936-219279936 on Build GRCh38
rs140751061 Chr.2: 219279913-219279913 on Build GRCh38
rs201236722 Chr.2: 219279948-219279948 on Build GRCh38
rs748766976 Chr.2: 219279945-219279945 on Build GRCh38
rs370901455 Chr.2: 219279811-219279811 on Build GRCh38
rs190841464 Chr.2: 219279961-219279961 on Build GRCh38
rs769407095 Chr.2: 219279942-219279942 on Build GRCh38
rs891806967 Chr.2: 219280018-219280018 on Build GRCh38
rs769158342 Chr.2: 219279808-219279808 on Build GRCh38
rs765136053 Chr.2: 219279828-219279828 on Build GRCh38
rs553915256 Chr.2: 219280165-219280165 on Build GRCh38
rs201783002 Chr.2: 219279800-219279800 on Build GRCh38
rs534163479 Chr.2: 219280118-219280118 on Build GRCh38
rs757154933 Chr.2: 219279784-219279786 on Build GRCh38
rs917518042 Chr.2: 219280184-219280184 on Build GRCh38
rs745505914 Chr.2: 219279937-219279937 on Build GRCh38
rs140932966 Chr.2: 219279861-219279861 on Build GRCh38
rs750395810 Chr.2: 219279940-219279940 on Build GRCh38
rs951929741 Chr.2: 219280150-219280150 on Build GRCh38
rs943765264 Chr.2: 219279777-219279777 on Build GRCh38
rs1041769412 Chr.2: 219279773-219279773 on Build GRCh38
rs571641215 Chr.2: 219280052-219280052 on Build GRCh38
rs753895049 Chr.2: 219279888-219279888 on Build GRCh38
rs879104936 Chr.2: 219279939-219279939 on Build GRCh38
rs532031093 Chr.2: 219279723-219279723 on Build GRCh38
rs1027503516 Chr.2: 219280086-219280086 on Build GRCh38
rs760873950 Chr.2: 219279868-219279868 on Build GRCh38
rs562983134 Chr.2: 219279719-219279719 on Build GRCh38
rs914225594 Chr.2: 219279730-219279730 on Build GRCh38
rs545301771 Chr.2: 219279724-219279724 on Build GRCh38
rs983206433 Chr.2: 219280151-219280151 on Build GRCh38
rs368516886 Chr.2: 219280032-219280032 on Build GRCh38
rs730882140 Chr.2: 219279847-219279847 on Build GRCh38
rs938963588 Chr.2: 219279734-219279734 on Build GRCh38
rs574072787 Chr.2: 219280186-219280186 on Build GRCh38
rs774449434 Chr.2: 219279932-219279932 on Build GRCh38
rs958490401 Chr.2: 219280041-219280041 on Build GRCh38
rs759190434 Chr.2: 219279933-219279933 on Build GRCh38
rs774702789 Chr.2: 219279841-219279841 on Build GRCh38
rs960398003 Chr.2: 219280176-219280176 on Build GRCh38
rs967688934 Chr.2: 219280150-219280152 on Build GRCh38
rs746732281 Chr.2: 219279789-219279789 on Build GRCh38

View Full Product Details