Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 241087015-241087520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTERF4
Entrez Gene ID
130916
Gene Name
mitochondrial transcription termination factor 4
Gene Aliases
MTERFD2
UniGene
Hs.159556
Gene Chromosome Location
Chr.2: 241072168-241103367 on Build GRCh38
Transcripts
NR_028049.1 NR_028050.1 NR_028051.1 XM_006712294.1 XM_006712295.3 XM_011510634.2 XM_017003379.1 XM_017003380.1 XM_017003381.1 XM_017003382.1 XM_017003383.1 XM_017003384.1 XM_017003385.1 XM_017003386.1 XM_017003387.1 XM_017003389.1 XM_017003390.1 XM_017003391.1 NM_001080437.1 XM_011510931.2 XM_011510932.2 XM_017003773.1 XM_017003774.1 XM_017003775.1 XM_017003776.1 XM_017003777.1 XM_017003778.1 XM_017003779.1 XM_017003780.1 XM_017003781.1 XM_017003782.1

Gene Symbol
SNED1
Entrez Gene ID
25992
Gene Name
sushi, nidogen and EGF like domains 1
Gene Aliases
SST3, Snep
UniGene
Hs.471834
Gene Chromosome Location
Chr.2: 240997409-241095568 on Build GRCh38
Transcripts
NR_028049.1 NR_028050.1 NR_028051.1 XM_006712294.1 XM_006712295.3 XM_011510634.2 XM_017003379.1 XM_017003380.1 XM_017003381.1 XM_017003382.1 XM_017003383.1 XM_017003384.1 XM_017003385.1 XM_017003386.1 XM_017003387.1 XM_017003389.1 XM_017003390.1 XM_017003391.1 NM_001080437.1 XM_011510931.2 XM_011510932.2 XM_017003773.1 XM_017003774.1 XM_017003775.1 XM_017003776.1 XM_017003777.1 XM_017003778.1 XM_017003779.1 XM_017003780.1 XM_017003781.1 XM_017003782.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5746071 c.4064A>G p.Y1355C Chr.2: 241087394-241087394 on Build GRCh38
4971630 c.4137C>T p.T1379T Chr.2: 241087467-241087467 on Build GRCh38
3991197 c.4080A>T p.R1360R Chr.2: 241087410-241087410 on Build GRCh38
263796 c.4124G>A p.S1375N Chr.2: 241087454-241087454 on Build GRCh38
5025880 c.4102A>T p.K1368* Chr.2: 241087432-241087432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs4035 Chr.2: 241087193-241087193 on Build GRCh38
rs112035079 Chr.2: 241087041-241087041 on Build GRCh38
rs895833551 Chr.2: 241087428-241087428 on Build GRCh38
rs372568480 Chr.2: 241087425-241087425 on Build GRCh38
rs780746868 Chr.2: 241087466-241087466 on Build GRCh38
rs752479794 Chr.2: 241087414-241087414 on Build GRCh38
rs530628665 Chr.2: 241087107-241087107 on Build GRCh38
rs1011929006 Chr.2: 241087228-241087228 on Build GRCh38
rs760773814 Chr.2: 241087349-241087349 on Build GRCh38
rs1032095302 Chr.2: 241087166-241087166 on Build GRCh38
rs759617899 Chr.2: 241087385-241087385 on Build GRCh38
rs923155254 Chr.2: 241087202-241087202 on Build GRCh38
rs776698657 Chr.2: 241087359-241087359 on Build GRCh38
rs541412404 Chr.2: 241087064-241087064 on Build GRCh38
rs1013478353 Chr.2: 241087095-241087095 on Build GRCh38
rs746134930 Chr.2: 241087082-241087082 on Build GRCh38
rs568339809 Chr.2: 241087444-241087444 on Build GRCh38
rs911161277 Chr.2: 241087066-241087066 on Build GRCh38
rs765911335 Chr.2: 241087311-241087311 on Build GRCh38
rs929280416 Chr.2: 241087423-241087423 on Build GRCh38
rs941909792 Chr.2: 241087270-241087270 on Build GRCh38
rs756678043 Chr.2: 241087396-241087397 on Build GRCh38
rs765274068 Chr.2: 241087400-241087400 on Build GRCh38
rs751218942 Chr.2: 241087451-241087451 on Build GRCh38
rs953442240 Chr.2: 241087469-241087469 on Build GRCh38
rs762407789 Chr.2: 241087279-241087279 on Build GRCh38
rs755416066 Chr.2: 241087494-241087494 on Build GRCh38
rs377479123 Chr.2: 241087260-241087260 on Build GRCh38
rs746314946 Chr.2: 241087501-241087501 on Build GRCh38
rs986483706 Chr.2: 241087248-241087248 on Build GRCh38
rs576745840 Chr.2: 241087140-241087142 on Build GRCh38
rs563179979 Chr.2: 241087081-241087081 on Build GRCh38
rs375760729 Chr.2: 241087446-241087446 on Build GRCh38
rs370177685 Chr.2: 241087358-241087358 on Build GRCh38
rs772302174 Chr.2: 241087104-241087104 on Build GRCh38
rs112333804 Chr.2: 241087147-241087147 on Build GRCh38
rs747470025 Chr.2: 241087170-241087170 on Build GRCh38
rs374367773 Chr.2: 241087195-241087198 on Build GRCh38
rs13411556 Chr.2: 241087234-241087234 on Build GRCh38
rs764499686 Chr.2: 241087239-241087239 on Build GRCh38
rs968908734 Chr.2: 241087142-241087142 on Build GRCh38
rs369008078 Chr.2: 241087478-241087478 on Build GRCh38

View Full Product Details