Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35548431-35548932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERGIC3
Entrez Gene ID
51614
Gene Name
ERGIC and golgi 3
Gene Aliases
C20orf47, CGI-54, Erv46, NY-BR-84, PRO0989, SDBCAG84, dJ477O4.2
UniGene
Hs.472558
Gene Chromosome Location
Chr.20: 35542029-35557634 on Build GRCh38
Transcripts
NM_015966.2 NM_198398.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3716520 c.509G>T p.G170V Chr.20: 35548556-35548556 on Build GRCh38
6309359 c.489G>T p.R163R Chr.20: 35548536-35548536 on Build GRCh38
3716519 c.508G>T p.G170C Chr.20: 35548555-35548555 on Build GRCh38
6258788 c.657G>T p.G219G Chr.20: 35548837-35548837 on Build GRCh38
1026255 c.604T>C p.Y202H Chr.20: 35548651-35548651 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752814923 Chr.20: 35548655-35548655 on Build GRCh38
rs760886960 Chr.20: 35548698-35548698 on Build GRCh38
rs762724761 Chr.20: 35548677-35548677 on Build GRCh38
rs775066956 Chr.20: 35548493-35548493 on Build GRCh38
rs756421513 Chr.20: 35548578-35548578 on Build GRCh38
rs748912772 Chr.20: 35548904-35548904 on Build GRCh38
rs759620258 Chr.20: 35548690-35548690 on Build GRCh38
rs915537140 Chr.20: 35548563-35548563 on Build GRCh38
rs199935905 Chr.20: 35548546-35548546 on Build GRCh38
rs757609562 Chr.20: 35548709-35548709 on Build GRCh38
rs1054657718 Chr.20: 35548744-35548744 on Build GRCh38
rs757775545 Chr.20: 35548595-35548595 on Build GRCh38
rs368439529 Chr.20: 35548903-35548903 on Build GRCh38
rs373639727 Chr.20: 35548489-35548489 on Build GRCh38
rs376497037 Chr.20: 35548879-35548879 on Build GRCh38
rs764363895 Chr.20: 35548701-35548701 on Build GRCh38
rs149433368 Chr.20: 35548536-35548536 on Build GRCh38
rs773239963 Chr.20: 35548542-35548542 on Build GRCh38
rs780955750 Chr.20: 35548659-35548659 on Build GRCh38
rs770890066 Chr.20: 35548685-35548685 on Build GRCh38
rs779344390 Chr.20: 35548634-35548634 on Build GRCh38
rs768338368 Chr.20: 35548527-35548527 on Build GRCh38
rs779446825 Chr.20: 35548713-35548713 on Build GRCh38
rs893060760 Chr.20: 35548805-35548805 on Build GRCh38
rs780777706 Chr.20: 35548886-35548886 on Build GRCh38
rs200914198 Chr.20: 35548459-35548459 on Build GRCh38
rs774786739 Chr.20: 35548786-35548787 on Build GRCh38
rs765543862 Chr.20: 35548877-35548877 on Build GRCh38
rs752858435 Chr.20: 35548697-35548697 on Build GRCh38
rs1036973147 Chr.20: 35548534-35548534 on Build GRCh38
rs74834954 Chr.20: 35548762-35548762 on Build GRCh38
rs769655761 Chr.20: 35548669-35548669 on Build GRCh38
rs969239936 Chr.20: 35548597-35548597 on Build GRCh38
rs750884905 Chr.20: 35548878-35548878 on Build GRCh38
rs374124575 Chr.20: 35548535-35548535 on Build GRCh38
rs758838039 Chr.20: 35548884-35548884 on Build GRCh38
rs146493836 Chr.20: 35548831-35548831 on Build GRCh38
rs747829579 Chr.20: 35548665-35548665 on Build GRCh38
rs367816715 Chr.20: 35548490-35548490 on Build GRCh38
rs777629170 Chr.20: 35548777-35548777 on Build GRCh38
rs765410361 Chr.20: 35548598-35548598 on Build GRCh38
rs752245559 Chr.20: 35548887-35548887 on Build GRCh38
rs921160779 Chr.20: 35548741-35548741 on Build GRCh38
rs763783785 Chr.20: 35548504-35548505 on Build GRCh38
rs746564496 Chr.20: 35548526-35548526 on Build GRCh38
rs139314230 Chr.20: 35548550-35548550 on Build GRCh38
rs958716105 Chr.20: 35548587-35548587 on Build GRCh38
rs764276399 Chr.20: 35548846-35548846 on Build GRCh38
rs776474299 Chr.20: 35548529-35548529 on Build GRCh38
rs768095192 Chr.20: 35548640-35548640 on Build GRCh38
rs976680854 Chr.20: 35548727-35548727 on Build GRCh38
rs778119325 Chr.20: 35548579-35548579 on Build GRCh38
rs760358675 Chr.20: 35548861-35548861 on Build GRCh38
rs770660482 Chr.20: 35548790-35548790 on Build GRCh38
rs749724539 Chr.20: 35548594-35548594 on Build GRCh38
rs77272989 Chr.20: 35548469-35548469 on Build GRCh38
rs202155461 Chr.20: 35548715-35548715 on Build GRCh38
rs746508284 Chr.20: 35548635-35548635 on Build GRCh38
rs755737719 Chr.20: 35548898-35548898 on Build GRCh38
rs182209758 Chr.20: 35548671-35548671 on Build GRCh38
rs934492507 Chr.20: 35548735-35548735 on Build GRCh38
rs762997368 Chr.20: 35548544-35548544 on Build GRCh38
rs749088943 Chr.20: 35548789-35548789 on Build GRCh38
rs531605955 Chr.20: 35548572-35548572 on Build GRCh38
rs754185898 Chr.20: 35548705-35548705 on Build GRCh38
rs752946417 Chr.20: 35548562-35548562 on Build GRCh38
rs867087855 Chr.20: 35548731-35548731 on Build GRCh38
rs547706779 Chr.20: 35548905-35548905 on Build GRCh38
rs200704504 Chr.20: 35548864-35548864 on Build GRCh38
rs932308355 Chr.20: 35548814-35548814 on Build GRCh38
rs751656462 Chr.20: 35548547-35548547 on Build GRCh38
rs769569948 Chr.20: 35548776-35548776 on Build GRCh38
rs760800923 Chr.20: 35548837-35548837 on Build GRCh38
rs747739875 Chr.20: 35548775-35548775 on Build GRCh38
rs775543043 Chr.20: 35548833-35548833 on Build GRCh38
rs144274449 Chr.20: 35548826-35548826 on Build GRCh38
rs754049984 Chr.20: 35548858-35548858 on Build GRCh38
rs767605074 Chr.20: 35548696-35548696 on Build GRCh38
rs532717308 Chr.20: 35548771-35548771 on Build GRCh38
rs774292745 Chr.20: 35548796-35548796 on Build GRCh38
rs754462476 Chr.20: 35548722-35548722 on Build GRCh38

View Full Product Details