Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63254234-63254759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ16779
Entrez Gene ID
100192386
Gene Name
uncharacterized LOC100192386
Gene Aliases
-
UniGene
Hs.111539
Gene Chromosome Location
Chr.20: 63253978-63261615 on Build GRCh38
Transcripts
NR_024389.1 NM_152864.3 XM_005260192.1 XM_011528527.1 XM_017027636.1

Gene Symbol
NKAIN4
Entrez Gene ID
128414
Gene Name
Na+/K+ transporting ATPase interacting 4
Gene Aliases
C20orf58, FAM77A, bA261N11.2
UniGene
Hs.46627
Gene Chromosome Location
Chr.20: 63240784-63254904 on Build GRCh38
Transcripts
NR_024389.1 NM_152864.3 XM_005260192.1 XM_011528527.1 XM_017027636.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7318380 c.39C>G p.L13L Chr.20: 63254412-63254412 on Build GRCh38
6860412 c.54+9C>T p.? Chr.20: 63254388-63254388 on Build GRCh38
5772932 c.54+7G>T p.? Chr.20: 63254390-63254390 on Build GRCh38
6046953 c.1A>G p.M1V Chr.20: 63254450-63254450 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760683400 Chr.20: 63254450-63254450 on Build GRCh38
rs554732868 Chr.20: 63254279-63254279 on Build GRCh38
rs894240315 Chr.20: 63254467-63254467 on Build GRCh38
rs559690731 Chr.20: 63254605-63254605 on Build GRCh38
rs35796278 Chr.20: 63254535-63254535 on Build GRCh38
rs940255032 Chr.20: 63254641-63254641 on Build GRCh38
rs900351685 Chr.20: 63254633-63254633 on Build GRCh38
rs931441208 Chr.20: 63254682-63254682 on Build GRCh38
rs957160259 Chr.20: 63254504-63254504 on Build GRCh38
rs757784874 Chr.20: 63254372-63254372 on Build GRCh38
rs758024523 Chr.20: 63254388-63254388 on Build GRCh38
rs998761787 Chr.20: 63254500-63254500 on Build GRCh38
rs954372182 Chr.20: 63254398-63254398 on Build GRCh38
rs990005725 Chr.20: 63254524-63254524 on Build GRCh38
rs891672932 Chr.20: 63254327-63254327 on Build GRCh38
rs775981629 Chr.20: 63254501-63254502 on Build GRCh38
rs542039776 Chr.20: 63254695-63254695 on Build GRCh38
rs1027216312 Chr.20: 63254723-63254723 on Build GRCh38
rs764894081 Chr.20: 63254380-63254380 on Build GRCh38
rs866320314 Chr.20: 63254565-63254565 on Build GRCh38
rs990230885 Chr.20: 63254354-63254354 on Build GRCh38
rs921779295 Chr.20: 63254502-63254510 on Build GRCh38
rs58426104 Chr.20: 63254383-63254383 on Build GRCh38
rs961620120 Chr.20: 63254455-63254455 on Build GRCh38
rs1023996499 Chr.20: 63254481-63254481 on Build GRCh38
rs998477202 Chr.20: 63254730-63254730 on Build GRCh38
rs995676795 Chr.20: 63254437-63254437 on Build GRCh38
rs1051113612 Chr.20: 63254374-63254374 on Build GRCh38
rs907116136 Chr.20: 63254484-63254484 on Build GRCh38
rs771932808 Chr.20: 63254255-63254256 on Build GRCh38
rs924476626 Chr.20: 63254297-63254297 on Build GRCh38
rs568234045 Chr.20: 63254470-63254491 on Build GRCh38
rs920077583 Chr.20: 63254480-63254480 on Build GRCh38
rs1042918289 Chr.20: 63254597-63254597 on Build GRCh38
rs576516019 Chr.20: 63254291-63254291 on Build GRCh38
rs1049253974 Chr.20: 63254444-63254444 on Build GRCh38
rs1012972092 Chr.20: 63254475-63254475 on Build GRCh38
rs1045802067 Chr.20: 63254704-63254704 on Build GRCh38
rs928695892 Chr.20: 63254495-63254495 on Build GRCh38
rs1004590547 Chr.20: 63254700-63254700 on Build GRCh38
rs932609492 Chr.20: 63254363-63254363 on Build GRCh38
rs541295527 Chr.20: 63254598-63254598 on Build GRCh38
rs1053043865 Chr.20: 63254533-63254533 on Build GRCh38
rs779731402 Chr.20: 63254415-63254415 on Build GRCh38
rs948525842 Chr.20: 63254702-63254702 on Build GRCh38
rs1028795853 Chr.20: 63254599-63254599 on Build GRCh38
rs898707106 Chr.20: 63254431-63254431 on Build GRCh38
rs957443988 Chr.20: 63254341-63254341 on Build GRCh38
rs537029699 Chr.20: 63254331-63254331 on Build GRCh38
rs867212486 Chr.20: 63254304-63254304 on Build GRCh38
rs954180959 Chr.20: 63254600-63254600 on Build GRCh38
rs912600290 Chr.20: 63254634-63254634 on Build GRCh38
rs754405977 Chr.20: 63254362-63254362 on Build GRCh38
rs894196948 Chr.20: 63254697-63254697 on Build GRCh38
rs1040653639 Chr.20: 63254482-63254500 on Build GRCh38
rs947290365 Chr.20: 63254580-63254580 on Build GRCh38
rs971356425 Chr.20: 63254274-63254274 on Build GRCh38
rs554316769 Chr.20: 63254542-63254543 on Build GRCh38
rs1052973218 Chr.20: 63254313-63254313 on Build GRCh38
rs773338934 Chr.20: 63254433-63254438 on Build GRCh38
rs754795807 Chr.20: 63254457-63254457 on Build GRCh38
rs6090334 Chr.20: 63254681-63254681 on Build GRCh38
rs1044434539 Chr.20: 63254253-63254253 on Build GRCh38
rs1031513289 Chr.20: 63254739-63254739 on Build GRCh38
rs780897949 Chr.20: 63254472-63254472 on Build GRCh38
rs1014591312 Chr.20: 63254710-63254710 on Build GRCh38
rs987194398 Chr.20: 63254621-63254621 on Build GRCh38
rs746592842 Chr.20: 63254435-63254435 on Build GRCh38
rs761052281 Chr.20: 63254663-63254663 on Build GRCh38
rs906916607 Chr.20: 63254720-63254720 on Build GRCh38
rs972941146 Chr.20: 63254642-63254642 on Build GRCh38
rs921359503 Chr.20: 63254359-63254359 on Build GRCh38
rs1031460917 Chr.20: 63254501-63254501 on Build GRCh38

View Full Product Details