Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 64092715-64093218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPRL1
Entrez Gene ID
4987
Gene Name
opioid related nociceptin receptor 1
Gene Aliases
KOR-3, NOCIR, NOP, NOPr, OOR, ORL1
UniGene
Hs.2859
Gene Chromosome Location
Chr.20: 64080082-64100643 on Build GRCh38
Transcripts
NM_000913.5 NM_001200019.1 NM_001318853.1 NM_001318854.1 NM_001318855.1 NM_182647.3 XM_011528828.2 XM_011528830.2 XM_017027852.1 XM_017027853.1 XM_017027854.1 XM_017027855.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4100566 c.233G>T p.R78M Chr.20: 64092953-64092953 on Build GRCh38
4709547 c.159C>A p.T53T Chr.20: 64092879-64092879 on Build GRCh38
137226 c.85C>T p.H29Y Chr.20: 64092805-64092805 on Build GRCh38
6160700 c.127G>T p.G43C Chr.20: 64092847-64092847 on Build GRCh38
3549223 c.199C>T p.L67L Chr.20: 64092919-64092919 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780063002 Chr.20: 64092895-64092895 on Build GRCh38
rs751386815 Chr.20: 64092806-64092806 on Build GRCh38
rs371951246 Chr.20: 64092842-64092842 on Build GRCh38
rs755332325 Chr.20: 64092783-64092783 on Build GRCh38
rs752591370 Chr.20: 64092867-64092867 on Build GRCh38
rs148535906 Chr.20: 64092736-64092736 on Build GRCh38
rs947699987 Chr.20: 64093030-64093030 on Build GRCh38
rs781195365 Chr.20: 64092816-64092816 on Build GRCh38
rs752611444 Chr.20: 64092963-64092963 on Build GRCh38
rs200138027 Chr.20: 64092939-64092939 on Build GRCh38
rs980611931 Chr.20: 64092819-64092819 on Build GRCh38
rs967962869 Chr.20: 64093010-64093010 on Build GRCh38
rs754819532 Chr.20: 64092815-64092815 on Build GRCh38
rs767590961 Chr.20: 64092951-64092951 on Build GRCh38
rs984776622 Chr.20: 64093019-64093019 on Build GRCh38
rs754188071 Chr.20: 64092762-64092762 on Build GRCh38
rs371386684 Chr.20: 64092738-64092738 on Build GRCh38
rs755983791 Chr.20: 64092967-64092967 on Build GRCh38
rs774642693 Chr.20: 64092741-64092741 on Build GRCh38
rs113541384 Chr.20: 64092737-64092737 on Build GRCh38
rs573091811 Chr.20: 64092753-64092753 on Build GRCh38
rs930223125 Chr.20: 64092798-64092798 on Build GRCh38
rs776074996 Chr.20: 64092834-64092834 on Build GRCh38
rs529551188 Chr.20: 64093049-64093049 on Build GRCh38
rs1044775824 Chr.20: 64092843-64092843 on Build GRCh38
rs771974896 Chr.20: 64092831-64092831 on Build GRCh38
rs769147015 Chr.20: 64092836-64092836 on Build GRCh38
rs763850004 Chr.20: 64092969-64092969 on Build GRCh38
rs146075237 Chr.20: 64092849-64092849 on Build GRCh38
rs770079623 Chr.20: 64092910-64092910 on Build GRCh38
rs772932512 Chr.20: 64092917-64092917 on Build GRCh38
rs368400897 Chr.20: 64092821-64092821 on Build GRCh38
rs757799727 Chr.20: 64092784-64092784 on Build GRCh38
rs761551555 Chr.20: 64093002-64093003 on Build GRCh38
rs979056099 Chr.20: 64093067-64093067 on Build GRCh38
rs768088736 Chr.20: 64092744-64092744 on Build GRCh38
rs547162294 Chr.20: 64093018-64093018 on Build GRCh38
rs781248458 Chr.20: 64092865-64092865 on Build GRCh38
rs1051265124 Chr.20: 64092874-64092874 on Build GRCh38
rs767058347 Chr.20: 64093004-64093004 on Build GRCh38
rs751069614 Chr.20: 64092860-64092860 on Build GRCh38
rs764836434 Chr.20: 64092764-64092764 on Build GRCh38
rs777074407 Chr.20: 64092901-64092901 on Build GRCh38
rs764893260 Chr.20: 64092846-64092846 on Build GRCh38
rs147164184 Chr.20: 64093097-64093097 on Build GRCh38
rs754400783 Chr.20: 64092743-64092743 on Build GRCh38
rs201975936 Chr.20: 64092751-64092751 on Build GRCh38
rs995514462 Chr.20: 64092776-64092776 on Build GRCh38
rs918758456 Chr.20: 64093003-64093003 on Build GRCh38
rs138017190 Chr.20: 64092894-64092894 on Build GRCh38
rs112318144 Chr.20: 64092850-64092850 on Build GRCh38
rs375635141 Chr.20: 64092739-64092739 on Build GRCh38
rs533394564 Chr.20: 64092995-64092995 on Build GRCh38
rs745988682 Chr.20: 64092824-64092824 on Build GRCh38
rs775816175 Chr.20: 64092746-64092746 on Build GRCh38
rs2229204 Chr.20: 64092882-64092882 on Build GRCh38
rs575515212 Chr.20: 64093022-64093022 on Build GRCh38
rs1010761729 Chr.20: 64093170-64093170 on Build GRCh38
rs146923933 Chr.20: 64092822-64092822 on Build GRCh38
rs374016312 Chr.20: 64092909-64092909 on Build GRCh38
rs562130539 Chr.20: 64092817-64092817 on Build GRCh38
rs113943566 Chr.20: 64092740-64092740 on Build GRCh38
rs892289013 Chr.20: 64093109-64093109 on Build GRCh38
rs940140688 Chr.20: 64093135-64093135 on Build GRCh38
rs753661159 Chr.20: 64092976-64092976 on Build GRCh38
rs927696574 Chr.20: 64093035-64093035 on Build GRCh38
rs550948910 Chr.20: 64093192-64093192 on Build GRCh38
rs55872076 Chr.20: 64092900-64092900 on Build GRCh38
rs762573593 Chr.20: 64092927-64092927 on Build GRCh38
rs756968755 Chr.20: 64092763-64092763 on Build GRCh38
rs780072581 Chr.20: 64092802-64092802 on Build GRCh38
rs759099539 Chr.20: 64092943-64092943 on Build GRCh38
rs755034870 Chr.20: 64092864-64092864 on Build GRCh38
rs932416924 Chr.20: 64093090-64093090 on Build GRCh38
rs1037253282 Chr.20: 64092899-64092899 on Build GRCh38
rs1037220410 Chr.20: 64093160-64093160 on Build GRCh38
rs889050043 Chr.20: 64093173-64093173 on Build GRCh38
rs1044723187 Chr.20: 64093033-64093033 on Build GRCh38
rs143480649 Chr.20: 64092994-64092994 on Build GRCh38
rs758586623 Chr.20: 64092979-64092979 on Build GRCh38
rs531179465 Chr.20: 64093172-64093172 on Build GRCh38
rs1034854856 Chr.20: 64092962-64092962 on Build GRCh38
rs773698884 Chr.20: 64092914-64092926 on Build GRCh38
rs6010720 Chr.20: 64093106-64093106 on Build GRCh38
rs750004258 Chr.20: 64092847-64092847 on Build GRCh38
rs751075811 Chr.20: 64092942-64092942 on Build GRCh38
rs779277717 Chr.20: 64092883-64092883 on Build GRCh38
rs6010719 Chr.20: 64093085-64093085 on Build GRCh38
rs749985655 Chr.20: 64092767-64092767 on Build GRCh38

View Full Product Details