Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 64199813-64200316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYT1
Entrez Gene ID
4661
Gene Name
myelin transcription factor 1
Gene Aliases
C20orf36, MTF1, MYTI, NZF2, PLPB1, ZC2H2C1, ZC2HC4A
UniGene
Hs.279562
Gene Chromosome Location
Chr.20: 64164474-64242253 on Build GRCh38
Transcripts
NM_004535.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1728372 c.68C>A p.T23N Chr.20: 64199904-64199904 on Build GRCh38
4100584 c.66G>T p.E22D Chr.20: 64199902-64199902 on Build GRCh38
4100582 c.66G>T p.E22D Chr.20: 64199902-64199902 on Build GRCh38
6160693 c.86+1G>T p.? Chr.20: 64199923-64199923 on Build GRCh38
1728371 c.68C>A p.T23N Chr.20: 64199904-64199904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201935947 Chr.20: 64199854-64199854 on Build GRCh38
rs989636267 Chr.20: 64199994-64199994 on Build GRCh38
rs373321123 Chr.20: 64199960-64199960 on Build GRCh38
rs762919782 Chr.20: 64199995-64199995 on Build GRCh38
rs754044323 Chr.20: 64200024-64200034 on Build GRCh38
rs532711755 Chr.20: 64200221-64200221 on Build GRCh38
rs545287866 Chr.20: 64200113-64200113 on Build GRCh38
rs764192787 Chr.20: 64200046-64200046 on Build GRCh38
rs3003138 Chr.20: 64200150-64200150 on Build GRCh38
rs957026061 Chr.20: 64199982-64199982 on Build GRCh38
rs918988420 Chr.20: 64200094-64200094 on Build GRCh38
rs75244365 Chr.20: 64200243-64200243 on Build GRCh38
rs765280642 Chr.20: 64199897-64199897 on Build GRCh38
rs570877728 Chr.20: 64199950-64199950 on Build GRCh38
rs367596232 Chr.20: 64200114-64200114 on Build GRCh38
rs1003524140 Chr.20: 64200160-64200160 on Build GRCh38
rs1052878606 Chr.20: 64200162-64200162 on Build GRCh38
rs753917257 Chr.20: 64199893-64199893 on Build GRCh38
rs767046254 Chr.20: 64200139-64200139 on Build GRCh38
rs750970145 Chr.20: 64199902-64199902 on Build GRCh38
rs780670285 Chr.20: 64199951-64199951 on Build GRCh38
rs758902158 Chr.20: 64199907-64199907 on Build GRCh38
rs374771555 Chr.20: 64199938-64199938 on Build GRCh38
rs1019115212 Chr.20: 64200233-64200233 on Build GRCh38
rs372202998 Chr.20: 64199889-64199889 on Build GRCh38
rs769881690 Chr.20: 64199868-64199868 on Build GRCh38
rs984984420 Chr.20: 64200289-64200289 on Build GRCh38
rs550087308 Chr.20: 64200208-64200208 on Build GRCh38
rs747680471 Chr.20: 64199953-64199953 on Build GRCh38
rs768185094 Chr.20: 64199848-64199848 on Build GRCh38
rs757279813 Chr.20: 64199896-64199896 on Build GRCh38
rs1022914888 Chr.20: 64200007-64200007 on Build GRCh38
rs971707457 Chr.20: 64200085-64200085 on Build GRCh38
rs911550720 Chr.20: 64200048-64200048 on Build GRCh38
rs761014186 Chr.20: 64199888-64199888 on Build GRCh38
rs766435549 Chr.20: 64200099-64200102 on Build GRCh38
rs908699089 Chr.20: 64200222-64200222 on Build GRCh38
rs576240250 Chr.20: 64200080-64200080 on Build GRCh38
rs371758296 Chr.20: 64200244-64200244 on Build GRCh38
rs897447748 Chr.20: 64199872-64199872 on Build GRCh38
rs573819351 Chr.20: 64199975-64199975 on Build GRCh38
rs778712633 Chr.20: 64199862-64199863 on Build GRCh38
rs748798194 Chr.20: 64200209-64200209 on Build GRCh38
rs911738990 Chr.20: 64200272-64200272 on Build GRCh38
rs201103523 Chr.20: 64199954-64199954 on Build GRCh38
rs542453881 Chr.20: 64200022-64200022 on Build GRCh38
rs527760844 Chr.20: 64200218-64200218 on Build GRCh38
rs2295452 Chr.20: 64200253-64200253 on Build GRCh38
rs961552239 Chr.20: 64200227-64200227 on Build GRCh38
rs774011894 Chr.20: 64199886-64199886 on Build GRCh38
rs780529595 Chr.20: 64199917-64199917 on Build GRCh38
rs1023619261 Chr.20: 64199943-64199943 on Build GRCh38
rs749300392 Chr.20: 64199965-64199965 on Build GRCh38
rs762958767 Chr.20: 64199879-64199879 on Build GRCh38
rs755781548 Chr.20: 64200163-64200163 on Build GRCh38
rs761343812 Chr.20: 64199861-64199861 on Build GRCh38
rs117718177 Chr.20: 64199877-64199877 on Build GRCh38
rs527635883 Chr.20: 64200133-64200133 on Build GRCh38
rs761081358 Chr.20: 64199887-64199887 on Build GRCh38
rs930401462 Chr.20: 64200097-64200097 on Build GRCh38
rs370240424 Chr.20: 64200138-64200138 on Build GRCh38
rs968419119 Chr.20: 64200016-64200016 on Build GRCh38
rs1035188674 Chr.20: 64200153-64200153 on Build GRCh38
rs2253857 Chr.20: 64200172-64200172 on Build GRCh38
rs921374623 Chr.20: 64200045-64200045 on Build GRCh38
rs1031047794 Chr.20: 64199979-64199979 on Build GRCh38
rs966283873 Chr.20: 64200236-64200236 on Build GRCh38
rs552425258 Chr.20: 64200232-64200232 on Build GRCh38
rs913905682 Chr.20: 64200003-64200003 on Build GRCh38
rs1036302128 Chr.20: 64200293-64200293 on Build GRCh38
rs951808858 Chr.20: 64200273-64200273 on Build GRCh38
rs191529672 Chr.20: 64200074-64200074 on Build GRCh38

View Full Product Details