Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 43024791-43025290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKNOX1
Entrez Gene ID
5316
Gene Name
PBX/knotted 1 homeobox 1
Gene Aliases
PREP1, pkonx1c
UniGene
Hs.431043
Gene Chromosome Location
Chr.21: 42974510-43033931 on Build GRCh38
Transcripts
NM_001286258.1 NM_001320694.1 NM_004571.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2820031 c.889G>A p.A297T Chr.21: 43024910-43024910 on Build GRCh38
4101831 c.857A>G p.Y286C Chr.21: 43024878-43024878 on Build GRCh38
5443249 c.912C>G p.L304L Chr.21: 43024933-43024933 on Build GRCh38
5816180 c.869A>T p.D290V Chr.21: 43024890-43024890 on Build GRCh38
4296513 c.889G>A p.A297T Chr.21: 43024910-43024910 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543578542 Chr.21: 43025076-43025076 on Build GRCh38
rs902135965 Chr.21: 43024841-43024841 on Build GRCh38
rs894242223 Chr.21: 43025113-43025113 on Build GRCh38
rs367706457 Chr.21: 43024981-43024981 on Build GRCh38
rs755996655 Chr.21: 43024814-43024814 on Build GRCh38
rs985387069 Chr.21: 43025240-43025240 on Build GRCh38
rs73906403 Chr.21: 43025049-43025049 on Build GRCh38
rs1016467449 Chr.21: 43025180-43025180 on Build GRCh38
rs771283980 Chr.21: 43024871-43024871 on Build GRCh38
rs902881447 Chr.21: 43025052-43025052 on Build GRCh38
rs771261316 Chr.21: 43024836-43024836 on Build GRCh38
rs955249350 Chr.21: 43025194-43025194 on Build GRCh38
rs747537686 Chr.21: 43024868-43024868 on Build GRCh38
rs947335891 Chr.21: 43025004-43025004 on Build GRCh38
rs532969633 Chr.21: 43025223-43025223 on Build GRCh38
rs748355411 Chr.21: 43024845-43024845 on Build GRCh38
rs993791590 Chr.21: 43025219-43025219 on Build GRCh38
rs373968439 Chr.21: 43024956-43024956 on Build GRCh38
rs111456054 Chr.21: 43024828-43024828 on Build GRCh38
rs234719 Chr.21: 43025075-43025075 on Build GRCh38
rs760602062 Chr.21: 43025106-43025106 on Build GRCh38
rs546026128 Chr.21: 43024857-43024857 on Build GRCh38
rs764229980 Chr.21: 43025159-43025159 on Build GRCh38
rs753797680 Chr.21: 43024829-43024829 on Build GRCh38
rs773915883 Chr.21: 43024955-43024955 on Build GRCh38
rs563834382 Chr.21: 43024985-43024985 on Build GRCh38
rs772935066 Chr.21: 43024984-43024984 on Build GRCh38
rs911023902 Chr.21: 43025241-43025241 on Build GRCh38
rs181025479 Chr.21: 43024986-43024986 on Build GRCh38
rs1013059138 Chr.21: 43025169-43025169 on Build GRCh38
rs778239127 Chr.21: 43024862-43024862 on Build GRCh38
rs779620444 Chr.21: 43024906-43024906 on Build GRCh38
rs770481809 Chr.21: 43024894-43024895 on Build GRCh38
rs545568820 Chr.21: 43024888-43024888 on Build GRCh38
rs768198993 Chr.21: 43024952-43024952 on Build GRCh38
rs578213121 Chr.21: 43024839-43024839 on Build GRCh38
rs117060078 Chr.21: 43025041-43025041 on Build GRCh38
rs368748944 Chr.21: 43024994-43024994 on Build GRCh38
rs748816719 Chr.21: 43024920-43024920 on Build GRCh38
rs946534648 Chr.21: 43025263-43025263 on Build GRCh38
rs1026759755 Chr.21: 43024825-43024825 on Build GRCh38
rs234720 Chr.21: 43025182-43025182 on Build GRCh38
rs766497858 Chr.21: 43024824-43024824 on Build GRCh38
rs148640127 Chr.21: 43025039-43025039 on Build GRCh38
rs772230089 Chr.21: 43024999-43024999 on Build GRCh38

View Full Product Details