Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 26540776-26541292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPST2
Entrez Gene ID
8459
Gene Name
tyrosylprotein sulfotransferase 2
Gene Aliases
TANGO13B
UniGene
Hs.632768
Gene Chromosome Location
Chr.22: 26524082-26590125 on Build GRCh38
Transcripts
NM_001008566.1 NM_003595.3 XM_006724338.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4834092 c.391G>A p.E131K Chr.22: 26541240-26541240 on Build GRCh38
6497740 c.693G>A p.E231E Chr.22: 26540938-26540938 on Build GRCh38
6807052 c.485C>T p.T162M Chr.22: 26541146-26541146 on Build GRCh38
5083071 c.353G>A p.R118H Chr.22: 26541278-26541278 on Build GRCh38
6807054 c.787G>A p.A263T Chr.22: 26540844-26540844 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758984700 Chr.22: 26541101-26541101 on Build GRCh38
rs200758682 Chr.22: 26541248-26541248 on Build GRCh38
rs370505161 Chr.22: 26541186-26541186 on Build GRCh38
rs747195748 Chr.22: 26540911-26540911 on Build GRCh38
rs781353266 Chr.22: 26541032-26541032 on Build GRCh38
rs745797442 Chr.22: 26540824-26540824 on Build GRCh38
rs781674333 Chr.22: 26540834-26540834 on Build GRCh38
rs550831870 Chr.22: 26540984-26540984 on Build GRCh38
rs758802800 Chr.22: 26540808-26540808 on Build GRCh38
rs5761587 Chr.22: 26541121-26541121 on Build GRCh38
rs767262747 Chr.22: 26541255-26541255 on Build GRCh38
rs760020260 Chr.22: 26540848-26540848 on Build GRCh38
rs371220641 Chr.22: 26540872-26540872 on Build GRCh38
rs776155338 Chr.22: 26541162-26541162 on Build GRCh38
rs778896187 Chr.22: 26541218-26541218 on Build GRCh38
rs780156467 Chr.22: 26540835-26540835 on Build GRCh38
rs940474636 Chr.22: 26541145-26541145 on Build GRCh38
rs769147490 Chr.22: 26541049-26541049 on Build GRCh38
rs369043628 Chr.22: 26541082-26541082 on Build GRCh38
rs140023376 Chr.22: 26540809-26540809 on Build GRCh38
rs773262198 Chr.22: 26540841-26540841 on Build GRCh38
rs771232915 Chr.22: 26541234-26541234 on Build GRCh38
rs148460389 Chr.22: 26540917-26540917 on Build GRCh38
rs776299836 Chr.22: 26541236-26541236 on Build GRCh38
rs754702298 Chr.22: 26540902-26540902 on Build GRCh38
rs762194832 Chr.22: 26541066-26541066 on Build GRCh38
rs749743940 Chr.22: 26541048-26541048 on Build GRCh38
rs988990085 Chr.22: 26540803-26540803 on Build GRCh38
rs183262802 Chr.22: 26540923-26540923 on Build GRCh38
rs762944926 Chr.22: 26541183-26541183 on Build GRCh38
rs780280233 Chr.22: 26541195-26541195 on Build GRCh38
rs776756599 Chr.22: 26540847-26540847 on Build GRCh38
rs763555643 Chr.22: 26540929-26540929 on Build GRCh38
rs1042029680 Chr.22: 26541268-26541268 on Build GRCh38
rs751141452 Chr.22: 26540871-26540871 on Build GRCh38
rs367946111 Chr.22: 26541063-26541063 on Build GRCh38
rs868105522 Chr.22: 26540843-26540843 on Build GRCh38
rs762095799 Chr.22: 26540956-26540956 on Build GRCh38
rs187643832 Chr.22: 26541229-26541229 on Build GRCh38
rs149746837 Chr.22: 26540862-26540862 on Build GRCh38
rs769834144 Chr.22: 26541043-26541043 on Build GRCh38
rs759049413 Chr.22: 26541171-26541171 on Build GRCh38
rs143783337 Chr.22: 26541266-26541266 on Build GRCh38
rs775980485 Chr.22: 26540857-26540857 on Build GRCh38
rs779424138 Chr.22: 26541128-26541128 on Build GRCh38
rs374367677 Chr.22: 26541012-26541012 on Build GRCh38
rs770123108 Chr.22: 26540856-26540856 on Build GRCh38
rs374069142 Chr.22: 26540817-26540817 on Build GRCh38
rs140525210 Chr.22: 26541232-26541232 on Build GRCh38
rs538398615 Chr.22: 26540823-26540823 on Build GRCh38
rs12169509 Chr.22: 26540931-26540931 on Build GRCh38
rs772767474 Chr.22: 26541228-26541228 on Build GRCh38
rs761996174 Chr.22: 26541036-26541036 on Build GRCh38
rs200350323 Chr.22: 26541031-26541031 on Build GRCh38
rs772530209 Chr.22: 26541135-26541135 on Build GRCh38
rs749274304 Chr.22: 26540899-26540899 on Build GRCh38
rs754057486 Chr.22: 26541256-26541256 on Build GRCh38
rs201685344 Chr.22: 26540935-26540935 on Build GRCh38
rs768530486 Chr.22: 26540839-26540839 on Build GRCh38
rs117701840 Chr.22: 26541173-26541173 on Build GRCh38
rs749896866 Chr.22: 26540882-26540882 on Build GRCh38
rs770653345 Chr.22: 26541146-26541146 on Build GRCh38
rs896000993 Chr.22: 26541269-26541269 on Build GRCh38
rs142098416 Chr.22: 26541078-26541078 on Build GRCh38
rs1041606752 Chr.22: 26541034-26541034 on Build GRCh38
rs145859999 Chr.22: 26541079-26541079 on Build GRCh38
rs764986870 Chr.22: 26541127-26541127 on Build GRCh38
rs370065051 Chr.22: 26541008-26541008 on Build GRCh38
rs538758533 Chr.22: 26540910-26540910 on Build GRCh38
rs201521913 Chr.22: 26540860-26540860 on Build GRCh38
rs750639533 Chr.22: 26541120-26541120 on Build GRCh38
rs767124818 Chr.22: 26540880-26540880 on Build GRCh38
rs201879530 Chr.22: 26540838-26540838 on Build GRCh38
rs753617880 Chr.22: 26541133-26541133 on Build GRCh38
rs753691008 Chr.22: 26540998-26540998 on Build GRCh38
rs750127856 Chr.22: 26540964-26540964 on Build GRCh38
rs752510173 Chr.22: 26541134-26541134 on Build GRCh38
rs758141984 Chr.22: 26541118-26541118 on Build GRCh38
rs765157243 Chr.22: 26541103-26541103 on Build GRCh38
rs371365617 Chr.22: 26541122-26541122 on Build GRCh38
rs921587084 Chr.22: 26541124-26541124 on Build GRCh38
rs765351148 Chr.22: 26541267-26541267 on Build GRCh38
rs780103356 Chr.22: 26540896-26540896 on Build GRCh38
rs1048426477 Chr.22: 26541220-26541220 on Build GRCh38
rs781403165 Chr.22: 26541125-26541125 on Build GRCh38
rs908057168 Chr.22: 26540908-26540908 on Build GRCh38
rs755633467 Chr.22: 26540965-26540965 on Build GRCh38
rs200871820 Chr.22: 26541083-26541083 on Build GRCh38
rs774835852 Chr.22: 26540938-26540938 on Build GRCh38
rs745883539 Chr.22: 26540920-26540920 on Build GRCh38
rs112497588 Chr.22: 26540962-26540962 on Build GRCh38
rs755059775 Chr.22: 26541215-26541215 on Build GRCh38
rs754025690 Chr.22: 26541198-26541198 on Build GRCh38
rs747921921 Chr.22: 26541015-26541015 on Build GRCh38
rs753448750 Chr.22: 26541093-26541093 on Build GRCh38
rs138925948 Chr.22: 26541249-26541249 on Build GRCh38
rs58274935 Chr.22: 26540800-26540800 on Build GRCh38
rs374214474 Chr.22: 26541223-26541223 on Build GRCh38
rs745557126 Chr.22: 26541241-26541241 on Build GRCh38
rs775612864 Chr.22: 26541045-26541045 on Build GRCh38
rs752609892 Chr.22: 26541115-26541115 on Build GRCh38
rs377669991 Chr.22: 26540844-26540844 on Build GRCh38
rs549915531 Chr.22: 26540796-26540796 on Build GRCh38
rs769912442 Chr.22: 26540831-26540831 on Build GRCh38
rs768953365 Chr.22: 26540916-26540916 on Build GRCh38
rs981597344 Chr.22: 26541193-26541193 on Build GRCh38
rs978932423 Chr.22: 26540819-26540819 on Build GRCh38
rs752659383 Chr.22: 26541178-26541178 on Build GRCh38
rs777594624 Chr.22: 26541119-26541119 on Build GRCh38
rs769317008 Chr.22: 26541244-26541244 on Build GRCh38
rs374640409 Chr.22: 26540977-26540977 on Build GRCh38
rs199686165 Chr.22: 26541067-26541067 on Build GRCh38
rs149409754 Chr.22: 26541253-26541253 on Build GRCh38
rs755012899 Chr.22: 26541001-26541001 on Build GRCh38
rs773784167 Chr.22: 26541142-26541142 on Build GRCh38
rs568213816 Chr.22: 26540845-26540845 on Build GRCh38
rs755692897 Chr.22: 26540888-26540888 on Build GRCh38
rs756700558 Chr.22: 26540812-26540812 on Build GRCh38
rs771245581 Chr.22: 26540912-26540912 on Build GRCh38
rs368243348 Chr.22: 26540892-26540892 on Build GRCh38
rs751295840 Chr.22: 26541187-26541187 on Build GRCh38
rs145597019 Chr.22: 26540863-26540863 on Build GRCh38

View Full Product Details