Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36516350-36516868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3D
Entrez Gene ID
8664
Gene Name
eukaryotic translation initiation factor 3 subunit D
Gene Aliases
EIF3S7, eIF3-p66, eIF3-zeta
UniGene
Hs.55682
Gene Chromosome Location
Chr.22: 36510850-36529431 on Build GRCh38
Transcripts
NM_003753.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5930100 c.1206+8G>T p.? Chr.22: 36516470-36516470 on Build GRCh38
4189060 c.1052A>G p.N351S Chr.22: 36516729-36516729 on Build GRCh38
1714339 c.1024C>T p.P342S Chr.22: 36516757-36516757 on Build GRCh38
6652812 c.1025C>T p.P342L Chr.22: 36516756-36516756 on Build GRCh38
1616419 c.1076+7C>A p.? Chr.22: 36516698-36516698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766992721 Chr.22: 36516756-36516756 on Build GRCh38
rs371499563 Chr.22: 36516546-36516546 on Build GRCh38
rs151303049 Chr.22: 36516591-36516591 on Build GRCh38
rs755487622 Chr.22: 36516699-36516699 on Build GRCh38
rs776668006 Chr.22: 36516656-36516656 on Build GRCh38
rs762899615 Chr.22: 36516742-36516742 on Build GRCh38
rs765079731 Chr.22: 36516662-36516662 on Build GRCh38
rs370408101 Chr.22: 36516814-36516814 on Build GRCh38
rs62230003 Chr.22: 36516391-36516391 on Build GRCh38
rs62230004 Chr.22: 36516401-36516401 on Build GRCh38
rs754331738 Chr.22: 36516696-36516696 on Build GRCh38
rs747026324 Chr.22: 36516716-36516716 on Build GRCh38
rs746314902 Chr.22: 36516457-36516457 on Build GRCh38
rs781040963 Chr.22: 36516553-36516553 on Build GRCh38
rs774038895 Chr.22: 36516840-36516840 on Build GRCh38
rs183188813 Chr.22: 36516722-36516722 on Build GRCh38
rs943701377 Chr.22: 36516728-36516728 on Build GRCh38
rs771073736 Chr.22: 36516813-36516813 on Build GRCh38
rs769050809 Chr.22: 36516618-36516618 on Build GRCh38
rs749729854 Chr.22: 36516471-36516471 on Build GRCh38
rs920321550 Chr.22: 36516641-36516641 on Build GRCh38
rs548651670 Chr.22: 36516453-36516453 on Build GRCh38
rs778101945 Chr.22: 36516715-36516715 on Build GRCh38
rs769845050 Chr.22: 36516828-36516828 on Build GRCh38
rs752360300 Chr.22: 36516547-36516547 on Build GRCh38
rs201479048 Chr.22: 36516806-36516806 on Build GRCh38
rs373088788 Chr.22: 36516433-36516433 on Build GRCh38
rs911007896 Chr.22: 36516727-36516727 on Build GRCh38
rs748577380 Chr.22: 36516710-36516710 on Build GRCh38
rs777383675 Chr.22: 36516745-36516745 on Build GRCh38
rs775618172 Chr.22: 36516829-36516829 on Build GRCh38
rs759493006 Chr.22: 36516729-36516729 on Build GRCh38
rs751020467 Chr.22: 36516679-36516679 on Build GRCh38
rs755472102 Chr.22: 36516767-36516767 on Build GRCh38
rs767535308 Chr.22: 36516517-36516517 on Build GRCh38
rs768458834 Chr.22: 36516837-36516837 on Build GRCh38
rs763632836 Chr.22: 36516743-36516743 on Build GRCh38
rs755713262 Chr.22: 36516821-36516824 on Build GRCh38
rs377061066 Chr.22: 36516470-36516470 on Build GRCh38
rs746015870 Chr.22: 36516824-36516824 on Build GRCh38
rs899970698 Chr.22: 36516468-36516468 on Build GRCh38
rs753557103 Chr.22: 36516543-36516543 on Build GRCh38
rs765935423 Chr.22: 36516825-36516826 on Build GRCh38
rs11558519 Chr.22: 36516583-36516583 on Build GRCh38
rs759574862 Chr.22: 36516658-36516658 on Build GRCh38
rs751267448 Chr.22: 36516751-36516751 on Build GRCh38
rs775425979 Chr.22: 36516737-36516737 on Build GRCh38
rs773353912 Chr.22: 36516523-36516523 on Build GRCh38
rs141517401 Chr.22: 36516511-36516511 on Build GRCh38
rs771000513 Chr.22: 36516721-36516721 on Build GRCh38
rs756840783 Chr.22: 36516688-36516688 on Build GRCh38
rs555447925 Chr.22: 36516844-36516844 on Build GRCh38
rs562285199 Chr.22: 36516827-36516827 on Build GRCh38
rs532212071 Chr.22: 36516622-36516622 on Build GRCh38
rs769863240 Chr.22: 36516736-36516736 on Build GRCh38
rs944270986 Chr.22: 36516417-36516417 on Build GRCh38
rs758833569 Chr.22: 36516782-36516782 on Build GRCh38
rs1022118796 Chr.22: 36516478-36516478 on Build GRCh38
rs746536790 Chr.22: 36516569-36516569 on Build GRCh38
rs943598744 Chr.22: 36516423-36516423 on Build GRCh38
rs778027509 Chr.22: 36516805-36516805 on Build GRCh38
rs955855839 Chr.22: 36516382-36516382 on Build GRCh38
rs761851541 Chr.22: 36516513-36516513 on Build GRCh38
rs772285279 Chr.22: 36516629-36516629 on Build GRCh38
rs778762531 Chr.22: 36516440-36516440 on Build GRCh38
rs929455560 Chr.22: 36516481-36516481 on Build GRCh38
rs753034103 Chr.22: 36516781-36516781 on Build GRCh38
rs2076083 Chr.22: 36516677-36516677 on Build GRCh38
rs202079294 Chr.22: 36516576-36516576 on Build GRCh38
rs774718957 Chr.22: 36516621-36516621 on Build GRCh38
rs773160724 Chr.22: 36516639-36516639 on Build GRCh38
rs371133317 Chr.22: 36516649-36516649 on Build GRCh38
rs547158688 Chr.22: 36516755-36516755 on Build GRCh38
rs117599987 Chr.22: 36516529-36516529 on Build GRCh38
rs999242162 Chr.22: 36516528-36516528 on Build GRCh38
rs140193656 Chr.22: 36516552-36516552 on Build GRCh38
rs200608758 Chr.22: 36516418-36516418 on Build GRCh38
rs138516680 Chr.22: 36516411-36516411 on Build GRCh38
rs749938618 Chr.22: 36516762-36516762 on Build GRCh38
rs955969753 Chr.22: 36516661-36516661 on Build GRCh38
rs754579373 Chr.22: 36516437-36516437 on Build GRCh38
rs780396237 Chr.22: 36516695-36516695 on Build GRCh38
rs758019848 Chr.22: 36516452-36516452 on Build GRCh38
rs754866318 Chr.22: 36516544-36516544 on Build GRCh38
rs770352309 Chr.22: 36516465-36516465 on Build GRCh38
rs779180489 Chr.22: 36516706-36516706 on Build GRCh38
rs201265305 Chr.22: 36516836-36516836 on Build GRCh38
rs767339874 Chr.22: 36516455-36516455 on Build GRCh38
rs752556419 Chr.22: 36516672-36516672 on Build GRCh38
rs929571707 Chr.22: 36516780-36516780 on Build GRCh38
rs16996838 Chr.22: 36516412-36516412 on Build GRCh38
rs768852788 Chr.22: 36516507-36516507 on Build GRCh38
rs764252752 Chr.22: 36516392-36516392 on Build GRCh38
rs376900932 Chr.22: 36516770-36516770 on Build GRCh38
rs748890507 Chr.22: 36516749-36516749 on Build GRCh38
rs750064458 Chr.22: 36516628-36516629 on Build GRCh38
rs762631466 Chr.22: 36516673-36516673 on Build GRCh38

View Full Product Details