Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39049201-39049698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC3F
Entrez Gene ID
200316
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3F
Gene Aliases
A3F, ARP8, BK150C2.4.MRNA, KA6
UniGene
Hs.659809
Gene Chromosome Location
Chr.22: 39020454-39055972 on Build GRCh38
Transcripts
NM_145298.5 XM_011529994.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4819322 c.689C>T p.P230L Chr.22: 39049547-39049547 on Build GRCh38
4662515 c.623G>A p.R208H Chr.22: 39049481-39049481 on Build GRCh38
159131 c.573G>A p.P191P Chr.22: 39049431-39049431 on Build GRCh38
140307 c.721C>T p.Q241* Chr.22: 39049579-39049579 on Build GRCh38
3151768 c.683A>G p.H228R Chr.22: 39049541-39049541 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753858317 Chr.22: 39049443-39049443 on Build GRCh38
rs769431557 Chr.22: 39049302-39049302 on Build GRCh38
rs767965103 Chr.22: 39049416-39049420 on Build GRCh38
rs2076101 Chr.22: 39049549-39049549 on Build GRCh38
rs770513913 Chr.22: 39049430-39049430 on Build GRCh38
rs759744454 Chr.22: 39049628-39049628 on Build GRCh38
rs2014881 Chr.22: 39049375-39049375 on Build GRCh38
rs771806377 Chr.22: 39049496-39049496 on Build GRCh38
rs369018699 Chr.22: 39049445-39049445 on Build GRCh38
rs780346083 Chr.22: 39049402-39049402 on Build GRCh38
rs372759662 Chr.22: 39049534-39049534 on Build GRCh38
rs768174453 Chr.22: 39049409-39049409 on Build GRCh38
rs376493009 Chr.22: 39049389-39049389 on Build GRCh38
rs1047466255 Chr.22: 39049582-39049582 on Build GRCh38
rs374667655 Chr.22: 39049467-39049467 on Build GRCh38
rs200231091 Chr.22: 39049485-39049485 on Build GRCh38
rs549550231 Chr.22: 39049545-39049545 on Build GRCh38
rs538526699 Chr.22: 39049664-39049664 on Build GRCh38
rs888472122 Chr.22: 39049598-39049598 on Build GRCh38
rs144119358 Chr.22: 39049310-39049310 on Build GRCh38
rs571232626 Chr.22: 39049663-39049663 on Build GRCh38
rs756069639 Chr.22: 39049376-39049376 on Build GRCh38
rs747325800 Chr.22: 39049464-39049464 on Build GRCh38
rs967666432 Chr.22: 39049658-39049658 on Build GRCh38
rs34578066 Chr.22: 39049551-39049551 on Build GRCh38
rs773937857 Chr.22: 39049490-39049490 on Build GRCh38
rs906825204 Chr.22: 39049563-39049563 on Build GRCh38
rs375405160 Chr.22: 39049442-39049442 on Build GRCh38
rs553003361 Chr.22: 39049221-39049221 on Build GRCh38
rs544624796 Chr.22: 39049256-39049256 on Build GRCh38
rs925733745 Chr.22: 39049253-39049253 on Build GRCh38
rs368249214 Chr.22: 39049390-39049390 on Build GRCh38
rs765418322 Chr.22: 39049446-39049446 on Build GRCh38
rs750873126 Chr.22: 39049534-39049557 on Build GRCh38
rs1029577811 Chr.22: 39049655-39049655 on Build GRCh38
rs760574777 Chr.22: 39049438-39049438 on Build GRCh38
rs746126402 Chr.22: 39049391-39049391 on Build GRCh38
rs369511676 Chr.22: 39049482-39049482 on Build GRCh38
rs750517144 Chr.22: 39049447-39049447 on Build GRCh38
rs540806428 Chr.22: 39049347-39049347 on Build GRCh38
rs1018399668 Chr.22: 39049652-39049652 on Build GRCh38
rs778451997 Chr.22: 39049548-39049548 on Build GRCh38
rs761754118 Chr.22: 39049501-39049501 on Build GRCh38
rs775354754 Chr.22: 39049434-39049434 on Build GRCh38
rs775299622 Chr.22: 39049497-39049497 on Build GRCh38
rs747403689 Chr.22: 39049412-39049412 on Build GRCh38
rs773754983 Chr.22: 39049629-39049629 on Build GRCh38
rs377406880 Chr.22: 39049555-39049555 on Build GRCh38
rs768209849 Chr.22: 39049562-39049562 on Build GRCh38
rs566918675 Chr.22: 39049567-39049567 on Build GRCh38
rs781519343 Chr.22: 39049481-39049481 on Build GRCh38
rs201034804 Chr.22: 39049444-39049444 on Build GRCh38
rs375133255 Chr.22: 39049351-39049351 on Build GRCh38
rs534531437 Chr.22: 39049573-39049573 on Build GRCh38
rs773182647 Chr.22: 39049586-39049586 on Build GRCh38
rs575027750 Chr.22: 39049270-39049270 on Build GRCh38
rs553102299 Chr.22: 39049615-39049615 on Build GRCh38
rs760360766 Chr.22: 39049498-39049498 on Build GRCh38
rs895322696 Chr.22: 39049259-39049259 on Build GRCh38
rs1005637334 Chr.22: 39049373-39049373 on Build GRCh38
rs374582155 Chr.22: 39049574-39049574 on Build GRCh38
rs189022097 Chr.22: 39049416-39049416 on Build GRCh38
rs745372879 Chr.22: 39049495-39049495 on Build GRCh38
rs767288632 Chr.22: 39049432-39049432 on Build GRCh38
rs758496258 Chr.22: 39049522-39049522 on Build GRCh38
rs760532357 Chr.22: 39049257-39049257 on Build GRCh38
rs765093100 Chr.22: 39049511-39049511 on Build GRCh38
rs963855393 Chr.22: 39049654-39049654 on Build GRCh38
rs766511374 Chr.22: 39049608-39049608 on Build GRCh38
rs994612767 Chr.22: 39049311-39049311 on Build GRCh38
rs764020546 Chr.22: 39049499-39049499 on Build GRCh38
rs551029584 Chr.22: 39049419-39049419 on Build GRCh38
rs33913356 Chr.22: 39049431-39049431 on Build GRCh38
rs866383625 Chr.22: 39049382-39049382 on Build GRCh38
rs201158254 Chr.22: 39049559-39049559 on Build GRCh38
rs758738330 Chr.22: 39049392-39049392 on Build GRCh38
rs373341511 Chr.22: 39049387-39049387 on Build GRCh38
rs562920885 Chr.22: 39049258-39049258 on Build GRCh38
rs780286886 Chr.22: 39049462-39049462 on Build GRCh38
rs748724922 Chr.22: 39049420-39049420 on Build GRCh38
rs577479783 Chr.22: 39049239-39049239 on Build GRCh38
rs776422590 Chr.22: 39049500-39049500 on Build GRCh38
rs370797211 Chr.22: 39049561-39049561 on Build GRCh38
rs755293573 Chr.22: 39049538-39049538 on Build GRCh38
rs112283226 Chr.22: 39049323-39049323 on Build GRCh38
rs767462293 Chr.22: 39049374-39049374 on Build GRCh38
rs372471972 Chr.22: 39049452-39049452 on Build GRCh38
rs202092816 Chr.22: 39049504-39049504 on Build GRCh38
rs753014868 Chr.22: 39049544-39049544 on Build GRCh38
rs755419532 Chr.22: 39049478-39049478 on Build GRCh38
rs1006020442 Chr.22: 39049646-39049646 on Build GRCh38
rs781662873 Chr.22: 39049539-39049539 on Build GRCh38
rs762988178 Chr.22: 39049590-39049590 on Build GRCh38
rs758554799 Chr.22: 39049460-39049460 on Build GRCh38
rs750464224 Chr.22: 39049518-39049518 on Build GRCh38
rs528680044 Chr.22: 39049326-39049326 on Build GRCh38
rs766566349 Chr.22: 39049529-39049529 on Build GRCh38

View Full Product Details