Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 43159118-43159607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPO
Entrez Gene ID
706
Gene Name
translocator protein
Gene Aliases
BPBS, BZRP, DBI, IBP, MBR, PBR, PBS, PKBS, PTBR, mDRC, pk18
UniGene
Hs.202
Gene Chromosome Location
Chr.22: 43151514-43163242 on Build GRCh38
Transcripts
NM_000714.5 NM_001256530.1 NM_001256531.1 NR_046308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6266207 c.61G>T p.V21L Chr.22: 43159299-43159299 on Build GRCh38
6299567 c.16G>T p.V6L Chr.22: 43159254-43159254 on Build GRCh38
4462028 c.122C>T p.S41L Chr.22: 43159360-43159360 on Build GRCh38
7245871 c.165G>T p.T55T Chr.22: 43159403-43159403 on Build GRCh38
6810687 c.8C>T p.P3L Chr.22: 43159246-43159246 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044995476 Chr.22: 43159541-43159541 on Build GRCh38
rs746005390 Chr.22: 43159539-43159539 on Build GRCh38
rs8141880 Chr.22: 43159584-43159584 on Build GRCh38
rs1047746677 Chr.22: 43159255-43159255 on Build GRCh38
rs940847568 Chr.22: 43159165-43159165 on Build GRCh38
rs765728030 Chr.22: 43159260-43159260 on Build GRCh38
rs1023579746 Chr.22: 43159319-43159319 on Build GRCh38
rs879002362 Chr.22: 43159423-43159424 on Build GRCh38
rs746605782 Chr.22: 43159163-43159163 on Build GRCh38
rs748778969 Chr.22: 43159303-43159303 on Build GRCh38
rs371632726 Chr.22: 43159446-43159446 on Build GRCh38
rs9333333 Chr.22: 43159529-43159529 on Build GRCh38
rs1039261487 Chr.22: 43159404-43159404 on Build GRCh38
rs868060402 Chr.22: 43159267-43159267 on Build GRCh38
rs1003504292 Chr.22: 43159575-43159575 on Build GRCh38
rs770801880 Chr.22: 43159307-43159307 on Build GRCh38
rs34963883 Chr.22: 43159574-43159575 on Build GRCh38
rs574309140 Chr.22: 43159576-43159576 on Build GRCh38
rs759332329 Chr.22: 43159195-43159195 on Build GRCh38
rs750349204 Chr.22: 43159283-43159283 on Build GRCh38
rs905118370 Chr.22: 43159569-43159569 on Build GRCh38
rs187866832 Chr.22: 43159279-43159279 on Build GRCh38
rs775741310 Chr.22: 43159201-43159201 on Build GRCh38
rs372615034 Chr.22: 43159280-43159280 on Build GRCh38
rs943162283 Chr.22: 43159169-43159169 on Build GRCh38
rs1014231414 Chr.22: 43159467-43159467 on Build GRCh38
rs751910716 Chr.22: 43159370-43159370 on Build GRCh38
rs1023655178 Chr.22: 43159137-43159137 on Build GRCh38
rs1048504922 Chr.22: 43159230-43159230 on Build GRCh38
rs942338615 Chr.22: 43159402-43159402 on Build GRCh38
rs867336465 Chr.22: 43159246-43159246 on Build GRCh38
rs771735915 Chr.22: 43159198-43159198 on Build GRCh38
rs568868318 Chr.22: 43159154-43159154 on Build GRCh38
rs1039152353 Chr.22: 43159183-43159183 on Build GRCh38
rs56135426 Chr.22: 43159437-43159437 on Build GRCh38
rs192201237 Chr.22: 43159403-43159403 on Build GRCh38
rs906108775 Chr.22: 43159513-43159513 on Build GRCh38
rs758845609 Chr.22: 43159276-43159276 on Build GRCh38
rs952337438 Chr.22: 43159341-43159341 on Build GRCh38
rs972264130 Chr.22: 43159173-43159173 on Build GRCh38
rs375039721 Chr.22: 43159343-43159343 on Build GRCh38
rs761915112 Chr.22: 43159340-43159340 on Build GRCh38
rs777431441 Chr.22: 43159299-43159299 on Build GRCh38
rs750994845 Chr.22: 43159360-43159360 on Build GRCh38
rs900940878 Chr.22: 43159289-43159289 on Build GRCh38
rs753398897 Chr.22: 43159430-43159430 on Build GRCh38
rs9333332 Chr.22: 43159331-43159331 on Build GRCh38
rs930926685 Chr.22: 43159227-43159227 on Build GRCh38
rs764229144 Chr.22: 43159243-43159243 on Build GRCh38
rs1033582672 Chr.22: 43159535-43159535 on Build GRCh38
rs778276873 Chr.22: 43159451-43159451 on Build GRCh38
rs930733534 Chr.22: 43159439-43159439 on Build GRCh38
rs763364246 Chr.22: 43159361-43159361 on Build GRCh38
rs913584046 Chr.22: 43159336-43159336 on Build GRCh38
rs749824230 Chr.22: 43159316-43159316 on Build GRCh38
rs760894949 Chr.22: 43159328-43159328 on Build GRCh38
rs778946143 Chr.22: 43159320-43159320 on Build GRCh38
rs910841261 Chr.22: 43159363-43159363 on Build GRCh38
rs376383649 Chr.22: 43159322-43159322 on Build GRCh38
rs550303992 Chr.22: 43159369-43159369 on Build GRCh38
rs1052644630 Chr.22: 43159504-43159504 on Build GRCh38
rs755760336 Chr.22: 43159298-43159298 on Build GRCh38
rs867721175 Chr.22: 43159265-43159265 on Build GRCh38
rs537752213 Chr.22: 43159429-43159429 on Build GRCh38
rs945283054 Chr.22: 43159147-43159147 on Build GRCh38
rs567132675 Chr.22: 43159266-43159266 on Build GRCh38
rs566580110 Chr.22: 43159332-43159332 on Build GRCh38
rs762240782 Chr.22: 43159259-43159259 on Build GRCh38
rs760878391 Chr.22: 43159224-43159224 on Build GRCh38
rs775263818 Chr.22: 43159327-43159327 on Build GRCh38
rs753849621 Chr.22: 43159251-43159251 on Build GRCh38
rs3937387 Chr.22: 43159188-43159188 on Build GRCh38
rs183593889 Chr.22: 43159153-43159153 on Build GRCh38

View Full Product Details