Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 43160964-43161468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPO
Entrez Gene ID
706
Gene Name
translocator protein
Gene Aliases
BPBS, BZRP, DBI, IBP, MBR, PBR, PBS, PKBS, PTBR, mDRC, pk18
UniGene
Hs.202
Gene Chromosome Location
Chr.22: 43151514-43163242 on Build GRCh38
Transcripts
NM_000714.5 NM_001256530.1 NM_001256531.1 NR_046308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5366566 c.5C>T p.A2V Chr.22: 43161155-43161155 on Build GRCh38
5950046 c.321+1_321+4delGTAA p.? Chr.22: 43161191-43161194 on Build GRCh38
6031622 c.286_287insC p.I98fs*>73 Chr.22: 43161155-43161156 on Build GRCh38
5366565 c.286C>T p.P96S Chr.22: 43161155-43161155 on Build GRCh38
7416624 c.289C>A p.P97T Chr.22: 43161158-43161158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373123834 Chr.22: 43161212-43161212 on Build GRCh38
rs777892439 Chr.22: 43161220-43161220 on Build GRCh38
rs896078930 Chr.22: 43161271-43161271 on Build GRCh38
rs781286079 Chr.22: 43161014-43161014 on Build GRCh38
rs765853866 Chr.22: 43161204-43161204 on Build GRCh38
rs373961705 Chr.22: 43161287-43161287 on Build GRCh38
rs759688508 Chr.22: 43161202-43161203 on Build GRCh38
rs142445069 Chr.22: 43161150-43161150 on Build GRCh38
rs746919529 Chr.22: 43161116-43161116 on Build GRCh38
rs776987614 Chr.22: 43161155-43161157 on Build GRCh38
rs949031213 Chr.22: 43161273-43161273 on Build GRCh38
rs1041489596 Chr.22: 43161401-43161401 on Build GRCh38
rs756386013 Chr.22: 43161219-43161219 on Build GRCh38
rs753588433 Chr.22: 43161061-43161061 on Build GRCh38
rs566547284 Chr.22: 43161056-43161056 on Build GRCh38
rs754840815 Chr.22: 43161206-43161206 on Build GRCh38
rs752385147 Chr.22: 43161214-43161214 on Build GRCh38
rs761543515 Chr.22: 43161179-43161179 on Build GRCh38
rs749336960 Chr.22: 43161154-43161154 on Build GRCh38
rs112658712 Chr.22: 43161193-43161193 on Build GRCh38
rs1015991137 Chr.22: 43161362-43161362 on Build GRCh38
rs544128172 Chr.22: 43161349-43161349 on Build GRCh38
rs749849080 Chr.22: 43161093-43161093 on Build GRCh38
rs8135638 Chr.22: 43161422-43161422 on Build GRCh38
rs1047408073 Chr.22: 43161284-43161284 on Build GRCh38
rs1020271436 Chr.22: 43161418-43161418 on Build GRCh38
rs968729748 Chr.22: 43161419-43161419 on Build GRCh38
rs574646673 Chr.22: 43161365-43161365 on Build GRCh38
rs760110771 Chr.22: 43161173-43161173 on Build GRCh38
rs181560394 Chr.22: 43161002-43161002 on Build GRCh38
rs770007222 Chr.22: 43161213-43161219 on Build GRCh38
rs771190871 Chr.22: 43161231-43161231 on Build GRCh38
rs756858058 Chr.22: 43161063-43161063 on Build GRCh38
rs762491836 Chr.22: 43161197-43161197 on Build GRCh38
rs763567442 Chr.22: 43161040-43161040 on Build GRCh38
rs758354864 Chr.22: 43161100-43161100 on Build GRCh38
rs754824182 Chr.22: 43161119-43161119 on Build GRCh38
rs376234950 Chr.22: 43161133-43161133 on Build GRCh38
rs890888020 Chr.22: 43161296-43161296 on Build GRCh38
rs878928844 Chr.22: 43161050-43161050 on Build GRCh38
rs1057387889 Chr.22: 43161250-43161250 on Build GRCh38
rs200728000 Chr.22: 43161020-43161020 on Build GRCh38
rs372235648 Chr.22: 43161102-43161102 on Build GRCh38
rs17003528 Chr.22: 43161379-43161379 on Build GRCh38
rs371088722 Chr.22: 43161366-43161366 on Build GRCh38
rs73418038 Chr.22: 43161357-43161357 on Build GRCh38
rs771101046 Chr.22: 43161157-43161157 on Build GRCh38
rs748303765 Chr.22: 43161036-43161036 on Build GRCh38
rs199899658 Chr.22: 43161079-43161079 on Build GRCh38
rs751563551 Chr.22: 43161205-43161205 on Build GRCh38
rs749497035 Chr.22: 43161230-43161230 on Build GRCh38
rs766940905 Chr.22: 43161049-43161049 on Build GRCh38
rs894315595 Chr.22: 43161022-43161022 on Build GRCh38
rs34936584 Chr.22: 43161045-43161046 on Build GRCh38
rs143915407 Chr.22: 43161186-43161186 on Build GRCh38
rs768899649 Chr.22: 43161028-43161028 on Build GRCh38
rs536354933 Chr.22: 43161037-43161037 on Build GRCh38
rs371895433 Chr.22: 43161001-43161001 on Build GRCh38
rs377098874 Chr.22: 43161124-43161124 on Build GRCh38
rs769743065 Chr.22: 43161196-43161196 on Build GRCh38
rs139234976 Chr.22: 43161057-43161057 on Build GRCh38
rs775654599 Chr.22: 43161177-43161177 on Build GRCh38
rs1010625534 Chr.22: 43161412-43161412 on Build GRCh38
rs938311016 Chr.22: 43161280-43161280 on Build GRCh38
rs954669511 Chr.22: 43161402-43161402 on Build GRCh38
rs771487582 Chr.22: 43161023-43161023 on Build GRCh38
rs368891259 Chr.22: 43161176-43161176 on Build GRCh38
rs746757983 Chr.22: 43161023-43161023 on Build GRCh38
rs775043588 Chr.22: 43161159-43161159 on Build GRCh38
rs965738132 Chr.22: 43161113-43161113 on Build GRCh38
rs769849973 Chr.22: 43161148-43161148 on Build GRCh38
rs776953146 Chr.22: 43161035-43161035 on Build GRCh38
rs943819288 Chr.22: 43161304-43161304 on Build GRCh38
rs73886413 Chr.22: 43161384-43161384 on Build GRCh38
rs908989218 Chr.22: 43161202-43161202 on Build GRCh38
rs927397418 Chr.22: 43161445-43161445 on Build GRCh38
rs376307141 Chr.22: 43161244-43161244 on Build GRCh38
rs143706367 Chr.22: 43161038-43161038 on Build GRCh38
rs937533744 Chr.22: 43161245-43161245 on Build GRCh38
rs774883859 Chr.22: 43161055-43161055 on Build GRCh38

View Full Product Details