Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 45345376-45345887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMC1B
Entrez Gene ID
27127
Gene Name
structural maintenance of chromosomes 1B
Gene Aliases
SMC1BETA, SMC1L2
UniGene
Hs.334176
Gene Chromosome Location
Chr.22: 45344063-45413671 on Build GRCh38
Transcripts
NM_001291501.1 NM_148674.4 XM_011530144.2 XM_011530145.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6221651 c.3581A>G p.D1194G Chr.22: 45345484-45345484 on Build GRCh38
5684167 c.3608C>T p.P1203L Chr.22: 45345463-45345463 on Build GRCh38
7310286 c.3520G>T p.E1174* Chr.22: 45345551-45345551 on Build GRCh38
3842850 c.3598G>C p.G1200R Chr.22: 45345473-45345473 on Build GRCh38
6221652 c.3587A>G p.D1196G Chr.22: 45345484-45345484 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572945287 Chr.22: 45345719-45345719 on Build GRCh38
rs1031890915 Chr.22: 45345713-45345713 on Build GRCh38
rs866091739 Chr.22: 45345714-45345714 on Build GRCh38
rs189944172 Chr.22: 45345844-45345844 on Build GRCh38
rs186666960 Chr.22: 45345660-45345660 on Build GRCh38
rs377608770 Chr.22: 45345487-45345487 on Build GRCh38
rs751741428 Chr.22: 45345523-45345523 on Build GRCh38
rs201216889 Chr.22: 45345471-45345471 on Build GRCh38
rs776159371 Chr.22: 45345609-45345609 on Build GRCh38
rs774161202 Chr.22: 45345442-45345442 on Build GRCh38
rs569024980 Chr.22: 45345567-45345567 on Build GRCh38
rs774966446 Chr.22: 45345591-45345591 on Build GRCh38
rs572780334 Chr.22: 45345816-45345817 on Build GRCh38
rs1020993457 Chr.22: 45345810-45345810 on Build GRCh38
rs5765260 Chr.22: 45345662-45345662 on Build GRCh38
rs749046928 Chr.22: 45345560-45345560 on Build GRCh38
rs757341596 Chr.22: 45345604-45345604 on Build GRCh38
rs1038666537 Chr.22: 45345665-45345665 on Build GRCh38
rs575185869 Chr.22: 45345637-45345637 on Build GRCh38
rs758989826 Chr.22: 45345446-45345446 on Build GRCh38
rs1032213088 Chr.22: 45345539-45345539 on Build GRCh38
rs767485898 Chr.22: 45345745-45345745 on Build GRCh38
rs898732639 Chr.22: 45345671-45345671 on Build GRCh38
rs969333637 Chr.22: 45345691-45345691 on Build GRCh38
rs1008493152 Chr.22: 45345482-45345482 on Build GRCh38
rs760298028 Chr.22: 45345470-45345470 on Build GRCh38
rs778862121 Chr.22: 45345581-45345581 on Build GRCh38
rs756265517 Chr.22: 45345805-45345805 on Build GRCh38
rs1038380362 Chr.22: 45345407-45345407 on Build GRCh38
rs750985241 Chr.22: 45345485-45345485 on Build GRCh38
rs949135305 Chr.22: 45345611-45345611 on Build GRCh38
rs771592397 Chr.22: 45345449-45345449 on Build GRCh38
rs9614646 Chr.22: 45345498-45345498 on Build GRCh38
rs267606272 Chr.22: 45345509-45345509 on Build GRCh38
rs897765567 Chr.22: 45345834-45345835 on Build GRCh38
rs764310972 Chr.22: 45345444-45345445 on Build GRCh38
rs530608032 Chr.22: 45345846-45345846 on Build GRCh38
rs763710587 Chr.22: 45345474-45345474 on Build GRCh38
rs745836832 Chr.22: 45345584-45345584 on Build GRCh38
rs118012587 Chr.22: 45345439-45345439 on Build GRCh38
rs781337680 Chr.22: 45345542-45345542 on Build GRCh38
rs1021696594 Chr.22: 45345681-45345681 on Build GRCh38
rs990482913 Chr.22: 45345580-45345580 on Build GRCh38
rs556788457 Chr.22: 45345585-45345586 on Build GRCh38
rs959327801 Chr.22: 45345576-45345576 on Build GRCh38
rs935030243 Chr.22: 45345401-45345401 on Build GRCh38
rs754223496 Chr.22: 45345479-45345479 on Build GRCh38
rs1008274655 Chr.22: 45345786-45345786 on Build GRCh38
rs768285206 Chr.22: 45345607-45345607 on Build GRCh38
rs922898451 Chr.22: 45345778-45345779 on Build GRCh38
rs369991859 Chr.22: 45345481-45345481 on Build GRCh38
rs9626312 Chr.22: 45345483-45345483 on Build GRCh38
rs1028852870 Chr.22: 45345480-45345480 on Build GRCh38
rs758508822 Chr.22: 45345486-45345486 on Build GRCh38
rs1036159465 Chr.22: 45345833-45345833 on Build GRCh38
rs749098001 Chr.22: 45345431-45345431 on Build GRCh38
rs56343022 Chr.22: 45345656-45345656 on Build GRCh38
rs117203058 Chr.22: 45345761-45345761 on Build GRCh38
rs775021378 Chr.22: 45345462-45345462 on Build GRCh38
rs61737925 Chr.22: 45345535-45345535 on Build GRCh38
rs552998946 Chr.22: 45345702-45345702 on Build GRCh38
rs565645951 Chr.22: 45345816-45345816 on Build GRCh38
rs537981348 Chr.22: 45345588-45345588 on Build GRCh38
rs9614647 Chr.22: 45345837-45345837 on Build GRCh38
rs975684027 Chr.22: 45345430-45345430 on Build GRCh38
rs866607530 Chr.22: 45345820-45345820 on Build GRCh38
rs760243039 Chr.22: 45345600-45345600 on Build GRCh38

View Full Product Details