Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14666725-14667233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC174
Entrez Gene ID
51244
Gene Name
coiled-coil domain containing 174
Gene Aliases
C3orf19, HSPC212, IHPM, IHPMR
UniGene
Hs.517820
Gene Chromosome Location
Chr.3: 14651683-14673186 on Build GRCh38
Transcripts
NM_016474.4 NR_135523.1 XM_017006554.1 XM_017006555.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4890244 c.647A>G p.Q216R Chr.3: 14666870-14666870 on Build GRCh38
5883746 c.663A>C p.E221D Chr.3: 14666886-14666886 on Build GRCh38
4909697 c.661G>A p.E221K Chr.3: 14666884-14666884 on Build GRCh38
5911842 c.695C>T p.P232L Chr.3: 14666918-14666918 on Build GRCh38
4114256 c.628A>G p.R210G Chr.3: 14666851-14666851 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139455452 Chr.3: 14667045-14667045 on Build GRCh38
rs781264205 Chr.3: 14666866-14666866 on Build GRCh38
rs537882704 Chr.3: 14666749-14666749 on Build GRCh38
rs1031864036 Chr.3: 14667001-14667001 on Build GRCh38
rs754397151 Chr.3: 14666956-14666956 on Build GRCh38
rs756167959 Chr.3: 14666948-14666948 on Build GRCh38
rs762777201 Chr.3: 14666845-14666845 on Build GRCh38
rs779345468 Chr.3: 14666962-14666962 on Build GRCh38
rs774272502 Chr.3: 14667028-14667028 on Build GRCh38
rs776338301 Chr.3: 14666912-14666912 on Build GRCh38
rs371601155 Chr.3: 14666968-14666968 on Build GRCh38
rs938519069 Chr.3: 14667136-14667136 on Build GRCh38
rs368146701 Chr.3: 14666916-14666916 on Build GRCh38
rs989826235 Chr.3: 14666990-14666990 on Build GRCh38
rs907985263 Chr.3: 14667158-14667158 on Build GRCh38
rs141772230 Chr.3: 14666972-14666972 on Build GRCh38
rs945712096 Chr.3: 14666750-14666750 on Build GRCh38
rs779104848 Chr.3: 14666880-14666880 on Build GRCh38
rs145478230 Chr.3: 14666920-14666920 on Build GRCh38
rs769461395 Chr.3: 14666988-14666988 on Build GRCh38
rs547951852 Chr.3: 14667137-14667137 on Build GRCh38
rs201236853 Chr.3: 14666772-14666772 on Build GRCh38
rs770760978 Chr.3: 14667024-14667024 on Build GRCh38
rs746764326 Chr.3: 14666977-14666978 on Build GRCh38
rs756846641 Chr.3: 14666890-14666891 on Build GRCh38
rs776701189 Chr.3: 14666831-14666831 on Build GRCh38
rs972041963 Chr.3: 14666941-14666941 on Build GRCh38
rs779844959 Chr.3: 14666976-14666977 on Build GRCh38
rs759379648 Chr.3: 14666900-14666900 on Build GRCh38
rs766201512 Chr.3: 14666927-14666927 on Build GRCh38
rs886421328 Chr.3: 14666809-14666809 on Build GRCh38
rs755125953 Chr.3: 14666863-14666863 on Build GRCh38
rs145512037 Chr.3: 14666872-14666872 on Build GRCh38
rs372914503 Chr.3: 14667096-14667096 on Build GRCh38
rs759815667 Chr.3: 14666935-14666935 on Build GRCh38
rs559778485 Chr.3: 14666819-14666819 on Build GRCh38
rs1038068225 Chr.3: 14667178-14667178 on Build GRCh38
rs532374267 Chr.3: 14667204-14667204 on Build GRCh38
rs888916642 Chr.3: 14667171-14667171 on Build GRCh38
rs746673913 Chr.3: 14666890-14666890 on Build GRCh38
rs759300396 Chr.3: 14667031-14667031 on Build GRCh38
rs970343761 Chr.3: 14667073-14667073 on Build GRCh38
rs545986135 Chr.3: 14666919-14666919 on Build GRCh38
rs983521504 Chr.3: 14667155-14667155 on Build GRCh38
rs570953092 Chr.3: 14667194-14667194 on Build GRCh38
rs771345868 Chr.3: 14666879-14666879 on Build GRCh38
rs980026298 Chr.3: 14667083-14667083 on Build GRCh38
rs565960157 Chr.3: 14666751-14666751 on Build GRCh38
rs139500358 Chr.3: 14666913-14666913 on Build GRCh38
rs754274911 Chr.3: 14666843-14666844 on Build GRCh38
rs369355501 Chr.3: 14666998-14666998 on Build GRCh38
rs775183407 Chr.3: 14666806-14666806 on Build GRCh38
rs773960569 Chr.3: 14666932-14666932 on Build GRCh38
rs1037947988 Chr.3: 14666855-14666855 on Build GRCh38
rs778172418 Chr.3: 14666766-14666766 on Build GRCh38
rs766041003 Chr.3: 14666847-14666847 on Build GRCh38
rs368563387 Chr.3: 14666788-14666788 on Build GRCh38
rs201598636 Chr.3: 14666764-14666764 on Build GRCh38
rs778314409 Chr.3: 14666875-14666875 on Build GRCh38
rs765253479 Chr.3: 14666835-14666835 on Build GRCh38
rs745524046 Chr.3: 14666974-14666975 on Build GRCh38
rs140126783 Chr.3: 14666832-14666832 on Build GRCh38
rs942040395 Chr.3: 14667160-14667160 on Build GRCh38
rs750249439 Chr.3: 14666836-14666836 on Build GRCh38
rs150513750 Chr.3: 14666987-14666987 on Build GRCh38
rs764027935 Chr.3: 14666954-14666954 on Build GRCh38
rs370223037 Chr.3: 14666867-14666867 on Build GRCh38
rs539944364 Chr.3: 14666818-14666818 on Build GRCh38
rs778278901 Chr.3: 14666946-14666947 on Build GRCh38
rs768316120 Chr.3: 14666979-14666979 on Build GRCh38
rs751840479 Chr.3: 14666849-14666849 on Build GRCh38
rs748659171 Chr.3: 14666762-14666762 on Build GRCh38
rs749666357 Chr.3: 14666877-14666877 on Build GRCh38
rs767725627 Chr.3: 14666938-14666938 on Build GRCh38
rs752817344 Chr.3: 14666944-14666944 on Build GRCh38
rs900509506 Chr.3: 14667205-14667205 on Build GRCh38
rs749262067 Chr.3: 14667009-14667009 on Build GRCh38
rs757634142 Chr.3: 14666958-14666958 on Build GRCh38

View Full Product Details