Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38555578-38556066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCN5A
Entrez Gene ID
6331
Gene Name
sodium voltage-gated channel alpha subunit 5
Gene Aliases
CDCD2, CMD1E, CMPD2, HB1, HB2, HBBD, HH1, ICCD, IVF, LQT3, Nav1.5, PFHB1, SSS1, VF1
UniGene
Hs.517898
Gene Chromosome Location
Chr.3: 38548061-38649673 on Build GRCh38
Transcripts
NM_000335.4 NM_001099404.1 NM_001099405.1 NM_001160160.1 NM_001160161.1 NM_198056.2 XM_011533991.2 XM_017007017.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5612079 c.4501C>T p.L1501L Chr.3: 38555697-38555697 on Build GRCh38
4917663 c.4445G>T p.G1482V Chr.3: 38555753-38555753 on Build GRCh38
1317120 c.4534C>T p.R1512W Chr.3: 38555664-38555664 on Build GRCh38
4495420 c.4539C>T p.P1513P Chr.3: 38555659-38555659 on Build GRCh38
5892914 c.4535G>A p.R1512Q Chr.3: 38555663-38555663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372002687 Chr.3: 38555668-38555668 on Build GRCh38
rs45548237 Chr.3: 38555689-38555689 on Build GRCh38
rs199473261 Chr.3: 38555718-38555718 on Build GRCh38
rs794728944 Chr.3: 38555726-38555726 on Build GRCh38
rs561231889 Chr.3: 38556040-38556040 on Build GRCh38
rs916556138 Chr.3: 38555993-38555993 on Build GRCh38
rs199473615 Chr.3: 38555742-38555742 on Build GRCh38
rs1028724324 Chr.3: 38555816-38555816 on Build GRCh38
rs368219299 Chr.3: 38555663-38555663 on Build GRCh38
rs746549884 Chr.3: 38555656-38555656 on Build GRCh38
rs199473616 Chr.3: 38555731-38555731 on Build GRCh38
rs549476218 Chr.3: 38555852-38555852 on Build GRCh38
rs567677951 Chr.3: 38555823-38555823 on Build GRCh38
rs868516592 Chr.3: 38555688-38555688 on Build GRCh38
rs794728886 Chr.3: 38555745-38555745 on Build GRCh38
rs888246311 Chr.3: 38555712-38555712 on Build GRCh38
rs900899636 Chr.3: 38555781-38555781 on Build GRCh38
rs199473265 Chr.3: 38555698-38555698 on Build GRCh38
rs909160173 Chr.3: 38555597-38555597 on Build GRCh38
rs779664400 Chr.3: 38555752-38555752 on Build GRCh38
rs953121664 Chr.3: 38555853-38555853 on Build GRCh38
rs1021763094 Chr.3: 38555955-38555955 on Build GRCh38
rs369245812 Chr.3: 38555746-38555746 on Build GRCh38
rs763373788 Chr.3: 38555673-38555673 on Build GRCh38
rs199473262 Chr.3: 38555714-38555714 on Build GRCh38
rs199473259 Chr.3: 38555735-38555735 on Build GRCh38
rs747594441 Chr.3: 38555770-38555770 on Build GRCh38
rs771686849 Chr.3: 38555634-38555634 on Build GRCh38
rs992882881 Chr.3: 38556022-38556022 on Build GRCh38
rs960392208 Chr.3: 38556009-38556009 on Build GRCh38
rs751787337 Chr.3: 38555686-38555686 on Build GRCh38
rs556875875 Chr.3: 38555999-38556000 on Build GRCh38
rs770359277 Chr.3: 38555939-38555939 on Build GRCh38
rs397514251 Chr.3: 38555679-38555687 on Build GRCh38
rs767968814 Chr.3: 38556008-38556008 on Build GRCh38
rs773829118 Chr.3: 38555792-38555792 on Build GRCh38
rs768122161 Chr.3: 38555773-38555773 on Build GRCh38
rs187180126 Chr.3: 38555936-38555936 on Build GRCh38
rs766554531 Chr.3: 38555681-38555681 on Build GRCh38
rs745648618 Chr.3: 38555633-38555633 on Build GRCh38
rs867527896 Chr.3: 38555646-38555646 on Build GRCh38
rs9825620 Chr.3: 38555854-38555854 on Build GRCh38
rs199473258 Chr.3: 38555739-38555739 on Build GRCh38
rs1016528553 Chr.3: 38555923-38555923 on Build GRCh38
rs199473260 Chr.3: 38555720-38555720 on Build GRCh38
rs199473257 Chr.3: 38555756-38555756 on Build GRCh38
rs767615184 Chr.3: 38555695-38555695 on Build GRCh38
rs199473266 Chr.3: 38555697-38555697 on Build GRCh38
rs368998090 Chr.3: 38555788-38555788 on Build GRCh38
rs375026424 Chr.3: 38555728-38555728 on Build GRCh38
rs779725328 Chr.3: 38555636-38555636 on Build GRCh38
rs41315503 Chr.3: 38555989-38555989 on Build GRCh38
rs199473342 Chr.3: 38555691-38555691 on Build GRCh38
rs191637035 Chr.3: 38555940-38555940 on Build GRCh38
rs567393423 Chr.3: 38555829-38555829 on Build GRCh38
rs961863224 Chr.3: 38555927-38555930 on Build GRCh38
rs137854602 Chr.3: 38555664-38555664 on Build GRCh38
rs372874010 Chr.3: 38555662-38555662 on Build GRCh38
rs544520508 Chr.3: 38556004-38556004 on Build GRCh38
rs758190533 Chr.3: 38555749-38555749 on Build GRCh38
rs199473263 Chr.3: 38555705-38555705 on Build GRCh38
rs794728887 Chr.3: 38555736-38555736 on Build GRCh38
rs794728923 Chr.3: 38555671-38555679 on Build GRCh38
rs199473264 Chr.3: 38555706-38555706 on Build GRCh38
rs939387864 Chr.3: 38555978-38555978 on Build GRCh38
rs557263322 Chr.3: 38555777-38555777 on Build GRCh38
rs77290410 Chr.3: 38556005-38556005 on Build GRCh38
rs182624016 Chr.3: 38555981-38555981 on Build GRCh38
rs200695594 Chr.3: 38555618-38555618 on Build GRCh38
rs869025522 Chr.3: 38555719-38555721 on Build GRCh38
rs199473267 Chr.3: 38555694-38555694 on Build GRCh38
rs369404890 Chr.3: 38555671-38555671 on Build GRCh38
rs199473268 Chr.3: 38555683-38555683 on Build GRCh38
rs535211542 Chr.3: 38555930-38555930 on Build GRCh38

View Full Product Details