Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46375982-46376477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR5
Entrez Gene ID
1234
Gene Name
C-C motif chemokine receptor 5 (gene/pseudogene)
Gene Aliases
CC-CKR-5, CCCKR5, CCR-5, CD195, CKR-5, CKR5, CMKBR5, IDDM22
UniGene
Hs.450802
Gene Chromosome Location
Chr.3: 46370142-46376206 on Build GRCh38
Transcripts
NM_000579.3 NM_001100168.1 NR_125406.1

Gene Symbol
LOC102724297
Entrez Gene ID
102724297
Gene Name
uncharacterized LOC102724297
Gene Aliases
-
UniGene
Hs.622021
Gene Chromosome Location
Chr.3: 46364660-46407059 on Build GRCh38
Transcripts
NM_000579.3 NM_001100168.1 NR_125406.1

SNP Detail

SNP ID Chromosome Location
rs370386700 Chr.3: 46376007-46376007 on Build GRCh38
rs1015601952 Chr.3: 46376230-46376230 on Build GRCh38
rs756987625 Chr.3: 46376023-46376023 on Build GRCh38
rs745454829 Chr.3: 46376215-46376215 on Build GRCh38
rs919817302 Chr.3: 46376300-46376303 on Build GRCh38
rs572814964 Chr.3: 46376403-46376403 on Build GRCh38
rs890684227 Chr.3: 46376144-46376144 on Build GRCh38
rs917724862 Chr.3: 46376375-46376375 on Build GRCh38
rs1042525719 Chr.3: 46376384-46376384 on Build GRCh38
rs374785441 Chr.3: 46376079-46376079 on Build GRCh38
rs997004635 Chr.3: 46376091-46376091 on Build GRCh38
rs990625315 Chr.3: 46376351-46376351 on Build GRCh38
rs541816244 Chr.3: 46376185-46376185 on Build GRCh38
rs959225030 Chr.3: 46376320-46376320 on Build GRCh38
rs912262083 Chr.3: 46376058-46376058 on Build GRCh38
rs924365789 Chr.3: 46376068-46376068 on Build GRCh38
rs943639762 Chr.3: 46376166-46376166 on Build GRCh38
rs994930270 Chr.3: 46376258-46376258 on Build GRCh38
rs779659274 Chr.3: 46376327-46376327 on Build GRCh38
rs188577829 Chr.3: 46376034-46376034 on Build GRCh38
rs893330800 Chr.3: 46376065-46376065 on Build GRCh38
rs570463527 Chr.3: 46376018-46376018 on Build GRCh38
rs559231171 Chr.3: 46376370-46376370 on Build GRCh38
rs535314315 Chr.3: 46376013-46376013 on Build GRCh38
rs1022622443 Chr.3: 46376444-46376444 on Build GRCh38
rs1008277574 Chr.3: 46376217-46376217 on Build GRCh38
rs955460524 Chr.3: 46376185-46376186 on Build GRCh38
rs1004022890 Chr.3: 46376340-46376340 on Build GRCh38
rs539027881 Chr.3: 46376240-46376240 on Build GRCh38
rs142076881 Chr.3: 46376377-46376377 on Build GRCh38
rs1051893584 Chr.3: 46376113-46376113 on Build GRCh38
rs978516613 Chr.3: 46376047-46376047 on Build GRCh38
rs923672058 Chr.3: 46376387-46376387 on Build GRCh38
rs934795954 Chr.3: 46376070-46376070 on Build GRCh38
rs959966350 Chr.3: 46376398-46376398 on Build GRCh38
rs150855419 Chr.3: 46376455-46376455 on Build GRCh38
rs771492567 Chr.3: 46376277-46376277 on Build GRCh38

View Full Product Details