Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49929517-49930005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MON1A
Entrez Gene ID
84315
Gene Name
MON1 homolog A, secretory trafficking associated
Gene Aliases
SAND1
UniGene
Hs.655014
Gene Chromosome Location
Chr.3: 49908869-49930012 on Build GRCh38
Transcripts
NM_001142501.1 NM_032355.3 XM_006713345.3

SNP Detail

SNP ID Chromosome Location
rs896797187 Chr.3: 49929608-49929608 on Build GRCh38
rs955519597 Chr.3: 49929956-49929956 on Build GRCh38
rs1026560828 Chr.3: 49929621-49929621 on Build GRCh38
rs889501107 Chr.3: 49929555-49929555 on Build GRCh38
rs563561715 Chr.3: 49929677-49929677 on Build GRCh38
rs760374377 Chr.3: 49929859-49929859 on Build GRCh38
rs763566298 Chr.3: 49929925-49929925 on Build GRCh38
rs770907124 Chr.3: 49929638-49929638 on Build GRCh38
rs574804045 Chr.3: 49929671-49929671 on Build GRCh38
rs1022451091 Chr.3: 49929879-49929879 on Build GRCh38
rs373514881 Chr.3: 49929626-49929626 on Build GRCh38
rs989763824 Chr.3: 49929643-49929643 on Build GRCh38
rs1036179011 Chr.3: 49929952-49929952 on Build GRCh38
rs775695509 Chr.3: 49929575-49929576 on Build GRCh38
rs1023161478 Chr.3: 49929924-49929924 on Build GRCh38
rs755223623 Chr.3: 49929750-49929750 on Build GRCh38
rs542293932 Chr.3: 49929672-49929672 on Build GRCh38
rs978113547 Chr.3: 49929682-49929682 on Build GRCh38
rs756080268 Chr.3: 49929789-49929789 on Build GRCh38
rs114918681 Chr.3: 49929537-49929537 on Build GRCh38
rs751039250 Chr.3: 49929588-49929588 on Build GRCh38
rs781340975 Chr.3: 49929752-49929752 on Build GRCh38
rs1037637748 Chr.3: 49929799-49929799 on Build GRCh38
rs1040692385 Chr.3: 49929607-49929607 on Build GRCh38
rs767241550 Chr.3: 49929670-49929670 on Build GRCh38
rs1002990865 Chr.3: 49929945-49929945 on Build GRCh38
rs769792471 Chr.3: 49929622-49929622 on Build GRCh38
rs1025924154 Chr.3: 49929560-49929560 on Build GRCh38
rs535089749 Chr.3: 49929942-49929942 on Build GRCh38
rs774268559 Chr.3: 49929639-49929639 on Build GRCh38
rs749245453 Chr.3: 49929635-49929635 on Build GRCh38
rs770963899 Chr.3: 49929800-49929800 on Build GRCh38
rs371051417 Chr.3: 49929811-49929811 on Build GRCh38
rs564730205 Chr.3: 49929797-49929797 on Build GRCh38
rs745613303 Chr.3: 49929825-49929825 on Build GRCh38
rs1011519135 Chr.3: 49929873-49929873 on Build GRCh38
rs993372315 Chr.3: 49929806-49929806 on Build GRCh38
rs112513448 Chr.3: 49929805-49929805 on Build GRCh38
rs993096579 Chr.3: 49929550-49929550 on Build GRCh38
rs1006412752 Chr.3: 49929886-49929886 on Build GRCh38
rs759303301 Chr.3: 49929650-49929650 on Build GRCh38
rs906174823 Chr.3: 49929829-49929829 on Build GRCh38
rs966991907 Chr.3: 49929906-49929906 on Build GRCh38
rs970489854 Chr.3: 49929566-49929566 on Build GRCh38
rs867689487 Chr.3: 49929853-49929853 on Build GRCh38
rs748272365 Chr.3: 49929760-49929760 on Build GRCh38
rs781288342 Chr.3: 49929580-49929580 on Build GRCh38
rs530925784 Chr.3: 49929707-49929707 on Build GRCh38
rs755166801 Chr.3: 49929562-49929562 on Build GRCh38
rs1025969814 Chr.3: 49929827-49929827 on Build GRCh38
rs943173689 Chr.3: 49929770-49929770 on Build GRCh38
rs546853089 Chr.3: 49929832-49929832 on Build GRCh38
rs761122218 Chr.3: 49929923-49929923 on Build GRCh38
rs985581880 Chr.3: 49929982-49929982 on Build GRCh38
rs113960203 Chr.3: 49929850-49929850 on Build GRCh38
rs896853532 Chr.3: 49929839-49929839 on Build GRCh38
rs866564918 Chr.3: 49929861-49929861 on Build GRCh38
rs922597513 Chr.3: 49929684-49929684 on Build GRCh38
rs780018576 Chr.3: 49929866-49929866 on Build GRCh38
rs766776924 Chr.3: 49929712-49929712 on Build GRCh38
rs982259515 Chr.3: 49929620-49929620 on Build GRCh38
rs553572587 Chr.3: 49929728-49929728 on Build GRCh38
rs566578324 Chr.3: 49929683-49929683 on Build GRCh38
rs1048016288 Chr.3: 49929852-49929852 on Build GRCh38
rs888958238 Chr.3: 49929769-49929769 on Build GRCh38
rs906011280 Chr.3: 49929936-49929936 on Build GRCh38
rs988740532 Chr.3: 49929959-49929959 on Build GRCh38
rs928680314 Chr.3: 49929744-49929744 on Build GRCh38
rs922607673 Chr.3: 49929937-49929937 on Build GRCh38

View Full Product Details