Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50611765-50612265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CISH
Entrez Gene ID
1154
Gene Name
cytokine inducible SH2 containing protein
Gene Aliases
BACTS2, CIS, CIS-1, G18, SOCS
UniGene
Hs.655334
Gene Chromosome Location
Chr.3: 50606454-50611831 on Build GRCh38
Transcripts
NM_013324.5 NM_145071.2 NM_001243926.1

Gene Symbol
MAPKAPK3
Entrez Gene ID
7867
Gene Name
mitogen-activated protein kinase-activated protein kinase 3
Gene Aliases
3PK, MAPKAP-K3, MAPKAP3, MAPKAPK-3, MK-3
UniGene
Hs.234521
Gene Chromosome Location
Chr.3: 50611862-50649297 on Build GRCh38
Transcripts
NM_013324.5 NM_145071.2 NM_001243926.1

SNP Detail

SNP ID Chromosome Location
rs923116708 Chr.3: 50612195-50612195 on Build GRCh38
rs961479287 Chr.3: 50612123-50612123 on Build GRCh38
rs1043351561 Chr.3: 50612109-50612109 on Build GRCh38
rs534334599 Chr.3: 50612024-50612024 on Build GRCh38
rs901696207 Chr.3: 50612071-50612071 on Build GRCh38
rs1029325601 Chr.3: 50612099-50612099 on Build GRCh38
rs536988377 Chr.3: 50611795-50611795 on Build GRCh38
rs986271739 Chr.3: 50612197-50612197 on Build GRCh38
rs866509149 Chr.3: 50611947-50611947 on Build GRCh38
rs542332876 Chr.3: 50611946-50611946 on Build GRCh38
rs917489605 Chr.3: 50611966-50611966 on Build GRCh38
rs927575905 Chr.3: 50612163-50612163 on Build GRCh38
rs999270829 Chr.3: 50611895-50611895 on Build GRCh38
rs6768300 Chr.3: 50611939-50611939 on Build GRCh38
rs940557132 Chr.3: 50612034-50612034 on Build GRCh38
rs391511 Chr.3: 50611943-50611943 on Build GRCh38
rs996067416 Chr.3: 50611882-50611882 on Build GRCh38
rs933420841 Chr.3: 50612067-50612067 on Build GRCh38
rs971564787 Chr.3: 50612145-50612145 on Build GRCh38
rs546321484 Chr.3: 50611810-50611810 on Build GRCh38
rs370153991 Chr.3: 50612240-50612240 on Build GRCh38
rs999050317 Chr.3: 50612117-50612117 on Build GRCh38
rs909031817 Chr.3: 50612031-50612031 on Build GRCh38
rs961532795 Chr.3: 50611945-50611945 on Build GRCh38
rs950257713 Chr.3: 50612004-50612004 on Build GRCh38
rs1054208321 Chr.3: 50612027-50612027 on Build GRCh38
rs542852951 Chr.3: 50612122-50612122 on Build GRCh38
rs576572466 Chr.3: 50612238-50612238 on Build GRCh38
rs1010058658 Chr.3: 50611827-50611827 on Build GRCh38
rs759335042 Chr.3: 50611794-50611794 on Build GRCh38
rs972832028 Chr.3: 50612140-50612140 on Build GRCh38
rs1035880153 Chr.3: 50612120-50612120 on Build GRCh38
rs926945831 Chr.3: 50612015-50612015 on Build GRCh38
rs866802776 Chr.3: 50611955-50611955 on Build GRCh38
rs527696165 Chr.3: 50612180-50612180 on Build GRCh38
rs561572321 Chr.3: 50612181-50612181 on Build GRCh38
rs954190701 Chr.3: 50611931-50611931 on Build GRCh38
rs763801137 Chr.3: 50612002-50612002 on Build GRCh38
rs999920700 Chr.3: 50611787-50611787 on Build GRCh38
rs889125258 Chr.3: 50612070-50612070 on Build GRCh38
rs1022174453 Chr.3: 50611890-50611890 on Build GRCh38
rs991793213 Chr.3: 50612002-50612002 on Build GRCh38
rs945865159 Chr.3: 50612059-50612059 on Build GRCh38
rs965957144 Chr.3: 50611832-50611832 on Build GRCh38
rs980341807 Chr.3: 50612154-50612154 on Build GRCh38
rs414171 Chr.3: 50612068-50612068 on Build GRCh38
rs985100064 Chr.3: 50612167-50612167 on Build GRCh38
rs1011840854 Chr.3: 50611824-50611824 on Build GRCh38
rs1006002676 Chr.3: 50612107-50612107 on Build GRCh38
rs1024451358 Chr.3: 50612141-50612141 on Build GRCh38
rs562779385 Chr.3: 50611933-50611933 on Build GRCh38
rs996013666 Chr.3: 50612098-50612098 on Build GRCh38
rs990245796 Chr.3: 50612179-50612179 on Build GRCh38
rs1044682704 Chr.3: 50612012-50612012 on Build GRCh38
rs1023306228 Chr.3: 50612008-50612008 on Build GRCh38
rs969670164 Chr.3: 50612010-50612010 on Build GRCh38
rs903393169 Chr.3: 50611891-50611891 on Build GRCh38
rs1023716567 Chr.3: 50612155-50612155 on Build GRCh38
rs1018803703 Chr.3: 50611828-50611828 on Build GRCh38
rs998318323 Chr.3: 50611820-50611837 on Build GRCh38
rs574503994 Chr.3: 50612102-50612102 on Build GRCh38
rs1017009516 Chr.3: 50612116-50612116 on Build GRCh38
rs867612206 Chr.3: 50612013-50612013 on Build GRCh38
rs1028294415 Chr.3: 50611915-50611915 on Build GRCh38
rs765534132 Chr.3: 50612047-50612047 on Build GRCh38
rs971841723 Chr.3: 50612182-50612182 on Build GRCh38
rs935667511 Chr.3: 50612164-50612164 on Build GRCh38
rs945769387 Chr.3: 50612236-50612236 on Build GRCh38

View Full Product Details