Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 68880592-68881137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM19A4
Entrez Gene ID
151647
Gene Name
family with sequence similarity 19 member A4, C-C motif chemokine like
Gene Aliases
TAFA-4, TAFA4
UniGene
Hs.187873
Gene Chromosome Location
Chr.3: 68731764-68932610 on Build GRCh38
Transcripts
NM_001005527.2 NM_182522.4 XM_011533371.1 XM_011533372.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3408868 c.84C>A p.C28* Chr.3: 68880776-68880776 on Build GRCh38
5628655 c.29C>A p.A10D Chr.3: 68880831-68880831 on Build GRCh38
6276555 c.130+6C>A p.? Chr.3: 68880724-68880724 on Build GRCh38
7125625 c.118C>T p.R40W Chr.3: 68880742-68880742 on Build GRCh38
1251860 c.70G>T p.V24L Chr.3: 68880790-68880790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991219535 Chr.3: 68880950-68880950 on Build GRCh38
rs751020598 Chr.3: 68880894-68880894 on Build GRCh38
rs192405598 Chr.3: 68880772-68880772 on Build GRCh38
rs934503055 Chr.3: 68881015-68881015 on Build GRCh38
rs1014714667 Chr.3: 68880922-68880922 on Build GRCh38
rs561089406 Chr.3: 68880632-68880632 on Build GRCh38
rs770350124 Chr.3: 68880998-68880998 on Build GRCh38
rs201443602 Chr.3: 68880736-68880736 on Build GRCh38
rs752269129 Chr.3: 68880763-68880763 on Build GRCh38
rs752644612 Chr.3: 68880874-68880874 on Build GRCh38
rs201386197 Chr.3: 68880712-68880712 on Build GRCh38
rs775670367 Chr.3: 68880720-68880720 on Build GRCh38
rs146433690 Chr.3: 68880716-68880716 on Build GRCh38
rs746682965 Chr.3: 68880779-68880779 on Build GRCh38
rs370734050 Chr.3: 68880791-68880791 on Build GRCh38
rs757662123 Chr.3: 68880935-68880935 on Build GRCh38
rs924509729 Chr.3: 68880976-68880976 on Build GRCh38
rs755519761 Chr.3: 68880689-68880689 on Build GRCh38
rs75705425 Chr.3: 68881091-68881091 on Build GRCh38
rs960125770 Chr.3: 68880674-68880674 on Build GRCh38
rs377392286 Chr.3: 68880864-68880864 on Build GRCh38
rs1021710430 Chr.3: 68880653-68880653 on Build GRCh38
rs754184954 Chr.3: 68880679-68880679 on Build GRCh38
rs1001870630 Chr.3: 68880660-68880660 on Build GRCh38
rs145690051 Chr.3: 68880742-68880742 on Build GRCh38
rs747576058 Chr.3: 68880704-68880704 on Build GRCh38
rs1025008778 Chr.3: 68880683-68880683 on Build GRCh38
rs778653238 Chr.3: 68880761-68880761 on Build GRCh38
rs913415177 Chr.3: 68881095-68881095 on Build GRCh38
rs557278163 Chr.3: 68880760-68880760 on Build GRCh38
rs746189741 Chr.3: 68880810-68880810 on Build GRCh38
rs201168159 Chr.3: 68880838-68880838 on Build GRCh38
rs774456248 Chr.3: 68880822-68880822 on Build GRCh38
rs962766601 Chr.3: 68880925-68880925 on Build GRCh38
rs766313661 Chr.3: 68880749-68880749 on Build GRCh38
rs914695035 Chr.3: 68881068-68881068 on Build GRCh38
rs771764731 Chr.3: 68880705-68880705 on Build GRCh38
rs981778037 Chr.3: 68880974-68880974 on Build GRCh38
rs904406098 Chr.3: 68880654-68880654 on Build GRCh38
rs909609371 Chr.3: 68880627-68880627 on Build GRCh38
rs1042700358 Chr.3: 68880836-68880836 on Build GRCh38
rs375573883 Chr.3: 68880719-68880719 on Build GRCh38
rs1014041972 Chr.3: 68880917-68880917 on Build GRCh38
rs777199118 Chr.3: 68880892-68880892 on Build GRCh38
rs779607944 Chr.3: 68881033-68881033 on Build GRCh38
rs753641196 Chr.3: 68880755-68880755 on Build GRCh38
rs1034164269 Chr.3: 68880663-68880663 on Build GRCh38
rs965620738 Chr.3: 68881063-68881063 on Build GRCh38
rs529389894 Chr.3: 68880637-68880637 on Build GRCh38
rs758327475 Chr.3: 68880883-68880883 on Build GRCh38
rs200368314 Chr.3: 68880735-68880735 on Build GRCh38
rs771354881 Chr.3: 68880715-68880715 on Build GRCh38
rs780756699 Chr.3: 68880896-68880896 on Build GRCh38
rs114565699 Chr.3: 68880812-68880812 on Build GRCh38
rs749302391 Chr.3: 68880685-68880685 on Build GRCh38
rs369496312 Chr.3: 68880695-68880695 on Build GRCh38
rs140722910 Chr.3: 68880800-68880800 on Build GRCh38
rs563304858 Chr.3: 68881103-68881103 on Build GRCh38
rs1027091827 Chr.3: 68880961-68880961 on Build GRCh38
rs751781952 Chr.3: 68880741-68880741 on Build GRCh38
rs994227659 Chr.3: 68880959-68880959 on Build GRCh38
rs991545721 Chr.3: 68880641-68880641 on Build GRCh38
rs77227101 Chr.3: 68880818-68880818 on Build GRCh38
rs754932379 Chr.3: 68880687-68880687 on Build GRCh38
rs911459754 Chr.3: 68880983-68880983 on Build GRCh38
rs753693343 Chr.3: 68880855-68880855 on Build GRCh38
rs952807930 Chr.3: 68880968-68880968 on Build GRCh38
rs545062046 Chr.3: 68880942-68880942 on Build GRCh38
rs1040801278 Chr.3: 68880636-68880636 on Build GRCh38
rs895771930 Chr.3: 68880901-68880901 on Build GRCh38
rs775801889 Chr.3: 68880813-68880813 on Build GRCh38
rs944177542 Chr.3: 68880744-68880744 on Build GRCh38
rs757905836 Chr.3: 68880765-68880765 on Build GRCh38
rs1001232691 Chr.3: 68880877-68880877 on Build GRCh38
rs564827210 Chr.3: 68881024-68881024 on Build GRCh38
rs768681819 Chr.3: 68880820-68880820 on Build GRCh38
rs762172046 Chr.3: 68880832-68880832 on Build GRCh38
rs765069037 Chr.3: 68880872-68880872 on Build GRCh38
rs1009903282 Chr.3: 68880645-68880645 on Build GRCh38
rs756932110 Chr.3: 68880895-68880895 on Build GRCh38
rs985510229 Chr.3: 68880970-68880970 on Build GRCh38

View Full Product Details