Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 87992082-87992611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR1F
Entrez Gene ID
3355
Gene Name
5-hydroxytryptamine receptor 1F
Gene Aliases
5-HT-1F, 5-HT1F, 5HT6, HTR1EL, MR77
UniGene
Hs.248136
Gene Chromosome Location
Chr.3: 87792767-87994856 on Build GRCh38
Transcripts
NM_000866.4 NM_001322208.1 NM_001322209.1 NM_001322210.1 XM_005264751.3 XM_011533664.2

SNP Detail

SNP ID Chromosome Location
rs1022086216 Chr.3: 87992496-87992500 on Build GRCh38
rs183024961 Chr.3: 87992120-87992120 on Build GRCh38
rs547937943 Chr.3: 87992305-87992305 on Build GRCh38
rs906335320 Chr.3: 87992316-87992316 on Build GRCh38
rs977558630 Chr.3: 87992171-87992171 on Build GRCh38
rs765120346 Chr.3: 87992499-87992503 on Build GRCh38
rs143654627 Chr.3: 87992530-87992530 on Build GRCh38
rs953115858 Chr.3: 87992577-87992577 on Build GRCh38
rs1033608046 Chr.3: 87992118-87992119 on Build GRCh38
rs975608706 Chr.3: 87992199-87992199 on Build GRCh38
rs965495758 Chr.3: 87992561-87992561 on Build GRCh38
rs998925203 Chr.3: 87992531-87992531 on Build GRCh38
rs1019670177 Chr.3: 87992532-87992532 on Build GRCh38
rs893724352 Chr.3: 87992452-87992452 on Build GRCh38
rs533553300 Chr.3: 87992337-87992337 on Build GRCh38
rs564208849 Chr.3: 87992114-87992114 on Build GRCh38
rs967903430 Chr.3: 87992163-87992163 on Build GRCh38
rs199952254 Chr.3: 87992119-87992120 on Build GRCh38
rs921533139 Chr.3: 87992203-87992203 on Build GRCh38
rs1048569303 Chr.3: 87992226-87992226 on Build GRCh38
rs186944680 Chr.3: 87992317-87992317 on Build GRCh38
rs776583941 Chr.3: 87992181-87992183 on Build GRCh38
rs1055007091 Chr.3: 87992345-87992345 on Build GRCh38
rs530752158 Chr.3: 87992185-87992185 on Build GRCh38
rs571661988 Chr.3: 87992118-87992118 on Build GRCh38
rs908282080 Chr.3: 87992244-87992244 on Build GRCh38
rs866285822 Chr.3: 87992266-87992266 on Build GRCh38
rs530334384 Chr.3: 87992210-87992210 on Build GRCh38
rs939775184 Chr.3: 87992281-87992281 on Build GRCh38
rs9310060 Chr.3: 87992130-87992130 on Build GRCh38
rs1046248521 Chr.3: 87992312-87992312 on Build GRCh38
rs191797735 Chr.3: 87992574-87992574 on Build GRCh38
rs769988749 Chr.3: 87992239-87992239 on Build GRCh38
rs569326582 Chr.3: 87992179-87992179 on Build GRCh38
rs975556213 Chr.3: 87992571-87992571 on Build GRCh38
rs2016224 Chr.3: 87992500-87992500 on Build GRCh38
rs1011570814 Chr.3: 87992477-87992477 on Build GRCh38
rs965587484 Chr.3: 87992195-87992195 on Build GRCh38

View Full Product Details