Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 132597144-132597686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAD11
Entrez Gene ID
84129
Gene Name
acyl-CoA dehydrogenase family member 11
Gene Aliases
ACAD-11
UniGene
Hs.441378
Gene Chromosome Location
Chr.3: 132558138-132660131 on Build GRCh38
Transcripts
NM_032169.4 NR_132426.1 NR_132427.1 NR_132428.1 NM_016557.3 NR_037804.1

Gene Symbol
ACKR4
Entrez Gene ID
51554
Gene Name
atypical chemokine receptor 4
Gene Aliases
CC-CKR-11, CCBP2, CCR-11, CCR10, CCR11, CCRL1, CCX CKR, CCX-CKR, CKR-11, PPR1, VSHK1
UniGene
Hs.310512
Gene Chromosome Location
Chr.3: 132597237-132602644 on Build GRCh38
Transcripts
NM_032169.4 NR_132426.1 NR_132427.1 NR_132428.1 NM_016557.3 NR_037804.1

Gene Symbol
NPHP3-ACAD11
Entrez Gene ID
100532724
Gene Name
NPHP3-ACAD11 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.511991
Gene Chromosome Location
Chr.3: 132558138-132722459 on Build GRCh38
Transcripts
NM_032169.4 NR_132426.1 NR_132427.1 NR_132428.1 NM_016557.3 NR_037804.1

SNP Detail

SNP ID Chromosome Location
rs988168751 Chr.3: 132597418-132597418 on Build GRCh38
rs966639951 Chr.3: 132597174-132597174 on Build GRCh38
rs577419842 Chr.3: 132597359-132597359 on Build GRCh38
rs537642040 Chr.3: 132597266-132597266 on Build GRCh38
rs577279796 Chr.3: 132597454-132597454 on Build GRCh38
rs376100309 Chr.3: 132597441-132597441 on Build GRCh38
rs562958996 Chr.3: 132597660-132597660 on Build GRCh38
rs927821312 Chr.3: 132597236-132597236 on Build GRCh38
rs377247014 Chr.3: 132597295-132597295 on Build GRCh38
rs534230098 Chr.3: 132597410-132597410 on Build GRCh38
rs898818641 Chr.3: 132597549-132597549 on Build GRCh38
rs556507285 Chr.3: 132597231-132597231 on Build GRCh38
rs925276449 Chr.3: 132597406-132597406 on Build GRCh38
rs553869127 Chr.3: 132597455-132597455 on Build GRCh38
rs778000523 Chr.3: 132597648-132597649 on Build GRCh38
rs1007193978 Chr.3: 132597176-132597176 on Build GRCh38
rs993182316 Chr.3: 132597570-132597570 on Build GRCh38
rs993325480 Chr.3: 132597279-132597279 on Build GRCh38
rs1052372703 Chr.3: 132597507-132597507 on Build GRCh38
rs13076630 Chr.3: 132597233-132597233 on Build GRCh38
rs1034905367 Chr.3: 132597514-132597514 on Build GRCh38
rs771943814 Chr.3: 132597623-132597623 on Build GRCh38
rs770004847 Chr.3: 132597546-132597546 on Build GRCh38
rs559297960 Chr.3: 132597523-132597523 on Build GRCh38
rs371085640 Chr.3: 132597366-132597366 on Build GRCh38
rs540669252 Chr.3: 132597361-132597361 on Build GRCh38
rs910767279 Chr.3: 132597522-132597522 on Build GRCh38
rs78680330 Chr.3: 132597626-132597626 on Build GRCh38
rs1048464520 Chr.3: 132597659-132597659 on Build GRCh38
rs181499241 Chr.3: 132597404-132597404 on Build GRCh38
rs6777377 Chr.3: 132597261-132597261 on Build GRCh38
rs1047961456 Chr.3: 132597360-132597360 on Build GRCh38
rs916387751 Chr.3: 132597604-132597604 on Build GRCh38
rs966359104 Chr.3: 132597501-132597501 on Build GRCh38
rs913994755 Chr.3: 132597477-132597477 on Build GRCh38
rs545681658 Chr.3: 132597355-132597355 on Build GRCh38
rs190566734 Chr.3: 132597374-132597374 on Build GRCh38
rs990640766 Chr.3: 132597601-132597601 on Build GRCh38
rs13076139 Chr.3: 132597262-132597262 on Build GRCh38
rs565591060 Chr.3: 132597332-132597332 on Build GRCh38
rs948831484 Chr.3: 132597615-132597615 on Build GRCh38
rs3217656 Chr.3: 132597362-132597362 on Build GRCh38
rs959876451 Chr.3: 132597338-132597338 on Build GRCh38
rs1014516366 Chr.3: 132597348-132597348 on Build GRCh38
rs117854933 Chr.3: 132597506-132597506 on Build GRCh38
rs756365289 Chr.3: 132597230-132597230 on Build GRCh38
rs1037993313 Chr.3: 132597531-132597531 on Build GRCh38
rs948736859 Chr.3: 132597312-132597312 on Build GRCh38
rs13076120 Chr.3: 132597227-132597227 on Build GRCh38
rs973457796 Chr.3: 132597616-132597617 on Build GRCh38
rs1010641155 Chr.3: 132597465-132597465 on Build GRCh38
rs995118668 Chr.3: 132597435-132597435 on Build GRCh38
rs185691726 Chr.3: 132597213-132597213 on Build GRCh38

View Full Product Details