Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 114103510-114104013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYT6
Entrez Gene ID
148281
Gene Name
synaptotagmin 6
Gene Aliases
sytVI
UniGene
Hs.370963
Gene Chromosome Location
Chr.1: 114089292-114153919 on Build GRCh38
Transcripts
NM_001270805.1 NM_205848.3 XM_017000371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4218042 c.1115T>A p.L372Q Chr.1: 114103678-114103678 on Build GRCh38
6576718 c.894delT p.K299fs*18 Chr.1: 114103644-114103644 on Build GRCh38
1267309 c.914C>T p.A305V Chr.1: 114103624-114103624 on Build GRCh38
6045255 c.1192+1G>T p.? Chr.1: 114103600-114103600 on Build GRCh38
201656 c.822C>T p.S274S Chr.1: 114103716-114103716 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768488507 Chr.1: 114103557-114103557 on Build GRCh38
rs932008801 Chr.1: 114103764-114103764 on Build GRCh38
rs749395181 Chr.1: 114103759-114103759 on Build GRCh38
rs757361800 Chr.1: 114103624-114103624 on Build GRCh38
rs760235846 Chr.1: 114103674-114103674 on Build GRCh38
rs368052418 Chr.1: 114103716-114103716 on Build GRCh38
rs778239423 Chr.1: 114103615-114103615 on Build GRCh38
rs747991912 Chr.1: 114103652-114103652 on Build GRCh38
rs533217908 Chr.1: 114103641-114103641 on Build GRCh38
rs754854811 Chr.1: 114103767-114103767 on Build GRCh38
rs780069747 Chr.1: 114103753-114103753 on Build GRCh38
rs1057343425 Chr.1: 114103936-114103936 on Build GRCh38
rs893985774 Chr.1: 114103726-114103726 on Build GRCh38
rs148778517 Chr.1: 114103913-114103913 on Build GRCh38
rs368439923 Chr.1: 114103568-114103568 on Build GRCh38
rs750707151 Chr.1: 114103744-114103744 on Build GRCh38
rs11585203 Chr.1: 114103977-114103977 on Build GRCh38
rs563313845 Chr.1: 114103972-114103972 on Build GRCh38
rs760270348 Chr.1: 114103578-114103578 on Build GRCh38
rs771229377 Chr.1: 114103684-114103684 on Build GRCh38
rs34039948 Chr.1: 114103671-114103671 on Build GRCh38
rs758800511 Chr.1: 114103614-114103614 on Build GRCh38
rs370807848 Chr.1: 114103606-114103606 on Build GRCh38
rs771560191 Chr.1: 114103660-114103660 on Build GRCh38
rs781479585 Chr.1: 114103632-114103632 on Build GRCh38
rs74112923 Chr.1: 114103563-114103563 on Build GRCh38
rs773970754 Chr.1: 114103650-114103650 on Build GRCh38
rs774025614 Chr.1: 114103560-114103560 on Build GRCh38
rs767620686 Chr.1: 114103720-114103720 on Build GRCh38
rs778917424 Chr.1: 114103769-114103769 on Build GRCh38
rs778968174 Chr.1: 114103715-114103715 on Build GRCh38
rs374176925 Chr.1: 114103637-114103637 on Build GRCh38
rs569763009 Chr.1: 114103771-114103771 on Build GRCh38
rs751973392 Chr.1: 114103623-114103623 on Build GRCh38
rs745931024 Chr.1: 114103636-114103636 on Build GRCh38
rs766024140 Chr.1: 114103678-114103678 on Build GRCh38
rs775471761 Chr.1: 114103551-114103551 on Build GRCh38
rs1019714772 Chr.1: 114103853-114103853 on Build GRCh38
rs1050544634 Chr.1: 114103787-114103787 on Build GRCh38
rs776468639 Chr.1: 114103831-114103831 on Build GRCh38
rs942134498 Chr.1: 114103809-114103809 on Build GRCh38
rs756156079 Chr.1: 114103747-114103747 on Build GRCh38
rs909249191 Chr.1: 114103808-114103808 on Build GRCh38
rs990737566 Chr.1: 114103554-114103554 on Build GRCh38
rs765839665 Chr.1: 114103591-114103591 on Build GRCh38
rs990686790 Chr.1: 114103951-114103951 on Build GRCh38
rs749264916 Chr.1: 114103647-114103647 on Build GRCh38
rs111876470 Chr.1: 114103546-114103546 on Build GRCh38
rs1009129990 Chr.1: 114103840-114103840 on Build GRCh38
rs952436654 Chr.1: 114103940-114103940 on Build GRCh38
rs768613392 Chr.1: 114103648-114103648 on Build GRCh38
rs778034859 Chr.1: 114103577-114103577 on Build GRCh38
rs764672894 Chr.1: 114103703-114103703 on Build GRCh38
rs112135140 Chr.1: 114103974-114103974 on Build GRCh38
rs564443599 Chr.1: 114103571-114103571 on Build GRCh38
rs535447842 Chr.1: 114103800-114103800 on Build GRCh38
rs534542012 Chr.1: 114103899-114103899 on Build GRCh38
rs549043278 Chr.1: 114103865-114103865 on Build GRCh38
rs894135462 Chr.1: 114103938-114103938 on Build GRCh38
rs1012572931 Chr.1: 114103985-114103985 on Build GRCh38
rs368169468 Chr.1: 114103539-114103539 on Build GRCh38
rs767225438 Chr.1: 114103569-114103569 on Build GRCh38
rs916405359 Chr.1: 114103975-114103975 on Build GRCh38
rs776309568 Chr.1: 114103680-114103680 on Build GRCh38
rs750905 Chr.1: 114103867-114103867 on Build GRCh38

View Full Product Details