Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 149045826-149046334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLTF
Entrez Gene ID
6596
Gene Name
helicase like transcription factor
Gene Aliases
HIP116, HIP116A, HLTF1, RNF80, SMARCA3, SNF2L3, ZBU1
UniGene
Hs.3068
Gene Chromosome Location
Chr.3: 149030064-149086554 on Build GRCh38
Transcripts
NM_001318934.1 NM_001318935.1 NM_003071.3 NM_139048.2 XM_011513091.1 XM_011513092.1 XM_011513093.1 XM_017007078.1 XM_017007079.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1039719 c.1946A>C p.K649T Chr.3: 149046206-149046206 on Build GRCh38
3002585 c.1895G>A p.R632H Chr.3: 149046257-149046257 on Build GRCh38
3660251 c.2002A>G p.I668V Chr.3: 149046150-149046150 on Build GRCh38
4114331 c.1982G>A p.R661H Chr.3: 149046170-149046170 on Build GRCh38
6680877 c.1998G>A p.Q666Q Chr.3: 149046154-149046154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572259697 Chr.3: 149046181-149046181 on Build GRCh38
rs775646317 Chr.3: 149046194-149046194 on Build GRCh38
rs770651808 Chr.3: 149046093-149046093 on Build GRCh38
rs982047162 Chr.3: 149045962-149045962 on Build GRCh38
rs369671613 Chr.3: 149046182-149046182 on Build GRCh38
rs967981708 Chr.3: 149045998-149045998 on Build GRCh38
rs757214237 Chr.3: 149046180-149046180 on Build GRCh38
rs752246801 Chr.3: 149046168-149046168 on Build GRCh38
rs756062602 Chr.3: 149046050-149046050 on Build GRCh38
rs367887879 Chr.3: 149045930-149045930 on Build GRCh38
rs780715352 Chr.3: 149046042-149046042 on Build GRCh38
rs371233960 Chr.3: 149046258-149046258 on Build GRCh38
rs1020687077 Chr.3: 149045938-149045938 on Build GRCh38
rs764697623 Chr.3: 149046128-149046128 on Build GRCh38
rs777212216 Chr.3: 149046129-149046129 on Build GRCh38
rs746059934 Chr.3: 149046115-149046115 on Build GRCh38
rs939890672 Chr.3: 149046280-149046280 on Build GRCh38
rs760727991 Chr.3: 149046127-149046127 on Build GRCh38
rs753382705 Chr.3: 149046139-149046140 on Build GRCh38
rs764582461 Chr.3: 149046268-149046268 on Build GRCh38
rs765649881 Chr.3: 149046140-149046140 on Build GRCh38
rs762423430 Chr.3: 149046137-149046137 on Build GRCh38
rs749228029 Chr.3: 149046060-149046060 on Build GRCh38
rs1049052447 Chr.3: 149046189-149046189 on Build GRCh38
rs776873058 Chr.3: 149046221-149046221 on Build GRCh38
rs777274567 Chr.3: 149046175-149046175 on Build GRCh38
rs927851202 Chr.3: 149045978-149045978 on Build GRCh38
rs762372116 Chr.3: 149046251-149046251 on Build GRCh38
rs754635603 Chr.3: 149046154-149046154 on Build GRCh38
rs137866464 Chr.3: 149046159-149046159 on Build GRCh38
rs1017489 Chr.3: 149046303-149046303 on Build GRCh38
rs772327180 Chr.3: 149046193-149046193 on Build GRCh38
rs772226314 Chr.3: 149046121-149046121 on Build GRCh38
rs889937209 Chr.3: 149046255-149046255 on Build GRCh38
rs146224624 Chr.3: 149046122-149046122 on Build GRCh38
rs752233699 Chr.3: 149046282-149046282 on Build GRCh38
rs182394060 Chr.3: 149045896-149045896 on Build GRCh38
rs753384192 Chr.3: 149046309-149046309 on Build GRCh38
rs112633508 Chr.3: 149046017-149046017 on Build GRCh38
rs186713897 Chr.3: 149046285-149046285 on Build GRCh38
rs755554282 Chr.3: 149046171-149046171 on Build GRCh38
rs768799200 Chr.3: 149046217-149046217 on Build GRCh38
rs897246384 Chr.3: 149046170-149046170 on Build GRCh38
rs747659820 Chr.3: 149046045-149046045 on Build GRCh38
rs774149457 Chr.3: 149046099-149046099 on Build GRCh38
rs1054685916 Chr.3: 149045872-149045872 on Build GRCh38
rs750785353 Chr.3: 149046148-149046148 on Build GRCh38
rs544609277 Chr.3: 149045961-149045961 on Build GRCh38
rs201978472 Chr.3: 149046257-149046257 on Build GRCh38
rs756822122 Chr.3: 149046230-149046231 on Build GRCh38
rs763293274 Chr.3: 149046272-149046272 on Build GRCh38
rs754611917 Chr.3: 149046033-149046033 on Build GRCh38
rs112020463 Chr.3: 149046208-149046208 on Build GRCh38
rs766771675 Chr.3: 149046273-149046273 on Build GRCh38
rs938697843 Chr.3: 149046059-149046059 on Build GRCh38
rs561083562 Chr.3: 149046107-149046107 on Build GRCh38
rs916385810 Chr.3: 149045927-149045927 on Build GRCh38
rs766002591 Chr.3: 149046138-149046138 on Build GRCh38
rs377004804 Chr.3: 149046177-149046177 on Build GRCh38

View Full Product Details