Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 159989051-159989497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12A
Entrez Gene ID
3592
Gene Name
interleukin 12A
Gene Aliases
CLMF, IL-12A, NFSK, NKSF1, P35
UniGene
Hs.673
Gene Chromosome Location
Chr.3: 159988836-159996019 on Build GRCh38
Transcripts
NM_000882.3 NR_108088.1

Gene Symbol
IL12A-AS1
Entrez Gene ID
101928376
Gene Name
IL12A antisense RNA 1
Gene Aliases
-
UniGene
Hs.565865
Gene Chromosome Location
Chr.3: 159913400-160207092 on Build GRCh38
Transcripts
NM_000882.3 NR_108088.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6096693 c.67G>T p.A23S Chr.3: 159989123-159989123 on Build GRCh38
5994715 c.70G>A p.A24T Chr.3: 159989126-159989126 on Build GRCh38
1420310 c.79G>A p.V27M Chr.3: 159989135-159989135 on Build GRCh38
6289638 c.51C>T p.A17A Chr.3: 159989107-159989107 on Build GRCh38
4692137 c.74G>C p.R25P Chr.3: 159989130-159989130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971085380 Chr.3: 159989105-159989105 on Build GRCh38
rs760290351 Chr.3: 159989224-159989224 on Build GRCh38
rs369334761 Chr.3: 159989441-159989441 on Build GRCh38
rs890907994 Chr.3: 159989455-159989455 on Build GRCh38
rs185884099 Chr.3: 159989465-159989465 on Build GRCh38
rs1043546966 Chr.3: 159989408-159989408 on Build GRCh38
rs201787657 Chr.3: 159989203-159989203 on Build GRCh38
rs1025088803 Chr.3: 159989296-159989296 on Build GRCh38
rs999262151 Chr.3: 159989426-159989426 on Build GRCh38
rs757703964 Chr.3: 159989188-159989188 on Build GRCh38
rs1053534390 Chr.3: 159989252-159989252 on Build GRCh38
rs373088816 Chr.3: 159989378-159989378 on Build GRCh38
rs759476225 Chr.3: 159989149-159989149 on Build GRCh38
rs1051754316 Chr.3: 159989191-159989191 on Build GRCh38
rs1055157526 Chr.3: 159989372-159989372 on Build GRCh38
rs144529637 Chr.3: 159989134-159989134 on Build GRCh38
rs559020447 Chr.3: 159989139-159989139 on Build GRCh38
rs566108640 Chr.3: 159989417-159989417 on Build GRCh38
rs777959510 Chr.3: 159989082-159989082 on Build GRCh38
rs1003421753 Chr.3: 159989088-159989088 on Build GRCh38
rs56098462 Chr.3: 159989449-159989449 on Build GRCh38
rs768957287 Chr.3: 159989130-159989130 on Build GRCh38
rs894950765 Chr.3: 159989123-159989123 on Build GRCh38
rs138453663 Chr.3: 159989421-159989421 on Build GRCh38
rs751582831 Chr.3: 159989145-159989145 on Build GRCh38
rs970400421 Chr.3: 159989304-159989304 on Build GRCh38
rs141519443 Chr.3: 159989195-159989195 on Build GRCh38
rs753412819 Chr.3: 159989208-159989209 on Build GRCh38
rs56120852 Chr.3: 159989243-159989243 on Build GRCh38
rs963552372 Chr.3: 159989394-159989394 on Build GRCh38
rs774147199 Chr.3: 159989208-159989208 on Build GRCh38
rs370390683 Chr.3: 159989117-159989117 on Build GRCh38
rs772730722 Chr.3: 159989193-159989193 on Build GRCh38
rs766958739 Chr.3: 159989157-159989157 on Build GRCh38
rs200995173 Chr.3: 159989212-159989212 on Build GRCh38
rs990443122 Chr.3: 159989086-159989086 on Build GRCh38
rs749701569 Chr.3: 159989084-159989084 on Build GRCh38
rs955235155 Chr.3: 159989177-159989177 on Build GRCh38
rs983943676 Chr.3: 159989268-159989268 on Build GRCh38
rs145899976 Chr.3: 159989390-159989390 on Build GRCh38
rs759427203 Chr.3: 159989209-159989209 on Build GRCh38
rs754124989 Chr.3: 159989364-159989364 on Build GRCh38
rs7615589 Chr.3: 159989399-159989399 on Build GRCh38
rs777530345 Chr.3: 159989183-159989183 on Build GRCh38
rs374402488 Chr.3: 159989189-159989189 on Build GRCh38
rs768480307 Chr.3: 159989282-159989282 on Build GRCh38
rs570750047 Chr.3: 159989172-159989172 on Build GRCh38
rs746315801 Chr.3: 159989190-159989190 on Build GRCh38
rs966445174 Chr.3: 159989144-159989144 on Build GRCh38
rs763620704 Chr.3: 159989200-159989212 on Build GRCh38
rs1005225612 Chr.3: 159989369-159989369 on Build GRCh38
rs1014815238 Chr.3: 159989393-159989393 on Build GRCh38
rs775722646 Chr.3: 159989124-159989124 on Build GRCh38
rs200139393 Chr.3: 159989089-159989089 on Build GRCh38
rs779528441 Chr.3: 159989095-159989095 on Build GRCh38
rs866444369 Chr.3: 159989080-159989080 on Build GRCh38
rs754139788 Chr.3: 159989186-159989186 on Build GRCh38
rs894934598 Chr.3: 159989289-159989289 on Build GRCh38
rs745934556 Chr.3: 159989115-159989115 on Build GRCh38
rs556712795 Chr.3: 159989258-159989258 on Build GRCh38
rs369660091 Chr.3: 159989205-159989205 on Build GRCh38
rs755783532 Chr.3: 159989074-159989074 on Build GRCh38
rs767481934 Chr.3: 159989210-159989210 on Build GRCh38
rs529786364 Chr.3: 159989338-159989338 on Build GRCh38
rs755730446 Chr.3: 159989176-159989176 on Build GRCh38
rs895044058 Chr.3: 159989380-159989380 on Build GRCh38
rs963218906 Chr.3: 159989132-159989132 on Build GRCh38
rs541740567 Chr.3: 159989232-159989232 on Build GRCh38
rs760932835 Chr.3: 159989129-159989129 on Build GRCh38
rs927962475 Chr.3: 159989291-159989291 on Build GRCh38
rs377562012 Chr.3: 159989194-159989194 on Build GRCh38
rs939360229 Chr.3: 159989306-159989306 on Build GRCh38

View Full Product Details