Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 170095806-170096317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHC3
Entrez Gene ID
80012
Gene Name
polyhomeotic homolog 3
Gene Aliases
EDR3, HPH3
UniGene
Hs.529592
Gene Chromosome Location
Chr.3: 170087580-170181779 on Build GRCh38
Transcripts
NM_001308116.1 NM_024947.3 XM_005247789.2 XM_006713753.1 XM_006713754.3 XM_006713756.2 XM_011513186.2 XM_011513187.2 XM_011513188.2 XM_017007235.1 XM_017007236.1 XM_017007237.1 XM_017007238.1 XM_017007239.1

SNP Detail

SNP ID Chromosome Location
rs774035019 Chr.3: 170095948-170095948 on Build GRCh38
rs568494216 Chr.3: 170096175-170096175 on Build GRCh38
rs931095620 Chr.3: 170096230-170096230 on Build GRCh38
rs995194627 Chr.3: 170095988-170095988 on Build GRCh38
rs1036132275 Chr.3: 170095840-170095840 on Build GRCh38
rs1025781692 Chr.3: 170095993-170095993 on Build GRCh38
rs923793491 Chr.3: 170096174-170096174 on Build GRCh38
rs772713170 Chr.3: 170096053-170096053 on Build GRCh38
rs772631604 Chr.3: 170096191-170096191 on Build GRCh38
rs1051003719 Chr.3: 170095924-170095924 on Build GRCh38
rs780496375 Chr.3: 170095863-170095863 on Build GRCh38
rs946720900 Chr.3: 170095897-170095897 on Build GRCh38
rs912487277 Chr.3: 170096189-170096190 on Build GRCh38
rs186669081 Chr.3: 170096034-170096034 on Build GRCh38
rs924061139 Chr.3: 170095946-170095946 on Build GRCh38
rs974590694 Chr.3: 170096139-170096139 on Build GRCh38
rs79070639 Chr.3: 170096189-170096189 on Build GRCh38
rs905494887 Chr.3: 170096017-170096017 on Build GRCh38
rs74898939 Chr.3: 170095845-170095845 on Build GRCh38
rs991507004 Chr.3: 170096076-170096076 on Build GRCh38
rs965868529 Chr.3: 170096138-170096138 on Build GRCh38
rs117315268 Chr.3: 170096006-170096006 on Build GRCh38
rs759978976 Chr.3: 170096080-170096080 on Build GRCh38
rs923738156 Chr.3: 170095919-170095919 on Build GRCh38
rs966383504 Chr.3: 170095867-170095867 on Build GRCh38
rs765756781 Chr.3: 170096114-170096114 on Build GRCh38
rs574922348 Chr.3: 170095830-170095830 on Build GRCh38
rs1039743678 Chr.3: 170095953-170095953 on Build GRCh38
rs567657979 Chr.3: 170096196-170096197 on Build GRCh38
rs950573783 Chr.3: 170095843-170095843 on Build GRCh38
rs1019162679 Chr.3: 170096112-170096112 on Build GRCh38
rs766973178 Chr.3: 170096013-170096013 on Build GRCh38
rs898253996 Chr.3: 170095970-170095970 on Build GRCh38
rs924036303 Chr.3: 170096245-170096245 on Build GRCh38
rs535869554 Chr.3: 170096200-170096200 on Build GRCh38
rs887317153 Chr.3: 170096269-170096269 on Build GRCh38
rs367813157 Chr.3: 170096005-170096005 on Build GRCh38
rs921913057 Chr.3: 170095893-170095893 on Build GRCh38
rs1043755866 Chr.3: 170095938-170095938 on Build GRCh38
rs968013108 Chr.3: 170096194-170096194 on Build GRCh38
rs911793816 Chr.3: 170095853-170095853 on Build GRCh38
rs1056571410 Chr.3: 170096274-170096274 on Build GRCh38
rs116114899 Chr.3: 170095877-170095877 on Build GRCh38
rs942573743 Chr.3: 170096193-170096197 on Build GRCh38
rs528405308 Chr.3: 170095980-170095980 on Build GRCh38
rs979275795 Chr.3: 170095902-170095902 on Build GRCh38
rs977487891 Chr.3: 170096153-170096153 on Build GRCh38
rs775910254 Chr.3: 170096189-170096191 on Build GRCh38
rs994541151 Chr.3: 170096042-170096042 on Build GRCh38
rs1035946135 Chr.3: 170096059-170096059 on Build GRCh38
rs147386464 Chr.3: 170095997-170095997 on Build GRCh38
rs1027826088 Chr.3: 170095837-170095837 on Build GRCh38
rs1004704038 Chr.3: 170096085-170096085 on Build GRCh38
rs935385581 Chr.3: 170096260-170096260 on Build GRCh38
rs890825164 Chr.3: 170095935-170095935 on Build GRCh38
rs935264103 Chr.3: 170095923-170095923 on Build GRCh38
rs550300759 Chr.3: 170096071-170096071 on Build GRCh38
rs879460688 Chr.3: 170096178-170096178 on Build GRCh38
rs200844615 Chr.3: 170096194-170096197 on Build GRCh38
rs951650201 Chr.3: 170095833-170095833 on Build GRCh38

View Full Product Details