Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 116134048-116134542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAB21L3
Entrez Gene ID
126868
Gene Name
mab-21 like 3
Gene Aliases
C1orf161
UniGene
Hs.376194
Gene Chromosome Location
Chr.1: 116072744-116135240 on Build GRCh38
Transcripts
NM_152367.2 XM_011540623.2 XM_011540625.2 XM_011540626.2 XM_017000260.1

SNP Detail

SNP ID Chromosome Location
rs944161066 Chr.1: 116134226-116134226 on Build GRCh38
rs770119611 Chr.1: 116134083-116134084 on Build GRCh38
rs1033352469 Chr.1: 116134429-116134429 on Build GRCh38
rs1027048214 Chr.1: 116134099-116134099 on Build GRCh38
rs928694174 Chr.1: 116134175-116134175 on Build GRCh38
rs1020564658 Chr.1: 116134476-116134476 on Build GRCh38
rs925123527 Chr.1: 116134260-116134260 on Build GRCh38
rs886724432 Chr.1: 116134393-116134393 on Build GRCh38
rs967053080 Chr.1: 116134485-116134485 on Build GRCh38
rs769399655 Chr.1: 116134101-116134102 on Build GRCh38
rs901822477 Chr.1: 116134090-116134090 on Build GRCh38
rs985744187 Chr.1: 116134449-116134449 on Build GRCh38
rs754731518 Chr.1: 116134304-116134304 on Build GRCh38
rs769428739 Chr.1: 116134474-116134474 on Build GRCh38
rs545329952 Chr.1: 116134109-116134109 on Build GRCh38
rs1319607 Chr.1: 116134258-116134258 on Build GRCh38
rs546294326 Chr.1: 116134454-116134454 on Build GRCh38
rs943468293 Chr.1: 116134289-116134289 on Build GRCh38
rs377465727 Chr.1: 116134327-116134327 on Build GRCh38
rs551736509 Chr.1: 116134394-116134394 on Build GRCh38
rs944253675 Chr.1: 116134468-116134468 on Build GRCh38
rs5777262 Chr.1: 116134276-116134279 on Build GRCh38
rs913862394 Chr.1: 116134256-116134256 on Build GRCh38
rs201294152 Chr.1: 116134279-116134279 on Build GRCh38
rs1039470675 Chr.1: 116134081-116134081 on Build GRCh38
rs947343286 Chr.1: 116134439-116134439 on Build GRCh38
rs189748784 Chr.1: 116134498-116134498 on Build GRCh38
rs114040663 Chr.1: 116134190-116134190 on Build GRCh38
rs934680011 Chr.1: 116134178-116134178 on Build GRCh38
rs869123679 Chr.1: 116134287-116134290 on Build GRCh38
rs1023938823 Chr.1: 116134334-116134334 on Build GRCh38
rs911441620 Chr.1: 116134456-116134456 on Build GRCh38
rs981498365 Chr.1: 116134165-116134165 on Build GRCh38
rs773582374 Chr.1: 116134194-116134194 on Build GRCh38
rs750962043 Chr.1: 116134113-116134113 on Build GRCh38
rs960330924 Chr.1: 116134251-116134251 on Build GRCh38
rs533655604 Chr.1: 116134355-116134355 on Build GRCh38
rs991822273 Chr.1: 116134475-116134475 on Build GRCh38
rs992041475 Chr.1: 116134252-116134252 on Build GRCh38
rs146290176 Chr.1: 116134423-116134423 on Build GRCh38
rs7521251 Chr.1: 116134349-116134349 on Build GRCh38
rs925122426 Chr.1: 116134513-116134513 on Build GRCh38
rs1023508968 Chr.1: 116134131-116134131 on Build GRCh38
rs148742356 Chr.1: 116134132-116134132 on Build GRCh38
rs761314286 Chr.1: 116134206-116134206 on Build GRCh38
rs1048581836 Chr.1: 116134504-116134504 on Build GRCh38
rs138078627 Chr.1: 116134102-116134102 on Build GRCh38
rs17035363 Chr.1: 116134102-116134102 on Build GRCh38
rs868584381 Chr.1: 116134208-116134208 on Build GRCh38
rs747308166 Chr.1: 116134291-116134291 on Build GRCh38
rs528637890 Chr.1: 116134114-116134114 on Build GRCh38
rs1319606 Chr.1: 116134232-116134232 on Build GRCh38
rs906006612 Chr.1: 116134412-116134412 on Build GRCh38
rs945289581 Chr.1: 116134257-116134257 on Build GRCh38
rs564299998 Chr.1: 116134074-116134074 on Build GRCh38
rs1041265159 Chr.1: 116134472-116134472 on Build GRCh38
rs142078833 Chr.1: 116134406-116134406 on Build GRCh38
rs988794188 Chr.1: 116134186-116134186 on Build GRCh38
rs981886447 Chr.1: 116134377-116134377 on Build GRCh38
rs889969905 Chr.1: 116134214-116134214 on Build GRCh38

View Full Product Details