Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 25389995-25390519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC4
Entrez Gene ID
29945
Gene Name
anaphase promoting complex subunit 4
Gene Aliases
APC4
UniGene
Hs.152173
Gene Chromosome Location
Chr.4: 25377226-25418498 on Build GRCh38
Transcripts
NM_001286756.1 NM_013367.2 XM_011513838.1 XM_011513839.1 XM_017008138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7167618 c.556G>C p.E186Q Chr.4: 25390176-25390176 on Build GRCh38
673928 c.529G>A p.V177I Chr.4: 25390149-25390149 on Build GRCh38
5866231 c.566C>T p.A189V Chr.4: 25390186-25390186 on Build GRCh38
733655 c.556G>T p.E186* Chr.4: 25390176-25390176 on Build GRCh38
6167277 c.597A>T p.T199T Chr.4: 25390217-25390217 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138210054 Chr.4: 25390255-25390255 on Build GRCh38
rs921960377 Chr.4: 25390320-25390320 on Build GRCh38
rs1007059061 Chr.4: 25390080-25390080 on Build GRCh38
rs181377202 Chr.4: 25390330-25390330 on Build GRCh38
rs777518592 Chr.4: 25390178-25390178 on Build GRCh38
rs1004061270 Chr.4: 25390139-25390139 on Build GRCh38
rs764282053 Chr.4: 25390100-25390100 on Build GRCh38
rs900585533 Chr.4: 25390053-25390053 on Build GRCh38
rs990579615 Chr.4: 25390283-25390283 on Build GRCh38
rs752115448 Chr.4: 25390149-25390149 on Build GRCh38
rs377456652 Chr.4: 25390210-25390210 on Build GRCh38
rs149911152 Chr.4: 25390148-25390148 on Build GRCh38
rs762895109 Chr.4: 25390241-25390241 on Build GRCh38
rs535218219 Chr.4: 25390173-25390173 on Build GRCh38
rs896841282 Chr.4: 25390307-25390307 on Build GRCh38
rs979290652 Chr.4: 25390212-25390212 on Build GRCh38
rs1004420924 Chr.4: 25390388-25390388 on Build GRCh38
rs374391579 Chr.4: 25390086-25390086 on Build GRCh38
rs564437217 Chr.4: 25390466-25390466 on Build GRCh38
rs201642475 Chr.4: 25390183-25390183 on Build GRCh38
rs917845884 Chr.4: 25390287-25390287 on Build GRCh38
rs763675094 Chr.4: 25390274-25390274 on Build GRCh38
rs200843781 Chr.4: 25390439-25390442 on Build GRCh38
rs776116087 Chr.4: 25390218-25390218 on Build GRCh38
rs768016403 Chr.4: 25390426-25390427 on Build GRCh38
rs370173906 Chr.4: 25390242-25390242 on Build GRCh38
rs1048655677 Chr.4: 25390351-25390351 on Build GRCh38
rs146440666 Chr.4: 25390205-25390205 on Build GRCh38
rs770194654 Chr.4: 25390214-25390214 on Build GRCh38
rs373309639 Chr.4: 25390272-25390272 on Build GRCh38
rs372522116 Chr.4: 25390304-25390304 on Build GRCh38
rs185809906 Chr.4: 25390348-25390348 on Build GRCh38
rs762037247 Chr.4: 25390267-25390267 on Build GRCh38
rs771350520 Chr.4: 25390209-25390209 on Build GRCh38
rs549611082 Chr.4: 25390070-25390070 on Build GRCh38
rs760426798 Chr.4: 25390106-25390106 on Build GRCh38
rs10001307 Chr.4: 25390361-25390361 on Build GRCh38
rs889497379 Chr.4: 25390358-25390358 on Build GRCh38
rs184720412 Chr.4: 25390113-25390113 on Build GRCh38
rs901325228 Chr.4: 25390392-25390392 on Build GRCh38
rs753752934 Chr.4: 25390119-25390119 on Build GRCh38
rs575939389 Chr.4: 25390302-25390302 on Build GRCh38
rs773877660 Chr.4: 25390076-25390076 on Build GRCh38
rs889990356 Chr.4: 25390064-25390064 on Build GRCh38
rs562446911 Chr.4: 25390305-25390306 on Build GRCh38
rs541201216 Chr.4: 25390453-25390453 on Build GRCh38
rs188580692 Chr.4: 25390246-25390246 on Build GRCh38
rs1020729158 Chr.4: 25390199-25390199 on Build GRCh38
rs1010936215 Chr.4: 25390454-25390454 on Build GRCh38
rs1036931881 Chr.4: 25390298-25390298 on Build GRCh38
rs988848095 Chr.4: 25390163-25390163 on Build GRCh38
rs111628527 Chr.4: 25390034-25390034 on Build GRCh38
rs754859496 Chr.4: 25390131-25390131 on Build GRCh38
rs962667806 Chr.4: 25390154-25390154 on Build GRCh38
rs555692788 Chr.4: 25390292-25390292 on Build GRCh38
rs995462976 Chr.4: 25390427-25390427 on Build GRCh38
rs370102686 Chr.4: 25390085-25390085 on Build GRCh38
rs1019772734 Chr.4: 25390387-25390387 on Build GRCh38

View Full Product Details