Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 39737310-39737826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2K
Entrez Gene ID
3093
Gene Name
ubiquitin conjugating enzyme E2 K
Gene Aliases
E2-25K, HIP2, HYPG, LIG, UBC1
UniGene
Hs.50308
Gene Chromosome Location
Chr.4: 39698044-39782792 on Build GRCh38
Transcripts
NM_001111112.1 NM_001111113.1 NM_001312646.1 NM_001312647.1 NM_001312648.1 NM_005339.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3603594 c.76C>T p.Q26* Chr.4: 39737432-39737432 on Build GRCh38
6413277 c.82A>G p.K28E Chr.4: 39737438-39737438 on Build GRCh38
6319545 c.68G>T p.S23I Chr.4: 39737424-39737424 on Build GRCh38
286232 c.157+8C>A p.? Chr.4: 39737521-39737521 on Build GRCh38
6815104 c.124G>T p.E42* Chr.4: 39737480-39737480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746598888 Chr.4: 39737384-39737384 on Build GRCh38
rs751841650 Chr.4: 39737449-39737449 on Build GRCh38
rs1020768947 Chr.4: 39737535-39737535 on Build GRCh38
rs760067106 Chr.4: 39737401-39737401 on Build GRCh38
rs776889350 Chr.4: 39737547-39737547 on Build GRCh38
rs779851573 Chr.4: 39737548-39737548 on Build GRCh38
rs1001786555 Chr.4: 39737367-39737367 on Build GRCh38
rs539687590 Chr.4: 39737744-39737744 on Build GRCh38
rs776932999 Chr.4: 39737398-39737398 on Build GRCh38
rs1011113447 Chr.4: 39737488-39737488 on Build GRCh38
rs763153644 Chr.4: 39737425-39737425 on Build GRCh38
rs548218674 Chr.4: 39737585-39737585 on Build GRCh38
rs1012894220 Chr.4: 39737740-39737740 on Build GRCh38
rs181301725 Chr.4: 39737771-39737771 on Build GRCh38
rs971569774 Chr.4: 39737345-39737345 on Build GRCh38
rs565808595 Chr.4: 39737534-39737534 on Build GRCh38
rs759663064 Chr.4: 39737629-39737629 on Build GRCh38
rs867623067 Chr.4: 39737504-39737504 on Build GRCh38
rs1054798606 Chr.4: 39737796-39737796 on Build GRCh38
rs375761297 Chr.4: 39737431-39737431 on Build GRCh38
rs201140126 Chr.4: 39737450-39737450 on Build GRCh38
rs113098017 Chr.4: 39737454-39737454 on Build GRCh38
rs766305230 Chr.4: 39737465-39737465 on Build GRCh38
rs557820136 Chr.4: 39737380-39737380 on Build GRCh38
rs1018832545 Chr.4: 39737582-39737582 on Build GRCh38
rs920630274 Chr.4: 39737610-39737610 on Build GRCh38
rs752571092 Chr.4: 39737536-39737536 on Build GRCh38
rs183791284 Chr.4: 39737714-39737714 on Build GRCh38
rs907965684 Chr.4: 39737640-39737640 on Build GRCh38
rs534416534 Chr.4: 39737546-39737546 on Build GRCh38
rs911606228 Chr.4: 39737611-39737611 on Build GRCh38
rs894498317 Chr.4: 39737725-39737725 on Build GRCh38
rs765898622 Chr.4: 39737656-39737656 on Build GRCh38
rs998991438 Chr.4: 39737360-39737360 on Build GRCh38
rs557252489 Chr.4: 39737685-39737685 on Build GRCh38
rs552508543 Chr.4: 39737422-39737422 on Build GRCh38
rs180680822 Chr.4: 39737605-39737605 on Build GRCh38
rs188502257 Chr.4: 39737768-39737768 on Build GRCh38
rs754706332 Chr.4: 39737527-39737527 on Build GRCh38
rs919076558 Chr.4: 39737650-39737650 on Build GRCh38
rs1049433037 Chr.4: 39737723-39737723 on Build GRCh38
rs914943413 Chr.4: 39737522-39737522 on Build GRCh38
rs983969765 Chr.4: 39737624-39737624 on Build GRCh38
rs777546794 Chr.4: 39737371-39737371 on Build GRCh38
rs906143767 Chr.4: 39737364-39737364 on Build GRCh38
rs879314475 Chr.4: 39737628-39737628 on Build GRCh38
rs1032343880 Chr.4: 39737363-39737363 on Build GRCh38
rs1036236771 Chr.4: 39737644-39737644 on Build GRCh38
rs753722032 Chr.4: 39737521-39737521 on Build GRCh38
rs563548473 Chr.4: 39737337-39737337 on Build GRCh38
rs74701834 Chr.4: 39737616-39737616 on Build GRCh38
rs770377034 Chr.4: 39737416-39737416 on Build GRCh38
rs376535981 Chr.4: 39737388-39737388 on Build GRCh38
rs1055207780 Chr.4: 39737387-39737387 on Build GRCh38

View Full Product Details