Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 109658869-109659282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCUB
Entrez Gene ID
55013
Gene Name
mitochondrial calcium uniporter dominant negative beta subunit
Gene Aliases
CCDC109B
UniGene
Hs.234149
Gene Chromosome Location
Chr.4: 109560199-109688719 on Build GRCh38
Transcripts
NM_017918.4 XM_006714246.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6469209 c.175+10C>T p.? Chr.4: 109659096-109659096 on Build GRCh38
6404299 c.106C>T p.R36C Chr.4: 109659017-109659017 on Build GRCh38
7359693 c.139C>G p.Q47E Chr.4: 109659050-109659050 on Build GRCh38
1426334 c.170C>T p.P57L Chr.4: 109659081-109659081 on Build GRCh38
6403793 c.110T>C p.V37A Chr.4: 109659021-109659021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918922849 Chr.4: 109658915-109658915 on Build GRCh38
rs943084177 Chr.4: 109659204-109659204 on Build GRCh38
rs567972526 Chr.4: 109659187-109659187 on Build GRCh38
rs190739225 Chr.4: 109659137-109659137 on Build GRCh38
rs1038907314 Chr.4: 109659007-109659007 on Build GRCh38
rs113161118 Chr.4: 109659095-109659095 on Build GRCh38
rs138159588 Chr.4: 109659154-109659154 on Build GRCh38
rs576183150 Chr.4: 109658969-109658969 on Build GRCh38
rs984518559 Chr.4: 109658918-109658918 on Build GRCh38
rs185634269 Chr.4: 109659120-109659120 on Build GRCh38
rs1055540256 Chr.4: 109658966-109658966 on Build GRCh38
rs376679283 Chr.4: 109659029-109659029 on Build GRCh38
rs867198846 Chr.4: 109659017-109659017 on Build GRCh38
rs904096055 Chr.4: 109658916-109658916 on Build GRCh38
rs943157562 Chr.4: 109658979-109658980 on Build GRCh38
rs774167265 Chr.4: 109659120-109659123 on Build GRCh38
rs891754276 Chr.4: 109659051-109659051 on Build GRCh38
rs974521000 Chr.4: 109659212-109659212 on Build GRCh38
rs1004034336 Chr.4: 109659053-109659053 on Build GRCh38
rs762262326 Chr.4: 109659000-109659000 on Build GRCh38
rs756560575 Chr.4: 109659200-109659200 on Build GRCh38
rs923093532 Chr.4: 109659216-109659216 on Build GRCh38
rs912300447 Chr.4: 109659232-109659232 on Build GRCh38
rs905974756 Chr.4: 109659071-109659071 on Build GRCh38
rs950380497 Chr.4: 109659153-109659153 on Build GRCh38
rs544852646 Chr.4: 109659089-109659089 on Build GRCh38
rs752396801 Chr.4: 109659070-109659070 on Build GRCh38
rs1543812 Chr.4: 109658928-109658928 on Build GRCh38
rs750963884 Chr.4: 109659192-109659192 on Build GRCh38
rs770851423 Chr.4: 109658972-109658973 on Build GRCh38
rs527411669 Chr.4: 109659199-109659199 on Build GRCh38
rs907057056 Chr.4: 109659050-109659050 on Build GRCh38
rs375249063 Chr.4: 109659058-109659058 on Build GRCh38
rs761602665 Chr.4: 109659020-109659020 on Build GRCh38
rs764375103 Chr.4: 109658912-109658912 on Build GRCh38
rs1035805791 Chr.4: 109659091-109659091 on Build GRCh38
rs1028414169 Chr.4: 109659172-109659172 on Build GRCh38
rs763818195 Chr.4: 109659113-109659113 on Build GRCh38
rs960518552 Chr.4: 109659116-109659116 on Build GRCh38
rs561429243 Chr.4: 109659193-109659193 on Build GRCh38

View Full Product Details