Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 140390503-140391019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLGN
Entrez Gene ID
1047
Gene Name
calmegin
Gene Aliases
-
UniGene
Hs.86368
Gene Chromosome Location
Chr.4: 140388453-140427661 on Build GRCh38
Transcripts
NM_001130675.1 NM_004362.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5521146 c.1714G>A p.E572K Chr.4: 140390666-140390666 on Build GRCh38
3600449 c.1732A>C p.K578Q Chr.4: 140390648-140390648 on Build GRCh38
1051442 c.1667C>A p.P556H Chr.4: 140390713-140390713 on Build GRCh38
1051441 c.1742C>A p.S581* Chr.4: 140390638-140390638 on Build GRCh38
1236404 c.1678A>T p.S560C Chr.4: 140390702-140390702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150241035 Chr.4: 140390673-140390673 on Build GRCh38
rs113979220 Chr.4: 140390704-140390704 on Build GRCh38
rs886995408 Chr.4: 140390919-140390919 on Build GRCh38
rs199887896 Chr.4: 140390596-140390596 on Build GRCh38
rs995455634 Chr.4: 140390565-140390565 on Build GRCh38
rs764608425 Chr.4: 140390646-140390646 on Build GRCh38
rs778559940 Chr.4: 140390723-140390723 on Build GRCh38
rs914803349 Chr.4: 140390807-140390807 on Build GRCh38
rs531190435 Chr.4: 140390795-140390795 on Build GRCh38
rs1026919372 Chr.4: 140390917-140390917 on Build GRCh38
rs775934904 Chr.4: 140390638-140390638 on Build GRCh38
rs112508276 Chr.4: 140390966-140390966 on Build GRCh38
rs899503289 Chr.4: 140390879-140390879 on Build GRCh38
rs748793236 Chr.4: 140390602-140390602 on Build GRCh38
rs770317342 Chr.4: 140390616-140390616 on Build GRCh38
rs528720060 Chr.4: 140390535-140390535 on Build GRCh38
rs771920685 Chr.4: 140390630-140390630 on Build GRCh38
rs1015585737 Chr.4: 140390789-140390789 on Build GRCh38
rs777104540 Chr.4: 140390778-140390778 on Build GRCh38
rs571822411 Chr.4: 140390809-140390809 on Build GRCh38
rs560965410 Chr.4: 140390734-140390734 on Build GRCh38
rs920352871 Chr.4: 140390849-140390849 on Build GRCh38
rs372814271 Chr.4: 140390660-140390660 on Build GRCh38
rs985812463 Chr.4: 140390937-140390937 on Build GRCh38
rs747363082 Chr.4: 140390768-140390768 on Build GRCh38
rs777593558 Chr.4: 140390598-140390598 on Build GRCh38
rs755892878 Chr.4: 140390687-140390687 on Build GRCh38
rs1039851410 Chr.4: 140390877-140390877 on Build GRCh38
rs202235605 Chr.4: 140390705-140390705 on Build GRCh38
rs765276633 Chr.4: 140390665-140390665 on Build GRCh38
rs200917325 Chr.4: 140390588-140390588 on Build GRCh38
rs358328 Chr.4: 140390553-140390553 on Build GRCh38
rs891271671 Chr.4: 140390863-140390863 on Build GRCh38
rs927351219 Chr.4: 140390788-140390788 on Build GRCh38
rs953080316 Chr.4: 140390894-140390894 on Build GRCh38
rs943842976 Chr.4: 140390871-140390871 on Build GRCh38
rs358329 Chr.4: 140390806-140390806 on Build GRCh38
rs899492479 Chr.4: 140390561-140390561 on Build GRCh38
rs772799147 Chr.4: 140390827-140390827 on Build GRCh38
rs1014530359 Chr.4: 140390782-140390782 on Build GRCh38
rs749474842 Chr.4: 140390646-140390648 on Build GRCh38
rs749157176 Chr.4: 140390707-140390707 on Build GRCh38
rs937310465 Chr.4: 140390803-140390803 on Build GRCh38
rs768874259 Chr.4: 140390769-140390769 on Build GRCh38
rs759251938 Chr.4: 140390626-140390626 on Build GRCh38
rs775309450 Chr.4: 140390765-140390765 on Build GRCh38
rs145828622 Chr.4: 140390720-140390720 on Build GRCh38
rs1010685733 Chr.4: 140390545-140390545 on Build GRCh38
rs774055681 Chr.4: 140390621-140390621 on Build GRCh38
rs533174138 Chr.4: 140390662-140390662 on Build GRCh38
rs752479617 Chr.4: 140390677-140390677 on Build GRCh38
rs771208708 Chr.4: 140390648-140390649 on Build GRCh38
rs773573080 Chr.4: 140390614-140390614 on Build GRCh38
rs545177462 Chr.4: 140390666-140390666 on Build GRCh38
rs995129693 Chr.4: 140390900-140390900 on Build GRCh38
rs748002859 Chr.4: 140390584-140390584 on Build GRCh38
rs760964931 Chr.4: 140390642-140390642 on Build GRCh38
rs780258346 Chr.4: 140390675-140390675 on Build GRCh38
rs771788072 Chr.4: 140390740-140390740 on Build GRCh38

View Full Product Details