Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 145138347-145138860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTUD4
Entrez Gene ID
54726
Gene Name
OTU deubiquitinase 4
Gene Aliases
DUBA6, HIN1, HSHIN1
UniGene
Hs.270851
Gene Chromosome Location
Chr.4: 145133650-145180019 on Build GRCh38
Transcripts
NM_001102653.1 XM_005263079.4 XM_011532040.2 XM_011532041.2 XM_011532042.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6736195 c.2378C>G p.P793R Chr.4: 145138397-145138397 on Build GRCh38
7326627 c.2211G>A p.K737K Chr.4: 145138564-145138564 on Build GRCh38
7115890 c.2140C>G p.Q714E Chr.4: 145138440-145138440 on Build GRCh38
4710574 c.2167C>A p.P723T Chr.4: 145138608-145138608 on Build GRCh38
6298955 c.2149C>A p.Q717K Chr.4: 145138431-145138431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs137890414 Chr.4: 145138411-145138411 on Build GRCh38
rs774191545 Chr.4: 145138678-145138678 on Build GRCh38
rs746004244 Chr.4: 145138446-145138446 on Build GRCh38
rs755767422 Chr.4: 145138684-145138686 on Build GRCh38
rs36226997 Chr.4: 145138381-145138381 on Build GRCh38
rs755138044 Chr.4: 145138659-145138659 on Build GRCh38
rs369523527 Chr.4: 145138699-145138699 on Build GRCh38
rs747258201 Chr.4: 145138658-145138658 on Build GRCh38
rs1054747519 Chr.4: 145138497-145138497 on Build GRCh38
rs554189943 Chr.4: 145138708-145138708 on Build GRCh38
rs775082267 Chr.4: 145138682-145138682 on Build GRCh38
rs778243257 Chr.4: 145138416-145138416 on Build GRCh38
rs773178191 Chr.4: 145138376-145138376 on Build GRCh38
rs534270992 Chr.4: 145138696-145138696 on Build GRCh38
rs774329693 Chr.4: 145138529-145138529 on Build GRCh38
rs200113018 Chr.4: 145138412-145138412 on Build GRCh38
rs752425537 Chr.4: 145138576-145138576 on Build GRCh38
rs368717800 Chr.4: 145138456-145138456 on Build GRCh38
rs781000012 Chr.4: 145138676-145138679 on Build GRCh38
rs397775091 Chr.4: 145138666-145138666 on Build GRCh38
rs778149515 Chr.4: 145138515-145138515 on Build GRCh38
rs140860184 Chr.4: 145138414-145138414 on Build GRCh38
rs1036386903 Chr.4: 145138703-145138703 on Build GRCh38
rs764564992 Chr.4: 145138598-145138598 on Build GRCh38
rs747980274 Chr.4: 145138680-145138681 on Build GRCh38
rs776637788 Chr.4: 145138714-145138714 on Build GRCh38
rs769612162 Chr.4: 145138482-145138482 on Build GRCh38
rs770199849 Chr.4: 145138409-145138409 on Build GRCh38
rs112177903 Chr.4: 145138469-145138469 on Build GRCh38
rs773151226 Chr.4: 145138542-145138542 on Build GRCh38
rs745756652 Chr.4: 145138680-145138680 on Build GRCh38
rs543729632 Chr.4: 145138728-145138728 on Build GRCh38
rs757646438 Chr.4: 145138431-145138431 on Build GRCh38
rs143080279 Chr.4: 145138415-145138415 on Build GRCh38
rs771268625 Chr.4: 145138643-145138643 on Build GRCh38
rs958950613 Chr.4: 145138704-145138704 on Build GRCh38
rs770734818 Chr.4: 145138666-145138666 on Build GRCh38
rs771728799 Chr.4: 145138681-145138681 on Build GRCh38
rs969335156 Chr.4: 145138808-145138808 on Build GRCh38
rs1005353584 Chr.4: 145138767-145138767 on Build GRCh38
rs574394439 Chr.4: 145138731-145138731 on Build GRCh38
rs759015189 Chr.4: 145138551-145138551 on Build GRCh38
rs182025062 Chr.4: 145138611-145138611 on Build GRCh38
rs558586803 Chr.4: 145138558-145138558 on Build GRCh38
rs79569624 Chr.4: 145138739-145138739 on Build GRCh38
rs772127605 Chr.4: 145138453-145138453 on Build GRCh38
rs923713223 Chr.4: 145138813-145138813 on Build GRCh38
rs780225506 Chr.4: 145138499-145138499 on Build GRCh38
rs991679081 Chr.4: 145138717-145138717 on Build GRCh38
rs779052504 Chr.4: 145138437-145138437 on Build GRCh38
rs754390918 Chr.4: 145138420-145138420 on Build GRCh38
rs150358837 Chr.4: 145138635-145138635 on Build GRCh38
rs749388482 Chr.4: 145138660-145138660 on Build GRCh38
rs112977068 Chr.4: 145138471-145138471 on Build GRCh38
rs374609116 Chr.4: 145138514-145138514 on Build GRCh38
rs772947085 Chr.4: 145138511-145138511 on Build GRCh38
rs762550741 Chr.4: 145138804-145138804 on Build GRCh38
rs754722217 Chr.4: 145138660-145138661 on Build GRCh38
rs1003921096 Chr.4: 145138811-145138811 on Build GRCh38
rs763445762 Chr.4: 145138823-145138823 on Build GRCh38
rs141253434 Chr.4: 145138613-145138613 on Build GRCh38
rs36226996 Chr.4: 145138661-145138661 on Build GRCh38
rs754082595 Chr.4: 145138609-145138609 on Build GRCh38
rs556075164 Chr.4: 145138689-145138690 on Build GRCh38
rs780071901 Chr.4: 145138656-145138656 on Build GRCh38
rs759370786 Chr.4: 145138726-145138726 on Build GRCh38
rs777630756 Chr.4: 145138662-145138662 on Build GRCh38
rs779348471 Chr.4: 145138619-145138619 on Build GRCh38
rs749091878 Chr.4: 145138664-145138664 on Build GRCh38
rs574378019 Chr.4: 145138722-145138722 on Build GRCh38
rs760354154 Chr.4: 145138593-145138593 on Build GRCh38
rs750977621 Chr.4: 145138397-145138397 on Build GRCh38
rs748823913 Chr.4: 145138695-145138696 on Build GRCh38
rs770709745 Chr.4: 145138525-145138525 on Build GRCh38
rs762964107 Chr.4: 145138517-145138517 on Build GRCh38

View Full Product Details