Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 154536338-154536887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLRG1
Entrez Gene ID
5356
Gene Name
pleiotropic regulator 1
Gene Aliases
Cwc1, PRL1, PRP46, PRPF46, TANGO4
UniGene
Hs.249996
Gene Chromosome Location
Chr.4: 154534997-154550433 on Build GRCh38
Transcripts
NM_001201564.1 NM_002669.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5507364 c.1486-8A>G p.? Chr.4: 154536752-154536752 on Build GRCh38
1618507 c.1486-9T>C p.? Chr.4: 154536753-154536753 on Build GRCh38
1052382 c.1520A>C p.E507A Chr.4: 154536710-154536710 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752054864 Chr.4: 154536660-154536661 on Build GRCh38
rs921902412 Chr.4: 154536628-154536628 on Build GRCh38
rs773215236 Chr.4: 154536786-154536786 on Build GRCh38
rs530498347 Chr.4: 154536644-154536644 on Build GRCh38
rs757000803 Chr.4: 154536734-154536734 on Build GRCh38
rs890754370 Chr.4: 154536697-154536697 on Build GRCh38
rs541721179 Chr.4: 154536428-154536428 on Build GRCh38
rs116491410 Chr.4: 154536406-154536406 on Build GRCh38
rs11943987 Chr.4: 154536478-154536478 on Build GRCh38
rs902242380 Chr.4: 154536423-154536423 on Build GRCh38
rs775341875 Chr.4: 154536792-154536792 on Build GRCh38
rs777075799 Chr.4: 154536778-154536778 on Build GRCh38
rs534385051 Chr.4: 154536531-154536531 on Build GRCh38
rs780490352 Chr.4: 154536763-154536763 on Build GRCh38
rs1027538331 Chr.4: 154536521-154536521 on Build GRCh38
rs981730610 Chr.4: 154536451-154536451 on Build GRCh38
rs768916648 Chr.4: 154536643-154536643 on Build GRCh38
rs7685666 Chr.4: 154536378-154536378 on Build GRCh38
rs763618326 Chr.4: 154536629-154536630 on Build GRCh38
rs746337936 Chr.4: 154536623-154536623 on Build GRCh38
rs946304476 Chr.4: 154536776-154536776 on Build GRCh38
rs764735351 Chr.4: 154536659-154536660 on Build GRCh38
rs966511465 Chr.4: 154536614-154536614 on Build GRCh38
rs758962686 Chr.4: 154536761-154536761 on Build GRCh38
rs775941141 Chr.4: 154536634-154536634 on Build GRCh38
rs760345462 Chr.4: 154536630-154536631 on Build GRCh38
rs767453572 Chr.4: 154536677-154536677 on Build GRCh38
rs57816603 Chr.4: 154536378-154536378 on Build GRCh38
rs745386813 Chr.4: 154536760-154536760 on Build GRCh38
rs959089122 Chr.4: 154536480-154536480 on Build GRCh38
rs1054679572 Chr.4: 154536747-154536747 on Build GRCh38
rs571216753 Chr.4: 154536851-154536851 on Build GRCh38
rs201170057 Chr.4: 154536669-154536669 on Build GRCh38
rs974920503 Chr.4: 154536375-154536375 on Build GRCh38
rs759398734 Chr.4: 154536673-154536673 on Build GRCh38
rs747392378 Chr.4: 154536641-154536641 on Build GRCh38
rs771968437 Chr.4: 154536791-154536791 on Build GRCh38
rs75682462 Chr.4: 154536629-154536629 on Build GRCh38
rs776757232 Chr.4: 154536646-154536646 on Build GRCh38
rs756701599 Chr.4: 154536651-154536652 on Build GRCh38
rs556669848 Chr.4: 154536369-154536369 on Build GRCh38
rs60732160 Chr.4: 154536449-154536449 on Build GRCh38
rs544670490 Chr.4: 154536582-154536582 on Build GRCh38
rs1042265370 Chr.4: 154536802-154536802 on Build GRCh38
rs770005253 Chr.4: 154536784-154536784 on Build GRCh38
rs753554475 Chr.4: 154536652-154536652 on Build GRCh38
rs200722097 Chr.4: 154536752-154536752 on Build GRCh38
rs1054729741 Chr.4: 154536370-154536371 on Build GRCh38
rs140279149 Chr.4: 154536659-154536659 on Build GRCh38
rs747367117 Chr.4: 154536766-154536766 on Build GRCh38
rs756770137 Chr.4: 154536612-154536612 on Build GRCh38
rs763075937 Chr.4: 154536789-154536789 on Build GRCh38
rs893268416 Chr.4: 154536370-154536370 on Build GRCh38
rs555981813 Chr.4: 154536386-154536386 on Build GRCh38
rs761929759 Chr.4: 154536651-154536651 on Build GRCh38
rs546441484 Chr.4: 154536800-154536800 on Build GRCh38
rs201310843 Chr.4: 154536671-154536671 on Build GRCh38
rs988922060 Chr.4: 154536399-154536399 on Build GRCh38
rs1012082387 Chr.4: 154536508-154536508 on Build GRCh38
rs371994884 Chr.4: 154536703-154536703 on Build GRCh38
rs772712154 Chr.4: 154536631-154536631 on Build GRCh38
rs748055769 Chr.4: 154536592-154536592 on Build GRCh38
rs763784369 Chr.4: 154536718-154536718 on Build GRCh38
rs374872052 Chr.4: 154536622-154536622 on Build GRCh38
rs752534937 Chr.4: 154536685-154536685 on Build GRCh38
rs937140873 Chr.4: 154536503-154536503 on Build GRCh38
rs748273412 Chr.4: 154536780-154536780 on Build GRCh38
rs1046817020 Chr.4: 154536662-154536662 on Build GRCh38
rs774470726 Chr.4: 154536630-154536630 on Build GRCh38
rs755534115 Chr.4: 154536660-154536663 on Build GRCh38
rs907383560 Chr.4: 154536807-154536807 on Build GRCh38
rs768949863 Chr.4: 154536769-154536769 on Build GRCh38
rs753386975 Chr.4: 154536725-154536725 on Build GRCh38
rs11945075 Chr.4: 154536856-154536856 on Build GRCh38
rs1003680905 Chr.4: 154536840-154536840 on Build GRCh38

View Full Product Details