Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 154587422-154587923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGA
Entrez Gene ID
2243
Gene Name
fibrinogen alpha chain
Gene Aliases
Fib2
UniGene
Hs.351593
Gene Chromosome Location
Chr.4: 154583126-154590766 on Build GRCh38
Transcripts
NM_000508.4 NM_021871.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
269168 c.510G>T p.E170D Chr.4: 154587512-154587512 on Build GRCh38
5400675 c.498G>A p.M166I Chr.4: 154587524-154587524 on Build GRCh38
6663310 c.385C>T p.R129* Chr.4: 154587637-154587637 on Build GRCh38
3660954 c.395A>G p.E132G Chr.4: 154587627-154587627 on Build GRCh38
5758441 c.467A>C p.Q156P Chr.4: 154587555-154587555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778196524 Chr.4: 154587855-154587855 on Build GRCh38
rs768136586 Chr.4: 154587673-154587673 on Build GRCh38
rs749264613 Chr.4: 154587784-154587792 on Build GRCh38
rs530960205 Chr.4: 154587805-154587805 on Build GRCh38
rs1004545235 Chr.4: 154587678-154587678 on Build GRCh38
rs565128682 Chr.4: 154587633-154587633 on Build GRCh38
rs901030475 Chr.4: 154587735-154587735 on Build GRCh38
rs2070030 Chr.4: 154587452-154587452 on Build GRCh38
rs758230567 Chr.4: 154587745-154587752 on Build GRCh38
rs758609385 Chr.4: 154587512-154587512 on Build GRCh38
rs549204248 Chr.4: 154587840-154587840 on Build GRCh38
rs567459765 Chr.4: 154587881-154587881 on Build GRCh38
rs113236303 Chr.4: 154587658-154587658 on Build GRCh38
rs368845778 Chr.4: 154587665-154587665 on Build GRCh38
rs757186085 Chr.4: 154587749-154587750 on Build GRCh38
rs757643821 Chr.4: 154587481-154587481 on Build GRCh38
rs758873385 Chr.4: 154587745-154587746 on Build GRCh38
rs774679289 Chr.4: 154587802-154587805 on Build GRCh38
rs762480524 Chr.4: 154587587-154587587 on Build GRCh38
rs915900570 Chr.4: 154587580-154587580 on Build GRCh38
rs67296980 Chr.4: 154587801-154587804 on Build GRCh38
rs138084209 Chr.4: 154587770-154587770 on Build GRCh38
rs113206878 Chr.4: 154587696-154587891 on Build GRCh38
rs1035843783 Chr.4: 154587898-154587898 on Build GRCh38
rs1021048121 Chr.4: 154587817-154587817 on Build GRCh38
rs763715993 Chr.4: 154587574-154587574 on Build GRCh38
rs368446857 Chr.4: 154587572-154587572 on Build GRCh38
rs771565963 Chr.4: 154587554-154587554 on Build GRCh38
rs1005698682 Chr.4: 154587702-154587702 on Build GRCh38
rs765085134 Chr.4: 154587463-154587463 on Build GRCh38
rs199828049 Chr.4: 154587547-154587547 on Build GRCh38
rs967629767 Chr.4: 154587836-154587836 on Build GRCh38
rs1016001599 Chr.4: 154587744-154587744 on Build GRCh38
rs928803453 Chr.4: 154587524-154587524 on Build GRCh38
rs766472109 Chr.4: 154587604-154587604 on Build GRCh38
rs770504846 Chr.4: 154587805-154587805 on Build GRCh38
rs375489804 Chr.4: 154587595-154587595 on Build GRCh38
rs777639722 Chr.4: 154587532-154587532 on Build GRCh38
rs749273086 Chr.4: 154587533-154587533 on Build GRCh38
rs531573623 Chr.4: 154587808-154587809 on Build GRCh38
rs879607786 Chr.4: 154587798-154587803 on Build GRCh38
rs993139606 Chr.4: 154587804-154587804 on Build GRCh38
rs60443975 Chr.4: 154587804-154587805 on Build GRCh38
rs147888314 Chr.4: 154587619-154587619 on Build GRCh38
rs551717426 Chr.4: 154587764-154587764 on Build GRCh38
rs765688392 Chr.4: 154587588-154587588 on Build GRCh38
rs113131575 Chr.4: 154587659-154587659 on Build GRCh38
rs979034065 Chr.4: 154587885-154587885 on Build GRCh38
rs750761333 Chr.4: 154587594-154587594 on Build GRCh38
rs532470007 Chr.4: 154587666-154587666 on Build GRCh38
rs746995811 Chr.4: 154587518-154587518 on Build GRCh38
rs1010532824 Chr.4: 154587816-154587816 on Build GRCh38
rs149605794 Chr.4: 154587802-154587803 on Build GRCh38
rs780339572 Chr.4: 154587513-154587513 on Build GRCh38
rs146387238 Chr.4: 154587511-154587511 on Build GRCh38
rs372161622 Chr.4: 154587586-154587586 on Build GRCh38
rs147211930 Chr.4: 154587496-154587496 on Build GRCh38
rs371697849 Chr.4: 154587746-154587749 on Build GRCh38
rs914869108 Chr.4: 154587504-154587504 on Build GRCh38
rs990801554 Chr.4: 154587497-154587497 on Build GRCh38
rs934668242 Chr.4: 154587590-154587591 on Build GRCh38
rs796207030 Chr.4: 154587804-154587805 on Build GRCh38
rs181612102 Chr.4: 154587453-154587453 on Build GRCh38
rs910497026 Chr.4: 154587841-154587841 on Build GRCh38
rs745581691 Chr.4: 154587654-154587654 on Build GRCh38
rs189308943 Chr.4: 154587451-154587451 on Build GRCh38
rs765774596 Chr.4: 154587688-154587688 on Build GRCh38
rs771930192 Chr.4: 154587664-154587664 on Build GRCh38
rs540438738 Chr.4: 154587540-154587540 on Build GRCh38
rs962142987 Chr.4: 154587786-154587791 on Build GRCh38
rs201265671 Chr.4: 154587803-154587804 on Build GRCh38
rs2070018 Chr.4: 154587475-154587475 on Build GRCh38
rs569935098 Chr.4: 154587768-154587768 on Build GRCh38
rs751832077 Chr.4: 154587629-154587629 on Build GRCh38
rs186082331 Chr.4: 154587755-154587755 on Build GRCh38
rs770680765 Chr.4: 154587534-154587534 on Build GRCh38
rs749285585 Chr.4: 154587484-154587484 on Build GRCh38
rs781289913 Chr.4: 154587637-154587637 on Build GRCh38
rs867367623 Chr.4: 154587519-154587519 on Build GRCh38
rs1027921021 Chr.4: 154587457-154587457 on Build GRCh38
rs752861134 Chr.4: 154587685-154587685 on Build GRCh38
rs527793968 Chr.4: 154587460-154587460 on Build GRCh38
rs112646442 Chr.4: 154587509-154587509 on Build GRCh38
rs201528325 Chr.4: 154587746-154587757 on Build GRCh38
rs778832765 Chr.4: 154587646-154587646 on Build GRCh38
rs757242809 Chr.4: 154587640-154587640 on Build GRCh38
rs748622041 Chr.4: 154587804-154587808 on Build GRCh38
rs760686922 Chr.4: 154587745-154587748 on Build GRCh38
rs537149728 Chr.4: 154587723-154587723 on Build GRCh38
rs773803625 Chr.4: 154587693-154587693 on Build GRCh38
rs372105398 Chr.4: 154587744-154587745 on Build GRCh38
rs763181366 Chr.4: 154587695-154587695 on Build GRCh38
rs765031780 Chr.4: 154587509-154587510 on Build GRCh38
rs755117226 Chr.4: 154587520-154587520 on Build GRCh38
rs372831271 Chr.4: 154587679-154587679 on Build GRCh38
rs776342437 Chr.4: 154587672-154587672 on Build GRCh38
rs113455932 Chr.4: 154587739-154587739 on Build GRCh38

View Full Product Details