Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 165992712-165993213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLL1
Entrez Gene ID
7092
Gene Name
tolloid like 1
Gene Aliases
ASD6, TLL
UniGene
Hs.106513
Gene Chromosome Location
Chr.4: 165873238-166105178 on Build GRCh38
Transcripts
NM_001204760.1 NM_012464.4 XM_011532214.1 XM_017008570.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5128831 c.308A>C p.K103T Chr.4: 165992831-165992831 on Build GRCh38
5665225 c.342G>T p.R114S Chr.4: 165992865-165992865 on Build GRCh38
1052968 c.319G>T p.A107S Chr.4: 165992842-165992842 on Build GRCh38
1052969 c.355G>T p.G119C Chr.4: 165992878-165992878 on Build GRCh38
3601695 c.292G>A p.D98N Chr.4: 165992815-165992815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147669973 Chr.4: 165993031-165993031 on Build GRCh38
rs147223461 Chr.4: 165992837-165992837 on Build GRCh38
rs764657488 Chr.4: 165992868-165992870 on Build GRCh38
rs552654988 Chr.4: 165993070-165993070 on Build GRCh38
rs549231272 Chr.4: 165993080-165993080 on Build GRCh38
rs771749988 Chr.4: 165992818-165992818 on Build GRCh38
rs1034420613 Chr.4: 165992762-165992762 on Build GRCh38
rs377042841 Chr.4: 165992817-165992817 on Build GRCh38
rs980689509 Chr.4: 165993125-165993125 on Build GRCh38
rs1034304725 Chr.4: 165993013-165993014 on Build GRCh38
rs748168573 Chr.4: 165992808-165992808 on Build GRCh38
rs942317592 Chr.4: 165992876-165992876 on Build GRCh38
rs760244525 Chr.4: 165992834-165992834 on Build GRCh38
rs776299665 Chr.4: 165992843-165992843 on Build GRCh38
rs891926411 Chr.4: 165992928-165992928 on Build GRCh38
rs535270477 Chr.4: 165992855-165992855 on Build GRCh38
rs960100588 Chr.4: 165992765-165992765 on Build GRCh38
rs373850201 Chr.4: 165992809-165992809 on Build GRCh38
rs747006247 Chr.4: 165993008-165993008 on Build GRCh38
rs984071945 Chr.4: 165993086-165993086 on Build GRCh38
rs766652780 Chr.4: 165992860-165992860 on Build GRCh38
rs138967219 Chr.4: 165992853-165992853 on Build GRCh38
rs111863557 Chr.4: 165992776-165992776 on Build GRCh38
rs200870855 Chr.4: 165992910-165992910 on Build GRCh38
rs754078827 Chr.4: 165992937-165992942 on Build GRCh38
rs142980270 Chr.4: 165992880-165992880 on Build GRCh38
rs971868974 Chr.4: 165993030-165993030 on Build GRCh38
rs997734898 Chr.4: 165992814-165992814 on Build GRCh38
rs76013943 Chr.4: 165992858-165992858 on Build GRCh38
rs200217590 Chr.4: 165992922-165992922 on Build GRCh38
rs953076597 Chr.4: 165992872-165992872 on Build GRCh38
rs781344924 Chr.4: 165992904-165992904 on Build GRCh38
rs776390238 Chr.4: 165992940-165992940 on Build GRCh38
rs765888239 Chr.4: 165992849-165992849 on Build GRCh38
rs761364561 Chr.4: 165992847-165992847 on Build GRCh38
rs909811752 Chr.4: 165993106-165993106 on Build GRCh38
rs775279862 Chr.4: 165992826-165992826 on Build GRCh38
rs768451909 Chr.4: 165992923-165992923 on Build GRCh38
rs558520791 Chr.4: 165992978-165992978 on Build GRCh38
rs756162133 Chr.4: 165992909-165992909 on Build GRCh38
rs768828893 Chr.4: 165992893-165992893 on Build GRCh38
rs370286337 Chr.4: 165992907-165992907 on Build GRCh38
rs764830494 Chr.4: 165992954-165992954 on Build GRCh38
rs929911038 Chr.4: 165992825-165992825 on Build GRCh38
rs761461023 Chr.4: 165992942-165992942 on Build GRCh38
rs375840245 Chr.4: 165992911-165992911 on Build GRCh38
rs529176580 Chr.4: 165993058-165993058 on Build GRCh38
rs781254168 Chr.4: 165992801-165992801 on Build GRCh38
rs184245905 Chr.4: 165992766-165992766 on Build GRCh38
rs73863517 Chr.4: 165993145-165993145 on Build GRCh38

View Full Product Details