Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 169989384-169989893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFAP3L
Entrez Gene ID
9848
Gene Name
microfibrillar associated protein 3 like
Gene Aliases
NYD-sp9
UniGene
Hs.593942
Gene Chromosome Location
Chr.4: 169986597-170033028 on Build GRCh38
Transcripts
NM_001009554.3 NM_001301647.1 NM_001301648.1 NM_021647.7 XM_005263366.3 XM_005263368.3 XM_017008864.1 XM_017008865.1 XM_017008866.1 XM_017008867.1 XM_017008868.1 XM_017008869.1 XM_017008870.1 XM_017008871.1

SNP Detail

SNP ID Chromosome Location
rs938431021 Chr.4: 169989442-169989442 on Build GRCh38
rs1008286590 Chr.4: 169989583-169989583 on Build GRCh38
rs895045611 Chr.4: 169989526-169989526 on Build GRCh38
rs1052376563 Chr.4: 169989570-169989570 on Build GRCh38
rs78227118 Chr.4: 169989670-169989670 on Build GRCh38
rs1010903141 Chr.4: 169989528-169989528 on Build GRCh38
rs551524805 Chr.4: 169989714-169989714 on Build GRCh38
rs948229103 Chr.4: 169989500-169989500 on Build GRCh38
rs762331333 Chr.4: 169989750-169989750 on Build GRCh38
rs1055482009 Chr.4: 169989515-169989515 on Build GRCh38
rs892435621 Chr.4: 169989811-169989811 on Build GRCh38
rs993006189 Chr.4: 169989466-169989466 on Build GRCh38
rs959988871 Chr.4: 169989716-169989716 on Build GRCh38
rs780774686 Chr.4: 169989421-169989421 on Build GRCh38
rs76904336 Chr.4: 169989699-169989699 on Build GRCh38
rs560924797 Chr.4: 169989571-169989571 on Build GRCh38
rs761255387 Chr.4: 169989809-169989809 on Build GRCh38
rs559112401 Chr.4: 169989656-169989656 on Build GRCh38
rs115449916 Chr.4: 169989540-169989540 on Build GRCh38
rs980899135 Chr.4: 169989758-169989758 on Build GRCh38
rs577904895 Chr.4: 169989491-169989491 on Build GRCh38
rs985632837 Chr.4: 169989596-169989596 on Build GRCh38
rs566952636 Chr.4: 169989492-169989492 on Build GRCh38
rs892487474 Chr.4: 169989579-169989579 on Build GRCh38
rs772562151 Chr.4: 169989711-169989711 on Build GRCh38
rs1052656467 Chr.4: 169989801-169989801 on Build GRCh38
rs1054966280 Chr.4: 169989755-169989755 on Build GRCh38
rs560470327 Chr.4: 169989502-169989502 on Build GRCh38
rs992556645 Chr.4: 169989723-169989723 on Build GRCh38
rs925484828 Chr.4: 169989782-169989782 on Build GRCh38
rs936908341 Chr.4: 169989783-169989783 on Build GRCh38
rs539654620 Chr.4: 169989712-169989712 on Build GRCh38
rs549462969 Chr.4: 169989685-169989685 on Build GRCh38
rs915649876 Chr.4: 169989737-169989737 on Build GRCh38
rs1047486427 Chr.4: 169989422-169989422 on Build GRCh38
rs944041734 Chr.4: 169989813-169989813 on Build GRCh38
rs769523431 Chr.4: 169989535-169989535 on Build GRCh38
rs955741801 Chr.4: 169989591-169989591 on Build GRCh38
rs939207875 Chr.4: 169989720-169989720 on Build GRCh38
rs114444126 Chr.4: 169989542-169989542 on Build GRCh38
rs1020128098 Chr.4: 169989654-169989654 on Build GRCh38
rs920264117 Chr.4: 169989683-169989683 on Build GRCh38
rs895090421 Chr.4: 169989756-169989756 on Build GRCh38
rs948512089 Chr.4: 169989742-169989742 on Build GRCh38
rs902541809 Chr.4: 169989860-169989860 on Build GRCh38
rs928914622 Chr.4: 169989694-169989694 on Build GRCh38
rs140284530 Chr.4: 169989614-169989615 on Build GRCh38
rs539162247 Chr.4: 169989509-169989509 on Build GRCh38

View Full Product Details