Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 34656536-34657041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC105374753
Entrez Gene ID
105374753
Gene Name
uncharacterized LOC105374753
Gene Aliases
-
UniGene
Hs.534795
Gene Chromosome Location
Chr.5: 34656306-34658489 on Build GRCh38
Transcripts
NM_001145520.1 NM_001145522.1 NM_015577.2 XM_011514024.1 XM_017009335.1 XM_017009336.1

Gene Symbol
RAI14
Entrez Gene ID
26064
Gene Name
retinoic acid induced 14
Gene Aliases
NORPEG, RAI13
UniGene
Hs.431400
Gene Chromosome Location
Chr.5: 34656328-34832612 on Build GRCh38
Transcripts
NM_001145520.1 NM_001145522.1 NM_015577.2 XM_011514024.1 XM_017009335.1 XM_017009336.1

SNP Detail

SNP ID Chromosome Location
rs957204108 Chr.5: 34656764-34656764 on Build GRCh38
rs893043427 Chr.5: 34656575-34656575 on Build GRCh38
rs1031110367 Chr.5: 34656980-34656980 on Build GRCh38
rs1017490049 Chr.5: 34656780-34656780 on Build GRCh38
rs75055357 Chr.5: 34656613-34656613 on Build GRCh38
rs1055339896 Chr.5: 34656629-34656629 on Build GRCh38
rs1011390220 Chr.5: 34656594-34656594 on Build GRCh38
rs982677645 Chr.5: 34656979-34656979 on Build GRCh38
rs780724714 Chr.5: 34656751-34656751 on Build GRCh38
rs923953213 Chr.5: 34656770-34656770 on Build GRCh38
rs989472112 Chr.5: 34656987-34656987 on Build GRCh38
rs893891178 Chr.5: 34656953-34656953 on Build GRCh38
rs368054720 Chr.5: 34656806-34656806 on Build GRCh38
rs947296815 Chr.5: 34656843-34656843 on Build GRCh38
rs1032854436 Chr.5: 34656756-34656756 on Build GRCh38
rs997238239 Chr.5: 34656632-34656632 on Build GRCh38
rs531023426 Chr.5: 34656769-34656769 on Build GRCh38
rs902494479 Chr.5: 34656908-34656908 on Build GRCh38
rs933081293 Chr.5: 34656911-34656911 on Build GRCh38
rs991022100 Chr.5: 34656710-34656710 on Build GRCh38
rs540156413 Chr.5: 34656912-34656912 on Build GRCh38
rs1024241340 Chr.5: 34656975-34656975 on Build GRCh38
rs970893713 Chr.5: 34656976-34656976 on Build GRCh38
rs1016554021 Chr.5: 34656653-34656653 on Build GRCh38
rs1015931345 Chr.5: 34656954-34656954 on Build GRCh38
rs971223002 Chr.5: 34656750-34656750 on Build GRCh38
rs922967514 Chr.5: 34656588-34656588 on Build GRCh38
rs975756552 Chr.5: 34656559-34656559 on Build GRCh38
rs900292075 Chr.5: 34656905-34656905 on Build GRCh38
rs1012466026 Chr.5: 34656968-34656968 on Build GRCh38
rs916721284 Chr.5: 34656737-34656737 on Build GRCh38
rs573617524 Chr.5: 34656574-34656574 on Build GRCh38
rs1030411582 Chr.5: 34656637-34656637 on Build GRCh38
rs956786167 Chr.5: 34656981-34656981 on Build GRCh38
rs461660 Chr.5: 34656920-34656920 on Build GRCh38
rs567141378 Chr.5: 34656941-34656941 on Build GRCh38
rs922915216 Chr.5: 34656814-34656814 on Build GRCh38
rs915725577 Chr.5: 34656823-34656823 on Build GRCh38
rs929039005 Chr.5: 34656591-34656591 on Build GRCh38
rs949619756 Chr.5: 34656901-34656901 on Build GRCh38
rs900281183 Chr.5: 34656625-34656625 on Build GRCh38
rs899424465 Chr.5: 34656959-34656959 on Build GRCh38
rs963603704 Chr.5: 34656690-34656690 on Build GRCh38
rs999552862 Chr.5: 34656755-34656755 on Build GRCh38
rs1007012306 Chr.5: 34656967-34656967 on Build GRCh38
rs935376675 Chr.5: 34656783-34656783 on Build GRCh38
rs950390327 Chr.5: 34656641-34656641 on Build GRCh38
rs914531698 Chr.5: 34656813-34656813 on Build GRCh38
rs528293645 Chr.5: 34656868-34656868 on Build GRCh38
rs942175746 Chr.5: 34656887-34656887 on Build GRCh38
rs1013824844 Chr.5: 34656684-34656684 on Build GRCh38
rs369173770 Chr.5: 34656556-34656557 on Build GRCh38
rs912641125 Chr.5: 34656889-34656900 on Build GRCh38
rs562087467 Chr.5: 34656643-34656643 on Build GRCh38
rs916794250 Chr.5: 34656898-34656898 on Build GRCh38
rs950373876 Chr.5: 34656815-34656816 on Build GRCh38
rs1005150002 Chr.5: 34656647-34656647 on Build GRCh38

View Full Product Details