Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 43445378-43445900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM267
Entrez Gene ID
64417
Gene Name
transmembrane protein 267
Gene Aliases
C5orf28
UniGene
Hs.732093
Gene Chromosome Location
Chr.5: 43444252-43484660 on Build GRCh38
Transcripts
NM_022483.4 XM_005248333.4 XM_005248335.3 XM_006714486.3 XM_011514074.2 XM_011514075.2

SNP Detail

SNP ID Chromosome Location
rs540584001 Chr.5: 43445695-43445695 on Build GRCh38
rs960681004 Chr.5: 43445847-43445847 on Build GRCh38
rs1027769671 Chr.5: 43445489-43445489 on Build GRCh38
rs956420703 Chr.5: 43445748-43445748 on Build GRCh38
rs891414248 Chr.5: 43445669-43445669 on Build GRCh38
rs939847642 Chr.5: 43445550-43445550 on Build GRCh38
rs575737470 Chr.5: 43445419-43445419 on Build GRCh38
rs188666581 Chr.5: 43445774-43445774 on Build GRCh38
rs1008880860 Chr.5: 43445701-43445701 on Build GRCh38
rs980471872 Chr.5: 43445811-43445811 on Build GRCh38
rs897826693 Chr.5: 43445479-43445479 on Build GRCh38
rs939246420 Chr.5: 43445516-43445516 on Build GRCh38
rs986322951 Chr.5: 43445526-43445526 on Build GRCh38
rs976340971 Chr.5: 43445626-43445626 on Build GRCh38
rs980399888 Chr.5: 43445451-43445451 on Build GRCh38
rs952217733 Chr.5: 43445523-43445523 on Build GRCh38
rs561325587 Chr.5: 43445488-43445488 on Build GRCh38
rs926528798 Chr.5: 43445473-43445473 on Build GRCh38
rs949846210 Chr.5: 43445865-43445865 on Build GRCh38
rs754112827 Chr.5: 43445773-43445776 on Build GRCh38
rs1031861235 Chr.5: 43445746-43445746 on Build GRCh38
rs544748560 Chr.5: 43445436-43445436 on Build GRCh38
rs898551824 Chr.5: 43445619-43445619 on Build GRCh38
rs760486528 Chr.5: 43445731-43445731 on Build GRCh38
rs926267810 Chr.5: 43445813-43445813 on Build GRCh38
rs910223638 Chr.5: 43445791-43445791 on Build GRCh38
rs1038874722 Chr.5: 43445612-43445612 on Build GRCh38
rs925749351 Chr.5: 43445772-43445772 on Build GRCh38
rs555547049 Chr.5: 43445541-43445541 on Build GRCh38
rs754082637 Chr.5: 43445653-43445653 on Build GRCh38
rs11547957 Chr.5: 43445753-43445753 on Build GRCh38
rs1055440819 Chr.5: 43445784-43445784 on Build GRCh38
rs1022061826 Chr.5: 43445765-43445765 on Build GRCh38
rs996807654 Chr.5: 43445633-43445633 on Build GRCh38
rs967717236 Chr.5: 43445450-43445450 on Build GRCh38
rs908354311 Chr.5: 43445545-43445545 on Build GRCh38
rs1017806104 Chr.5: 43445544-43445544 on Build GRCh38
rs967751523 Chr.5: 43445768-43445768 on Build GRCh38
rs541260040 Chr.5: 43445762-43445762 on Build GRCh38
rs530390230 Chr.5: 43445603-43445603 on Build GRCh38

View Full Product Details