Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55308053-55308542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SKIV2L2
Entrez Gene ID
23517
Gene Name
Ski2 like RNA helicase 2
Gene Aliases
Dob1, KIAA0052, Mtr4, fSAP118
UniGene
Hs.274531
Gene Chromosome Location
Chr.5: 55307748-55425581 on Build GRCh38
Transcripts
NM_015360.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3855610 c.124G>A p.D42N Chr.5: 55308137-55308137 on Build GRCh38
1068849 c.134+1G>A p.? Chr.5: 55308148-55308148 on Build GRCh38
3855609 c.47A>T p.D16V Chr.5: 55308060-55308060 on Build GRCh38
3344130 c.133G>C p.G45R Chr.5: 55308146-55308146 on Build GRCh38
5061118 c.92_93insG p.K33fs*17 Chr.5: 55308105-55308106 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139234738 Chr.5: 55308109-55308109 on Build GRCh38
rs894674601 Chr.5: 55308392-55308392 on Build GRCh38
rs998833580 Chr.5: 55308412-55308412 on Build GRCh38
rs1022137040 Chr.5: 55308404-55308404 on Build GRCh38
rs545864726 Chr.5: 55308473-55308473 on Build GRCh38
rs113691185 Chr.5: 55308246-55308246 on Build GRCh38
rs973138066 Chr.5: 55308512-55308512 on Build GRCh38
rs563438949 Chr.5: 55308091-55308091 on Build GRCh38
rs1056039825 Chr.5: 55308389-55308389 on Build GRCh38
rs769982886 Chr.5: 55308437-55308437 on Build GRCh38
rs34536927 Chr.5: 55308369-55308369 on Build GRCh38
rs374529416 Chr.5: 55308165-55308165 on Build GRCh38
rs767088388 Chr.5: 55308143-55308143 on Build GRCh38
rs953366019 Chr.5: 55308441-55308441 on Build GRCh38
rs753565286 Chr.5: 55308174-55308174 on Build GRCh38
rs920406862 Chr.5: 55308379-55308379 on Build GRCh38
rs1011739401 Chr.5: 55308396-55308396 on Build GRCh38
rs1014789424 Chr.5: 55308256-55308256 on Build GRCh38
rs779865125 Chr.5: 55308444-55308444 on Build GRCh38
rs923864316 Chr.5: 55308285-55308285 on Build GRCh38
rs185438749 Chr.5: 55308429-55308429 on Build GRCh38
rs977360742 Chr.5: 55308283-55308283 on Build GRCh38
rs757302853 Chr.5: 55308418-55308418 on Build GRCh38
rs865830335 Chr.5: 55308220-55308220 on Build GRCh38
rs907472378 Chr.5: 55308374-55308374 on Build GRCh38
rs940345742 Chr.5: 55308365-55308365 on Build GRCh38
rs1029872848 Chr.5: 55308455-55308455 on Build GRCh38
rs899052058 Chr.5: 55308216-55308217 on Build GRCh38
rs1033238879 Chr.5: 55308300-55308300 on Build GRCh38
rs756874353 Chr.5: 55308199-55308199 on Build GRCh38
rs1049322656 Chr.5: 55308322-55308322 on Build GRCh38
rs760501885 Chr.5: 55308159-55308159 on Build GRCh38
rs910321439 Chr.5: 55308501-55308501 on Build GRCh38
rs989062719 Chr.5: 55308315-55308315 on Build GRCh38
rs771959345 Chr.5: 55308240-55308240 on Build GRCh38
rs978868186 Chr.5: 55308294-55308294 on Build GRCh38
rs760397175 Chr.5: 55308255-55308255 on Build GRCh38
rs749016532 Chr.5: 55308151-55308195 on Build GRCh38
rs187566676 Chr.5: 55308341-55308341 on Build GRCh38
rs528498095 Chr.5: 55308177-55308177 on Build GRCh38
rs967451341 Chr.5: 55308407-55308407 on Build GRCh38
rs967818209 Chr.5: 55308270-55308270 on Build GRCh38
rs553621311 Chr.5: 55308391-55308391 on Build GRCh38
rs202230684 Chr.5: 55308095-55308095 on Build GRCh38
rs1003044409 Chr.5: 55308228-55308228 on Build GRCh38
rs113129565 Chr.5: 55308235-55308235 on Build GRCh38
rs760003998 Chr.5: 55308127-55308127 on Build GRCh38
rs771507477 Chr.5: 55308123-55308123 on Build GRCh38
rs752864541 Chr.5: 55308197-55308197 on Build GRCh38
rs541294144 Chr.5: 55308210-55308210 on Build GRCh38
rs542835433 Chr.5: 55308231-55308231 on Build GRCh38
rs755154979 Chr.5: 55308082-55308082 on Build GRCh38
rs748370811 Chr.5: 55308094-55308094 on Build GRCh38
rs530589027 Chr.5: 55308318-55308318 on Build GRCh38
rs750004976 Chr.5: 55308146-55308146 on Build GRCh38
rs148436384 Chr.5: 55308147-55308147 on Build GRCh38
rs376773669 Chr.5: 55308358-55308360 on Build GRCh38
rs913671595 Chr.5: 55308111-55308111 on Build GRCh38
rs575447318 Chr.5: 55308472-55308472 on Build GRCh38
rs958888885 Chr.5: 55308244-55308244 on Build GRCh38
rs569878829 Chr.5: 55308312-55308312 on Build GRCh38
rs778707790 Chr.5: 55308188-55308188 on Build GRCh38
rs943144491 Chr.5: 55308211-55308211 on Build GRCh38
rs796474058 Chr.5: 55308427-55308427 on Build GRCh38
rs761591150 Chr.5: 55308139-55308139 on Build GRCh38
rs568707225 Chr.5: 55308423-55308423 on Build GRCh38
rs2925531 Chr.5: 55308426-55308426 on Build GRCh38
rs111856852 Chr.5: 55308253-55308253 on Build GRCh38
rs113304584 Chr.5: 55308213-55308213 on Build GRCh38
rs146136432 Chr.5: 55308305-55308305 on Build GRCh38
rs377514775 Chr.5: 55308126-55308126 on Build GRCh38
rs987126148 Chr.5: 55308496-55308496 on Build GRCh38
rs774082027 Chr.5: 55308136-55308136 on Build GRCh38
rs765751923 Chr.5: 55308276-55308276 on Build GRCh38
rs371073092 Chr.5: 55308156-55308156 on Build GRCh38
rs145897480 Chr.5: 55308457-55308460 on Build GRCh38
rs375878338 Chr.5: 55308074-55308074 on Build GRCh38
rs772096950 Chr.5: 55308101-55308101 on Build GRCh38
rs965912967 Chr.5: 55308287-55308287 on Build GRCh38
rs760672905 Chr.5: 55308375-55308375 on Build GRCh38
rs564104416 Chr.5: 55308132-55308132 on Build GRCh38
rs751612007 Chr.5: 55308406-55308406 on Build GRCh38
rs190696250 Chr.5: 55308362-55308362 on Build GRCh38
rs956197622 Chr.5: 55308438-55308438 on Build GRCh38
rs753710402 Chr.5: 55308080-55308080 on Build GRCh38
rs1036648369 Chr.5: 55308208-55308208 on Build GRCh38
rs984605580 Chr.5: 55308511-55308511 on Build GRCh38
rs1004548872 Chr.5: 55308200-55308200 on Build GRCh38
rs180972005 Chr.5: 55308370-55308370 on Build GRCh38

View Full Product Details