Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55343049-55343548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SKIV2L2
Entrez Gene ID
23517
Gene Name
Ski2 like RNA helicase 2
Gene Aliases
Dob1, KIAA0052, Mtr4, fSAP118
UniGene
Hs.274531
Gene Chromosome Location
Chr.5: 55307748-55425581 on Build GRCh38
Transcripts
NM_015360.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1068892 c.784C>T p.R262C Chr.5: 55343333-55343333 on Build GRCh38
3920204 c.831C>T p.V277V Chr.5: 55343380-55343380 on Build GRCh38
6779022 c.849G>A p.S283S Chr.5: 55343398-55343398 on Build GRCh38
1068893 c.848C>T p.S283L Chr.5: 55343397-55343397 on Build GRCh38
1193524 c.810T>G p.I270M Chr.5: 55343359-55343359 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1045828810 Chr.5: 55343241-55343241 on Build GRCh38
rs374689113 Chr.5: 55343398-55343398 on Build GRCh38
rs930882236 Chr.5: 55343158-55343158 on Build GRCh38
rs184254099 Chr.5: 55343477-55343477 on Build GRCh38
rs377452171 Chr.5: 55343313-55343313 on Build GRCh38
rs886632369 Chr.5: 55343187-55343187 on Build GRCh38
rs1038677992 Chr.5: 55343126-55343126 on Build GRCh38
rs945140118 Chr.5: 55343075-55343075 on Build GRCh38
rs967712604 Chr.5: 55343201-55343201 on Build GRCh38
rs747521885 Chr.5: 55343418-55343418 on Build GRCh38
rs747494448 Chr.5: 55343302-55343302 on Build GRCh38
rs748297445 Chr.5: 55343296-55343296 on Build GRCh38
rs747776234 Chr.5: 55343479-55343479 on Build GRCh38
rs1021847036 Chr.5: 55343165-55343165 on Build GRCh38
rs576182903 Chr.5: 55343524-55343524 on Build GRCh38
rs760559056 Chr.5: 55343370-55343370 on Build GRCh38
rs746301119 Chr.5: 55343460-55343460 on Build GRCh38
rs536414559 Chr.5: 55343401-55343401 on Build GRCh38
rs572379310 Chr.5: 55343344-55343344 on Build GRCh38
rs777746852 Chr.5: 55343405-55343405 on Build GRCh38
rs956464141 Chr.5: 55343278-55343279 on Build GRCh38
rs1036440162 Chr.5: 55343074-55343074 on Build GRCh38
rs943053142 Chr.5: 55343099-55343099 on Build GRCh38
rs552522470 Chr.5: 55343078-55343078 on Build GRCh38
rs758458777 Chr.5: 55343400-55343400 on Build GRCh38
rs755192954 Chr.5: 55343392-55343392 on Build GRCh38
rs554823334 Chr.5: 55343431-55343431 on Build GRCh38
rs757752993 Chr.5: 55343430-55343430 on Build GRCh38
rs745590425 Chr.5: 55343315-55343318 on Build GRCh38
rs1035850564 Chr.5: 55343088-55343088 on Build GRCh38
rs757981606 Chr.5: 55343320-55343321 on Build GRCh38
rs770925800 Chr.5: 55343498-55343498 on Build GRCh38
rs770287451 Chr.5: 55343474-55343474 on Build GRCh38
rs899418753 Chr.5: 55343133-55343133 on Build GRCh38
rs766948286 Chr.5: 55343355-55343355 on Build GRCh38
rs938792669 Chr.5: 55343521-55343521 on Build GRCh38
rs777177165 Chr.5: 55343312-55343312 on Build GRCh38
rs979084099 Chr.5: 55343247-55343247 on Build GRCh38
rs774473685 Chr.5: 55343300-55343300 on Build GRCh38
rs201981107 Chr.5: 55343502-55343502 on Build GRCh38
rs538919904 Chr.5: 55343213-55343213 on Build GRCh38
rs147039795 Chr.5: 55343377-55343377 on Build GRCh38
rs768480581 Chr.5: 55343316-55343316 on Build GRCh38
rs199907263 Chr.5: 55343359-55343359 on Build GRCh38
rs1014361082 Chr.5: 55343227-55343227 on Build GRCh38
rs760633993 Chr.5: 55343495-55343495 on Build GRCh38
rs373463657 Chr.5: 55343303-55343303 on Build GRCh38
rs761318485 Chr.5: 55343350-55343350 on Build GRCh38
rs758321459 Chr.5: 55343521-55343524 on Build GRCh38
rs140748362 Chr.5: 55343076-55343076 on Build GRCh38
rs553736439 Chr.5: 55343267-55343267 on Build GRCh38
rs772635923 Chr.5: 55343490-55343490 on Build GRCh38
rs774086585 Chr.5: 55343475-55343475 on Build GRCh38
rs570796665 Chr.5: 55343163-55343163 on Build GRCh38
rs758838224 Chr.5: 55343095-55343095 on Build GRCh38
rs377597877 Chr.5: 55343486-55343486 on Build GRCh38

View Full Product Details