Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 61158753-61159253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM15
Entrez Gene ID
643155
Gene Name
small integral membrane protein 15
Gene Aliases
C5orf43
UniGene
Hs.508479
Gene Chromosome Location
Chr.5: 61157709-61162475 on Build GRCh38
Transcripts
NM_001048249.3

SNP Detail

SNP ID Chromosome Location
rs76253290 Chr.5: 61158799-61158799 on Build GRCh38
rs530880532 Chr.5: 61159003-61159003 on Build GRCh38
rs140536797 Chr.5: 61158805-61158805 on Build GRCh38
rs900529519 Chr.5: 61159216-61159216 on Build GRCh38
rs983571203 Chr.5: 61159025-61159025 on Build GRCh38
rs531765423 Chr.5: 61159070-61159070 on Build GRCh38
rs920159829 Chr.5: 61159076-61159077 on Build GRCh38
rs949584330 Chr.5: 61159102-61159102 on Build GRCh38
rs72759240 Chr.5: 61159103-61159103 on Build GRCh38
rs1021049991 Chr.5: 61158865-61158865 on Build GRCh38
rs963036952 Chr.5: 61159075-61159075 on Build GRCh38
rs549433993 Chr.5: 61159132-61159132 on Build GRCh38
rs896547811 Chr.5: 61158829-61158829 on Build GRCh38
rs74917535 Chr.5: 61159004-61159004 on Build GRCh38
rs1014062361 Chr.5: 61158854-61158854 on Build GRCh38
rs144862587 Chr.5: 61158938-61158938 on Build GRCh38
rs918048367 Chr.5: 61159091-61159091 on Build GRCh38
rs551961107 Chr.5: 61159076-61159076 on Build GRCh38
rs180768088 Chr.5: 61158973-61158973 on Build GRCh38
rs540198600 Chr.5: 61158909-61158909 on Build GRCh38
rs1038591429 Chr.5: 61158779-61158779 on Build GRCh38
rs765146867 Chr.5: 61159018-61159018 on Build GRCh38
rs907626363 Chr.5: 61159060-61159064 on Build GRCh38
rs1053739104 Chr.5: 61159201-61159201 on Build GRCh38
rs755969211 Chr.5: 61158976-61158976 on Build GRCh38
rs938284775 Chr.5: 61159083-61159083 on Build GRCh38
rs1055727810 Chr.5: 61158808-61158808 on Build GRCh38
rs1000810771 Chr.5: 61159106-61159106 on Build GRCh38
rs898787490 Chr.5: 61158781-61158781 on Build GRCh38
rs974584565 Chr.5: 61158998-61158998 on Build GRCh38
rs1000862998 Chr.5: 61158945-61158945 on Build GRCh38
rs762997000 Chr.5: 61159126-61159126 on Build GRCh38
rs775831887 Chr.5: 61158889-61158889 on Build GRCh38
rs1032214726 Chr.5: 61158956-61158956 on Build GRCh38
rs753312568 Chr.5: 61159228-61159228 on Build GRCh38
rs1055397270 Chr.5: 61159086-61159086 on Build GRCh38
rs902642837 Chr.5: 61159105-61159105 on Build GRCh38

View Full Product Details