Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80650470-80650977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHFR
Entrez Gene ID
1719
Gene Name
dihydrofolate reductase
Gene Aliases
DHFRP1, DYR
UniGene
Hs.592364
Gene Chromosome Location
Chr.5: 80626226-80654981 on Build GRCh38
Transcripts
NM_000791.3 NM_001290354.1 NM_001290357.1 NR_110936.1 NM_001190470.1

Gene Symbol
MTRNR2L2
Entrez Gene ID
100462981
Gene Name
MT-RNR2-like 2
Gene Aliases
HN2
UniGene
Hs.666077
Gene Chromosome Location
Chr.5: 80650000-80651035 on Build GRCh38
Transcripts
NM_000791.3 NM_001290354.1 NM_001290357.1 NR_110936.1 NM_001190470.1

SNP Detail

SNP ID Chromosome Location
rs537353690 Chr.5: 80650728-80650728 on Build GRCh38
rs3991268 Chr.5: 80650713-80650713 on Build GRCh38
rs941722714 Chr.5: 80650931-80650931 on Build GRCh38
rs761815562 Chr.5: 80650863-80650863 on Build GRCh38
rs3958541 Chr.5: 80650662-80650662 on Build GRCh38
rs71328893 Chr.5: 80650818-80650818 on Build GRCh38
rs972528712 Chr.5: 80650510-80650510 on Build GRCh38
rs553534345 Chr.5: 80650791-80650791 on Build GRCh38
rs910189223 Chr.5: 80650921-80650921 on Build GRCh38
rs3958542 Chr.5: 80650741-80650741 on Build GRCh38
rs374520236 Chr.5: 80650951-80650952 on Build GRCh38
rs901617871 Chr.5: 80650652-80650652 on Build GRCh38
rs967600662 Chr.5: 80650813-80650813 on Build GRCh38
rs970818386 Chr.5: 80650765-80650765 on Build GRCh38
rs776446720 Chr.5: 80650734-80650734 on Build GRCh38
rs3958540 Chr.5: 80650638-80650638 on Build GRCh38
rs546929803 Chr.5: 80650951-80650951 on Build GRCh38
rs1050122946 Chr.5: 80650668-80650668 on Build GRCh38
rs1041837216 Chr.5: 80650621-80650621 on Build GRCh38
rs560727756 Chr.5: 80650515-80650515 on Build GRCh38
rs544457992 Chr.5: 80650508-80650508 on Build GRCh38
rs836820 Chr.5: 80650561-80650561 on Build GRCh38
rs932959904 Chr.5: 80650657-80650657 on Build GRCh38
rs74464984 Chr.5: 80650575-80650575 on Build GRCh38
rs567568145 Chr.5: 80650745-80650745 on Build GRCh38
rs761920985 Chr.5: 80650550-80650550 on Build GRCh38
rs559798398 Chr.5: 80650607-80650607 on Build GRCh38
rs914515760 Chr.5: 80650809-80650809 on Build GRCh38
rs1016529111 Chr.5: 80650731-80650731 on Build GRCh38
rs551555903 Chr.5: 80650669-80650669 on Build GRCh38
rs937297498 Chr.5: 80650576-80650576 on Build GRCh38
rs71628868 Chr.5: 80650894-80650894 on Build GRCh38
rs933097102 Chr.5: 80650862-80650862 on Build GRCh38
rs77008859 Chr.5: 80650499-80650499 on Build GRCh38
rs183768463 Chr.5: 80650752-80650752 on Build GRCh38
rs764257446 Chr.5: 80650775-80650775 on Build GRCh38
rs558205606 Chr.5: 80650582-80650583 on Build GRCh38
rs79627144 Chr.5: 80650556-80650556 on Build GRCh38
rs71328894 Chr.5: 80650829-80650829 on Build GRCh38
rs774425738 Chr.5: 80650810-80650810 on Build GRCh38
rs528686734 Chr.5: 80650641-80650641 on Build GRCh38
rs2405873 Chr.5: 80650835-80650835 on Build GRCh38
rs958040567 Chr.5: 80650802-80650802 on Build GRCh38
rs112558161 Chr.5: 80650647-80650647 on Build GRCh38
rs878881803 Chr.5: 80650914-80650914 on Build GRCh38
rs548625287 Chr.5: 80650554-80650554 on Build GRCh38
rs970684087 Chr.5: 80650516-80650516 on Build GRCh38
rs372335365 Chr.5: 80650497-80650497 on Build GRCh38
rs945916530 Chr.5: 80650596-80650596 on Build GRCh38
rs371631948 Chr.5: 80650935-80650935 on Build GRCh38
rs990087671 Chr.5: 80650579-80650579 on Build GRCh38
rs879207460 Chr.5: 80650938-80650938 on Build GRCh38
rs1046585021 Chr.5: 80650948-80650948 on Build GRCh38
rs770705037 Chr.5: 80650729-80650729 on Build GRCh38
rs368360907 Chr.5: 80650740-80650740 on Build GRCh38
rs1033599666 Chr.5: 80650783-80650783 on Build GRCh38
rs878991233 Chr.5: 80650830-80650830 on Build GRCh38
rs3991269 Chr.5: 80650774-80650774 on Build GRCh38
rs536633066 Chr.5: 80650770-80650770 on Build GRCh38
rs369735282 Chr.5: 80650496-80650496 on Build GRCh38
rs571947725 Chr.5: 80650717-80650717 on Build GRCh38
rs879073578 Chr.5: 80650907-80650907 on Build GRCh38
rs2897261 Chr.5: 80650876-80650876 on Build GRCh38
rs538879458 Chr.5: 80650878-80650878 on Build GRCh38
rs990446657 Chr.5: 80650808-80650808 on Build GRCh38
rs923008353 Chr.5: 80650840-80650840 on Build GRCh38

View Full Product Details