Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 112891883-112892420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REEP5
Entrez Gene ID
7905
Gene Name
receptor accessory protein 5
Gene Aliases
C5orf18, D5S346, DP1, TB2, YOP1, Yip2e
UniGene
Hs.429608
Gene Chromosome Location
Chr.5: 112876384-112922334 on Build GRCh38
Transcripts
NM_005669.4 XM_017009844.1 NM_001204199.1

Gene Symbol
SRP19
Entrez Gene ID
6728
Gene Name
signal recognition particle 19
Gene Aliases
-
UniGene
Hs.637001
Gene Chromosome Location
Chr.5: 112861188-112893079 on Build GRCh38
Transcripts
NM_005669.4 XM_017009844.1 NM_001204199.1

Gene Symbol
ZRSR1
Entrez Gene ID
7310
Gene Name
zinc finger CCCH-type, RNA binding motif and serine/arginine rich 1
Gene Aliases
U2AF1-RS1, U2AF1L1, U2AF1P, U2AF1RS1, U2AFBPL, ZC3H21
UniGene
Hs.637001
Gene Chromosome Location
Chr.5: 112891616-112893097 on Build GRCh38
Transcripts
NM_005669.4 XM_017009844.1 NM_001204199.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6038835 c.421G>A p.V141M Chr.5: 112892060-112892060 on Build GRCh38
7217870 c.456A>G p.L152L Chr.5: 112892095-112892095 on Build GRCh38
7242236 c.616C>T p.L206F Chr.5: 112892255-112892255 on Build GRCh38
6050491 c.255G>A p.Q85Q Chr.5: 112891894-112891894 on Build GRCh38
149898 c.603T>C p.S201S Chr.5: 112892242-112892242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757920678 Chr.5: 112892034-112892036 on Build GRCh38
rs751804570 Chr.5: 112891936-112891938 on Build GRCh38
rs747673529 Chr.5: 112892220-112892220 on Build GRCh38
rs758469884 Chr.5: 112892087-112892087 on Build GRCh38
rs751536325 Chr.5: 112892057-112892057 on Build GRCh38
rs985272280 Chr.5: 112892174-112892174 on Build GRCh38
rs773937476 Chr.5: 112892152-112892152 on Build GRCh38
rs761442929 Chr.5: 112892153-112892153 on Build GRCh38
rs770588712 Chr.5: 112892103-112892104 on Build GRCh38
rs538839279 Chr.5: 112892002-112892002 on Build GRCh38
rs1032053607 Chr.5: 112892156-112892156 on Build GRCh38
rs550072734 Chr.5: 112891938-112891938 on Build GRCh38
rs750379852 Chr.5: 112892159-112892159 on Build GRCh38
rs756493903 Chr.5: 112892197-112892197 on Build GRCh38
rs1013032986 Chr.5: 112892090-112892090 on Build GRCh38
rs916321921 Chr.5: 112892274-112892274 on Build GRCh38
rs745651207 Chr.5: 112891955-112891955 on Build GRCh38
rs764810500 Chr.5: 112891916-112891916 on Build GRCh38
rs772191308 Chr.5: 112891968-112891968 on Build GRCh38
rs779357335 Chr.5: 112892366-112892366 on Build GRCh38
rs781103889 Chr.5: 112891949-112891949 on Build GRCh38
rs755182345 Chr.5: 112891958-112891958 on Build GRCh38
rs534068387 Chr.5: 112892127-112892127 on Build GRCh38
rs754358702 Chr.5: 112892015-112892017 on Build GRCh38
rs371589323 Chr.5: 112892169-112892169 on Build GRCh38
rs756517401 Chr.5: 112892286-112892286 on Build GRCh38
rs759302583 Chr.5: 112892316-112892316 on Build GRCh38
rs745678178 Chr.5: 112892256-112892257 on Build GRCh38
rs768370547 Chr.5: 112892215-112892215 on Build GRCh38
rs572159454 Chr.5: 112892271-112892271 on Build GRCh38
rs712666 Chr.5: 112892242-112892242 on Build GRCh38
rs753636327 Chr.5: 112892166-112892166 on Build GRCh38
rs758011778 Chr.5: 112891937-112891937 on Build GRCh38
rs766973731 Chr.5: 112892342-112892342 on Build GRCh38
rs549770313 Chr.5: 112892357-112892357 on Build GRCh38
rs116824915 Chr.5: 112891939-112891939 on Build GRCh38
rs141868335 Chr.5: 112892222-112892222 on Build GRCh38
rs191227399 Chr.5: 112892334-112892334 on Build GRCh38
rs761870820 Chr.5: 112892321-112892321 on Build GRCh38
rs772720429 Chr.5: 112892017-112892017 on Build GRCh38
rs748197210 Chr.5: 112891965-112891965 on Build GRCh38
rs766355052 Chr.5: 112892249-112892249 on Build GRCh38
rs767462791 Chr.5: 112892158-112892158 on Build GRCh38
rs764268042 Chr.5: 112892265-112892265 on Build GRCh38
rs778699193 Chr.5: 112892292-112892292 on Build GRCh38
rs746419233 Chr.5: 112892180-112892180 on Build GRCh38
rs61741889 Chr.5: 112892300-112892300 on Build GRCh38
rs777304934 Chr.5: 112892039-112892042 on Build GRCh38
rs776362524 Chr.5: 112892261-112892261 on Build GRCh38
rs770817305 Chr.5: 112892307-112892307 on Build GRCh38
rs183342096 Chr.5: 112892390-112892390 on Build GRCh38
rs781707380 Chr.5: 112892171-112892171 on Build GRCh38
rs777764337 Chr.5: 112892095-112892095 on Build GRCh38
rs761771394 Chr.5: 112892065-112892065 on Build GRCh38
rs759839764 Chr.5: 112892021-112892021 on Build GRCh38
rs377229917 Chr.5: 112892131-112892131 on Build GRCh38
rs1036674405 Chr.5: 112891996-112891996 on Build GRCh38
rs751827938 Chr.5: 112892266-112892266 on Build GRCh38
rs560842182 Chr.5: 112892305-112892305 on Build GRCh38
rs746641681 Chr.5: 112892102-112892102 on Build GRCh38
rs965261027 Chr.5: 112892214-112892214 on Build GRCh38
rs767945021 Chr.5: 112891962-112891963 on Build GRCh38
rs930992041 Chr.5: 112892240-112892240 on Build GRCh38
rs546611568 Chr.5: 112892278-112892278 on Build GRCh38
rs531248504 Chr.5: 112892101-112892101 on Build GRCh38
rs550786485 Chr.5: 112892320-112892320 on Build GRCh38
rs551539844 Chr.5: 112892070-112892070 on Build GRCh38
rs750418854 Chr.5: 112892336-112892336 on Build GRCh38
rs778241711 Chr.5: 112891992-112891992 on Build GRCh38
rs750517346 Chr.5: 112892285-112892285 on Build GRCh38
rs578080478 Chr.5: 112892191-112892191 on Build GRCh38
rs753119613 Chr.5: 112892384-112892384 on Build GRCh38
rs755907382 Chr.5: 112892072-112892072 on Build GRCh38
rs771432824 Chr.5: 112892005-112892005 on Build GRCh38
rs780922415 Chr.5: 112892256-112892258 on Build GRCh38
rs755394365 Chr.5: 112892360-112892360 on Build GRCh38
rs749681827 Chr.5: 112892213-112892213 on Build GRCh38
rs750831767 Chr.5: 112892005-112892008 on Build GRCh38
rs762529349 Chr.5: 112892055-112892055 on Build GRCh38
rs535521257 Chr.5: 112892301-112892301 on Build GRCh38
rs538127313 Chr.5: 112892163-112892163 on Build GRCh38
rs771419377 Chr.5: 112892111-112892111 on Build GRCh38
rs768423518 Chr.5: 112892148-112892148 on Build GRCh38
rs1022151456 Chr.5: 112892338-112892338 on Build GRCh38
rs779176035 Chr.5: 112892075-112892075 on Build GRCh38
rs776143293 Chr.5: 112891907-112891907 on Build GRCh38
rs775043127 Chr.5: 112892053-112892053 on Build GRCh38
rs199946572 Chr.5: 112892297-112892297 on Build GRCh38
rs918209061 Chr.5: 112892230-112892230 on Build GRCh38
rs765074322 Chr.5: 112892318-112892318 on Build GRCh38
rs777514138 Chr.5: 112892387-112892387 on Build GRCh38
rs752201917 Chr.5: 112891934-112891934 on Build GRCh38
rs573726695 Chr.5: 112892140-112892140 on Build GRCh38
rs752976701 Chr.5: 112892081-112892081 on Build GRCh38
rs769447970 Chr.5: 112892051-112892051 on Build GRCh38
rs756246625 Chr.5: 112892339-112892339 on Build GRCh38
rs746209964 Chr.5: 112892121-112892121 on Build GRCh38
rs757102297 Chr.5: 112891946-112891946 on Build GRCh38
rs764339826 Chr.5: 112891931-112891932 on Build GRCh38
rs771543724 Chr.5: 112892226-112892226 on Build GRCh38
rs763752661 Chr.5: 112892056-112892056 on Build GRCh38
rs762179897 Chr.5: 112891954-112891956 on Build GRCh38
rs712665 Chr.5: 112892102-112892102 on Build GRCh38
rs780625800 Chr.5: 112892009-112892013 on Build GRCh38
rs767365374 Chr.5: 112892069-112892069 on Build GRCh38
rs763805816 Chr.5: 112892349-112892349 on Build GRCh38
rs545585712 Chr.5: 112892219-112892219 on Build GRCh38
rs563712600 Chr.5: 112892005-112892006 on Build GRCh38
rs780495653 Chr.5: 112892209-112892209 on Build GRCh38
rs757940873 Chr.5: 112892385-112892385 on Build GRCh38
rs781580621 Chr.5: 112892112-112892112 on Build GRCh38
rs755445568 Chr.5: 112892290-112892290 on Build GRCh38
rs61995948 Chr.5: 112892167-112892167 on Build GRCh38

View Full Product Details