Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 113025269-113025793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCC
Entrez Gene ID
4163
Gene Name
mutated in colorectal cancers
Gene Aliases
MCC1
UniGene
Hs.593171
Gene Chromosome Location
Chr.5: 113022099-113488830 on Build GRCh38
Transcripts
NM_001085377.1 NM_002387.2 XM_005271991.2 XM_017009473.1 XM_017009474.1

SNP Detail

SNP ID Chromosome Location
rs981063350 Chr.5: 113025314-113025314 on Build GRCh38
rs777187397 Chr.5: 113025686-113025686 on Build GRCh38
rs897665882 Chr.5: 113025609-113025609 on Build GRCh38
rs569433530 Chr.5: 113025587-113025587 on Build GRCh38
rs755292364 Chr.5: 113025495-113025495 on Build GRCh38
rs139897552 Chr.5: 113025311-113025311 on Build GRCh38
rs10660020 Chr.5: 113025575-113025576 on Build GRCh38
rs528864584 Chr.5: 113025398-113025398 on Build GRCh38
rs1021395355 Chr.5: 113025364-113025364 on Build GRCh38
rs557637826 Chr.5: 113025549-113025549 on Build GRCh38
rs35690462 Chr.5: 113025609-113025610 on Build GRCh38
rs1044173249 Chr.5: 113025350-113025350 on Build GRCh38
rs528592883 Chr.5: 113025335-113025335 on Build GRCh38
rs201055332 Chr.5: 113025573-113025574 on Build GRCh38
rs58750504 Chr.5: 113025610-113025611 on Build GRCh38
rs535774136 Chr.5: 113025519-113025519 on Build GRCh38
rs950330532 Chr.5: 113025440-113025440 on Build GRCh38
rs180784494 Chr.5: 113025673-113025673 on Build GRCh38
rs1030556565 Chr.5: 113025718-113025718 on Build GRCh38
rs1035318002 Chr.5: 113025319-113025319 on Build GRCh38
rs748940682 Chr.5: 113025687-113025687 on Build GRCh38
rs909279555 Chr.5: 113025553-113025553 on Build GRCh38
rs778979968 Chr.5: 113025618-113025618 on Build GRCh38
rs557859355 Chr.5: 113025630-113025630 on Build GRCh38
rs999052318 Chr.5: 113025678-113025678 on Build GRCh38
rs546665030 Chr.5: 113025390-113025390 on Build GRCh38
rs1036189371 Chr.5: 113025607-113025607 on Build GRCh38
rs555058055 Chr.5: 113025724-113025724 on Build GRCh38
rs189775839 Chr.5: 113025326-113025326 on Build GRCh38
rs924139790 Chr.5: 113025423-113025423 on Build GRCh38
rs781157838 Chr.5: 113025357-113025357 on Build GRCh38
rs903703741 Chr.5: 113025367-113025367 on Build GRCh38
rs1021739804 Chr.5: 113025671-113025671 on Build GRCh38
rs945465921 Chr.5: 113025347-113025347 on Build GRCh38
rs999891893 Chr.5: 113025585-113025585 on Build GRCh38
rs938546880 Chr.5: 113025611-113025611 on Build GRCh38
rs1038979830 Chr.5: 113025403-113025403 on Build GRCh38
rs931995186 Chr.5: 113025521-113025521 on Build GRCh38
rs375443387 Chr.5: 113025574-113025575 on Build GRCh38
rs909237669 Chr.5: 113025730-113025730 on Build GRCh38
rs78255454 Chr.5: 113025617-113025617 on Build GRCh38
rs9326871 Chr.5: 113025592-113025593 on Build GRCh38
rs33949689 Chr.5: 113025576-113025577 on Build GRCh38
rs972566594 Chr.5: 113025763-113025763 on Build GRCh38
rs1031128640 Chr.5: 113025682-113025682 on Build GRCh38
rs7709534 Chr.5: 113025422-113025422 on Build GRCh38
rs1025990084 Chr.5: 113025426-113025426 on Build GRCh38
rs942138392 Chr.5: 113025560-113025560 on Build GRCh38
rs923239237 Chr.5: 113025512-113025512 on Build GRCh38
rs986596914 Chr.5: 113025542-113025542 on Build GRCh38
rs548747305 Chr.5: 113025618-113025621 on Build GRCh38
rs574641158 Chr.5: 113025329-113025330 on Build GRCh38
rs941495146 Chr.5: 113025302-113025302 on Build GRCh38
rs563628023 Chr.5: 113025593-113025593 on Build GRCh38
rs573625144 Chr.5: 113025737-113025737 on Build GRCh38
rs1016402487 Chr.5: 113025478-113025478 on Build GRCh38
rs976274329 Chr.5: 113025706-113025706 on Build GRCh38
rs956949007 Chr.5: 113025469-113025469 on Build GRCh38
rs962931578 Chr.5: 113025614-113025614 on Build GRCh38
rs901755592 Chr.5: 113025417-113025417 on Build GRCh38
rs143503170 Chr.5: 113025328-113025328 on Build GRCh38
rs6869100 Chr.5: 113025634-113025634 on Build GRCh38
rs953653619 Chr.5: 113025720-113025720 on Build GRCh38

View Full Product Details