Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 159161462-159161958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF145
Entrez Gene ID
153830
Gene Name
ring finger protein 145
Gene Aliases
-
UniGene
Hs.349306
Gene Chromosome Location
Chr.5: 159157409-159210053 on Build GRCh38
Transcripts
NM_001199380.1 NM_001199381.1 NM_001199382.1 NM_001199383.1 NM_144726.2 XM_017009138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6395752 c.1376A>T p.Y459F Chr.5: 159161600-159161600 on Build GRCh38
4943308 c.1495G>A p.A499T Chr.5: 159161481-159161481 on Build GRCh38
6389857 c.1378G>T p.V460F Chr.5: 159161598-159161598 on Build GRCh38
3853261 c.1394A>C p.E465A Chr.5: 159161582-159161582 on Build GRCh38
6856401 c.1354-7_1354-4delTTTT p.? Chr.5: 159161626-159161629 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763336053 Chr.5: 159161575-159161575 on Build GRCh38
rs1001308236 Chr.5: 159161826-159161826 on Build GRCh38
rs753180463 Chr.5: 159161614-159161614 on Build GRCh38
rs1021482112 Chr.5: 159161744-159161744 on Build GRCh38
rs189954648 Chr.5: 159161819-159161819 on Build GRCh38
rs567969991 Chr.5: 159161735-159161735 on Build GRCh38
rs766786617 Chr.5: 159161581-159161581 on Build GRCh38
rs753482431 Chr.5: 159161641-159161641 on Build GRCh38
rs763652016 Chr.5: 159161636-159161636 on Build GRCh38
rs533689653 Chr.5: 159161641-159161641 on Build GRCh38
rs76548644 Chr.5: 159161630-159161630 on Build GRCh38
rs974854861 Chr.5: 159161877-159161877 on Build GRCh38
rs552772641 Chr.5: 159161668-159161668 on Build GRCh38
rs1054020398 Chr.5: 159161836-159161836 on Build GRCh38
rs17056583 Chr.5: 159161741-159161741 on Build GRCh38
rs750265237 Chr.5: 159161660-159161660 on Build GRCh38
rs532972199 Chr.5: 159161840-159161840 on Build GRCh38
rs1020196705 Chr.5: 159161862-159161862 on Build GRCh38
rs932277962 Chr.5: 159161855-159161855 on Build GRCh38
rs908041391 Chr.5: 159161687-159161687 on Build GRCh38
rs757032286 Chr.5: 159161497-159161497 on Build GRCh38
rs370946043 Chr.5: 159161611-159161611 on Build GRCh38
rs780382744 Chr.5: 159161625-159161629 on Build GRCh38
rs188056900 Chr.5: 159161915-159161915 on Build GRCh38
rs140347447 Chr.5: 159161889-159161889 on Build GRCh38
rs987565868 Chr.5: 159161563-159161563 on Build GRCh38
rs758167832 Chr.5: 159161661-159161661 on Build GRCh38
rs796866742 Chr.5: 159161907-159161907 on Build GRCh38
rs772627933 Chr.5: 159161625-159161626 on Build GRCh38
rs897879499 Chr.5: 159161864-159161864 on Build GRCh38
rs770716803 Chr.5: 159161626-159161627 on Build GRCh38
rs374310644 Chr.5: 159161646-159161646 on Build GRCh38
rs747630902 Chr.5: 159161626-159161631 on Build GRCh38
rs367907107 Chr.5: 159161528-159161528 on Build GRCh38
rs201485871 Chr.5: 159161501-159161501 on Build GRCh38
rs371278772 Chr.5: 159161485-159161485 on Build GRCh38
rs758277636 Chr.5: 159161627-159161631 on Build GRCh38
rs201369350 Chr.5: 159161493-159161493 on Build GRCh38
rs778833400 Chr.5: 159161516-159161516 on Build GRCh38
rs540581579 Chr.5: 159161595-159161595 on Build GRCh38
rs544289245 Chr.5: 159161911-159161911 on Build GRCh38
rs892832551 Chr.5: 159161842-159161842 on Build GRCh38
rs760179941 Chr.5: 159161626-159161629 on Build GRCh38
rs569010291 Chr.5: 159161678-159161678 on Build GRCh38
rs1020060984 Chr.5: 159161490-159161490 on Build GRCh38
rs1057108436 Chr.5: 159161869-159161869 on Build GRCh38
rs532569671 Chr.5: 159161718-159161718 on Build GRCh38
rs532619291 Chr.5: 159161667-159161667 on Build GRCh38
rs773559436 Chr.5: 159161557-159161557 on Build GRCh38
rs759909130 Chr.5: 159161602-159161602 on Build GRCh38
rs1021952436 Chr.5: 159161876-159161876 on Build GRCh38
rs965129441 Chr.5: 159161874-159161874 on Build GRCh38
rs530723654 Chr.5: 159161863-159161863 on Build GRCh38
rs780019659 Chr.5: 159161662-159161662 on Build GRCh38
rs530466069 Chr.5: 159161585-159161585 on Build GRCh38
rs968525577 Chr.5: 159161757-159161757 on Build GRCh38
rs983698879 Chr.5: 159161892-159161892 on Build GRCh38
rs753517327 Chr.5: 159161719-159161719 on Build GRCh38
rs930870324 Chr.5: 159161648-159161648 on Build GRCh38
rs986777646 Chr.5: 159161857-159161858 on Build GRCh38
rs374061231 Chr.5: 159161491-159161491 on Build GRCh38
rs563283556 Chr.5: 159161767-159161767 on Build GRCh38
rs748229151 Chr.5: 159161542-159161542 on Build GRCh38
rs200947571 Chr.5: 159161510-159161510 on Build GRCh38
rs908083911 Chr.5: 159161895-159161895 on Build GRCh38
rs950330660 Chr.5: 159161904-159161904 on Build GRCh38
rs531415099 Chr.5: 159161695-159161695 on Build GRCh38
rs548722417 Chr.5: 159161867-159161867 on Build GRCh38
rs952067356 Chr.5: 159161891-159161891 on Build GRCh38
rs1010089814 Chr.5: 159161849-159161849 on Build GRCh38
rs553801809 Chr.5: 159161757-159161757 on Build GRCh38
rs773623177 Chr.5: 159161636-159161641 on Build GRCh38
rs752240857 Chr.5: 159161737-159161737 on Build GRCh38
rs565121121 Chr.5: 159161645-159161645 on Build GRCh38
rs59997682 Chr.5: 159161636-159161640 on Build GRCh38
rs909421255 Chr.5: 159161857-159161857 on Build GRCh38
rs769174313 Chr.5: 159161626-159161626 on Build GRCh38
rs754715187 Chr.5: 159161724-159161724 on Build GRCh38
rs977201772 Chr.5: 159161777-159161777 on Build GRCh38
rs1048033967 Chr.5: 159161651-159161651 on Build GRCh38
rs965043497 Chr.5: 159161830-159161830 on Build GRCh38
rs183315606 Chr.5: 159161870-159161870 on Build GRCh38
rs745724719 Chr.5: 159161518-159161518 on Build GRCh38
rs764862565 Chr.5: 159161727-159161727 on Build GRCh38
rs548256172 Chr.5: 159161725-159161725 on Build GRCh38
rs988702595 Chr.5: 159161489-159161489 on Build GRCh38
rs139682876 Chr.5: 159161626-159161630 on Build GRCh38
rs927495262 Chr.5: 159161910-159161910 on Build GRCh38

View Full Product Details